Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 878271-878766 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRD9
Entrez Gene ID
65980
Gene Name
bromodomain containing 9
Gene Aliases
LAVS3040, PRO9856
UniGene
Hs.449278
Gene Chromosome Location
Chr.5: 863735-892824 on Build GRCh38
Transcripts
NM_001009877.2 NM_001317951.1 NM_023924.4 NR_134296.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4160046 c.1190T>C p.V397A Chr.5: 878436-878436 on Build GRCh38
4416425 c.1243G>A p.D415N Chr.5: 878383-878383 on Build GRCh38
3140173 c.981G>A p.L327L Chr.5: 878357-878357 on Build GRCh38
4425892 c.1248G>C p.E416D Chr.5: 878378-878378 on Build GRCh38
3140175 c.966C>T p.G322G Chr.5: 878372-878372 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766846882 Chr.5: 878305-878305 on Build GRCh38
rs112921031 Chr.5: 878345-878345 on Build GRCh38
rs752478693 Chr.5: 878500-878500 on Build GRCh38
rs776335129 Chr.5: 878483-878483 on Build GRCh38
rs750542254 Chr.5: 878522-878522 on Build GRCh38
rs758370497 Chr.5: 878449-878449 on Build GRCh38
rs752361048 Chr.5: 878351-878351 on Build GRCh38
rs145107515 Chr.5: 878436-878436 on Build GRCh38
rs112329685 Chr.5: 878708-878708 on Build GRCh38
rs752346504 Chr.5: 878402-878402 on Build GRCh38
rs755685034 Chr.5: 878353-878353 on Build GRCh38
rs769507523 Chr.5: 878539-878539 on Build GRCh38
rs750386081 Chr.5: 878446-878446 on Build GRCh38
rs568142311 Chr.5: 878463-878463 on Build GRCh38
rs150581830 Chr.5: 878459-878459 on Build GRCh38
rs76774211 Chr.5: 878428-878428 on Build GRCh38
rs767405019 Chr.5: 878499-878499 on Build GRCh38
rs778090180 Chr.5: 878420-878420 on Build GRCh38
rs541919388 Chr.5: 878390-878390 on Build GRCh38
rs200748824 Chr.5: 878388-878388 on Build GRCh38
rs768451401 Chr.5: 878305-878306 on Build GRCh38
rs777247838 Chr.5: 878355-878355 on Build GRCh38
rs557158947 Chr.5: 878462-878462 on Build GRCh38
rs199521397 Chr.5: 878478-878478 on Build GRCh38
rs375460308 Chr.5: 878497-878497 on Build GRCh38
rs368074125 Chr.5: 878387-878387 on Build GRCh38
rs768794422 Chr.5: 878331-878331 on Build GRCh38
rs550624553 Chr.5: 878501-878501 on Build GRCh38
rs750344995 Chr.5: 878414-878414 on Build GRCh38
rs141776263 Chr.5: 878454-878454 on Build GRCh38
rs77993339 Chr.5: 878492-878492 on Build GRCh38
rs955603111 Chr.5: 878382-878382 on Build GRCh38
rs766186216 Chr.5: 878451-878451 on Build GRCh38
rs763674604 Chr.5: 878403-878403 on Build GRCh38
rs1017315995 Chr.5: 878291-878291 on Build GRCh38
rs543226350 Chr.5: 878316-878316 on Build GRCh38
rs534644668 Chr.5: 878736-878736 on Build GRCh38
rs111644085 Chr.5: 878421-878421 on Build GRCh38
rs753380128 Chr.5: 878407-878407 on Build GRCh38
rs936137899 Chr.5: 878468-878468 on Build GRCh38
rs764739306 Chr.5: 878484-878484 on Build GRCh38
rs778767370 Chr.5: 878413-878413 on Build GRCh38
rs765914354 Chr.5: 878501-878502 on Build GRCh38
rs747631392 Chr.5: 878308-878308 on Build GRCh38
rs773923249 Chr.5: 878339-878339 on Build GRCh38
rs537201571 Chr.5: 878570-878570 on Build GRCh38
rs747828561 Chr.5: 878339-878340 on Build GRCh38
rs753291718 Chr.5: 878358-878358 on Build GRCh38
rs769963721 Chr.5: 878338-878338 on Build GRCh38
rs376194540 Chr.5: 878550-878550 on Build GRCh38
rs371708847 Chr.5: 878372-878372 on Build GRCh38
rs781551309 Chr.5: 878535-878535 on Build GRCh38
rs508016 Chr.5: 878410-878410 on Build GRCh38
rs60153305 Chr.5: 878642-878642 on Build GRCh38
rs758388910 Chr.5: 878528-878528 on Build GRCh38
rs758194342 Chr.5: 878416-878416 on Build GRCh38
rs775929609 Chr.5: 878470-878470 on Build GRCh38
rs745612844 Chr.5: 878324-878324 on Build GRCh38
rs775841419 Chr.5: 878430-878430 on Build GRCh38
rs564800119 Chr.5: 878431-878431 on Build GRCh38
rs746369071 Chr.5: 878429-878429 on Build GRCh38
rs781310944 Chr.5: 878368-878368 on Build GRCh38
rs76072224 Chr.5: 878346-878346 on Build GRCh38
rs749214187 Chr.5: 878314-878314 on Build GRCh38
rs73733935 Chr.5: 878333-878333 on Build GRCh38
rs749427415 Chr.5: 878425-878425 on Build GRCh38
rs761354730 Chr.5: 878440-878440 on Build GRCh38
rs776247216 Chr.5: 878439-878439 on Build GRCh38
rs747512178 Chr.5: 878534-878534 on Build GRCh38
rs111758433 Chr.5: 878319-878319 on Build GRCh38
rs577057556 Chr.5: 878648-878648 on Build GRCh38
rs778872586 Chr.5: 878361-878361 on Build GRCh38
rs779060319 Chr.5: 878461-878461 on Build GRCh38
rs748377915 Chr.5: 878371-878371 on Build GRCh38
rs139057857 Chr.5: 878334-878334 on Build GRCh38
rs748395756 Chr.5: 878537-878537 on Build GRCh38
rs751422052 Chr.5: 878452-878452 on Build GRCh38
rs41283155 Chr.5: 878684-878684 on Build GRCh38
rs868397287 Chr.5: 878601-878601 on Build GRCh38
rs73733934 Chr.5: 878306-878306 on Build GRCh38
rs145445077 Chr.5: 878360-878360 on Build GRCh38
rs756436277 Chr.5: 878418-878418 on Build GRCh38
rs778966110 Chr.5: 878521-878521 on Build GRCh38
rs199826285 Chr.5: 878376-878376 on Build GRCh38
rs559125334 Chr.5: 878367-878367 on Build GRCh38
rs767122037 Chr.5: 878350-878350 on Build GRCh38
rs764801062 Chr.5: 878441-878441 on Build GRCh38
rs757335221 Chr.5: 878460-878460 on Build GRCh38
rs780773076 Chr.5: 878327-878327 on Build GRCh38
rs41283151 Chr.5: 878632-878632 on Build GRCh38
rs529793007 Chr.5: 878385-878385 on Build GRCh38
rs367613682 Chr.5: 878340-878340 on Build GRCh38
rs372141822 Chr.5: 878494-878494 on Build GRCh38
rs375651328 Chr.5: 878426-878426 on Build GRCh38
rs368820375 Chr.5: 878498-878498 on Build GRCh38
rs554318861 Chr.5: 878299-878299 on Build GRCh38
rs780581409 Chr.5: 878533-878533 on Build GRCh38
rs745675628 Chr.5: 878366-878366 on Build GRCh38
rs747426013 Chr.5: 878473-878473 on Build GRCh38
rs372409324 Chr.5: 878392-878392 on Build GRCh38
rs772809053 Chr.5: 878485-878485 on Build GRCh38
rs531087363 Chr.5: 878417-878417 on Build GRCh38
rs41283153 Chr.5: 878645-878645 on Build GRCh38
rs754796515 Chr.5: 878453-878453 on Build GRCh38
rs569016921 Chr.5: 878541-878541 on Build GRCh38
rs777723817 Chr.5: 878312-878312 on Build GRCh38
rs755595468 Chr.5: 878310-878310 on Build GRCh38
rs757503157 Chr.5: 878507-878507 on Build GRCh38

View Full Product Details