Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 887274-887767 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRD9
Entrez Gene ID
65980
Gene Name
bromodomain containing 9
Gene Aliases
LAVS3040, PRO9856
UniGene
Hs.449278
Gene Chromosome Location
Chr.5: 863735-892824 on Build GRCh38
Transcripts
NM_001009877.2 NM_001317951.1 NM_023924.4 NR_134296.1 XM_006714494.3 XM_017009780.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3776914 c.348A>T p.A116A Chr.5: 887382-887382 on Build GRCh38
5061253 c.345C>T p.H115H Chr.5: 887385-887385 on Build GRCh38
354502 c.607G>C p.A203P Chr.5: 887471-887471 on Build GRCh38
7167748 c.711G>A p.M237I Chr.5: 887367-887367 on Build GRCh38
6171809 c.276G>T p.M92I Chr.5: 887454-887454 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1047318719 Chr.5: 887307-887307 on Build GRCh38
rs1024524610 Chr.5: 887571-887571 on Build GRCh38
rs543410636 Chr.5: 887628-887628 on Build GRCh38
rs183710737 Chr.5: 887572-887572 on Build GRCh38
rs933989709 Chr.5: 887629-887629 on Build GRCh38
rs770557326 Chr.5: 887462-887462 on Build GRCh38
rs780966841 Chr.5: 887319-887319 on Build GRCh38
rs747423821 Chr.5: 887314-887314 on Build GRCh38
rs564984069 Chr.5: 887295-887295 on Build GRCh38
rs761948572 Chr.5: 887384-887384 on Build GRCh38
rs73733979 Chr.5: 887744-887744 on Build GRCh38
rs566253425 Chr.5: 887424-887424 on Build GRCh38
rs560080716 Chr.5: 887338-887338 on Build GRCh38
rs377290566 Chr.5: 887392-887392 on Build GRCh38
rs564971955 Chr.5: 887633-887633 on Build GRCh38
rs552340981 Chr.5: 887493-887493 on Build GRCh38
rs577055055 Chr.5: 887639-887639 on Build GRCh38
rs188474105 Chr.5: 887687-887687 on Build GRCh38
rs529227290 Chr.5: 887497-887497 on Build GRCh38
rs898942006 Chr.5: 887529-887529 on Build GRCh38
rs985525155 Chr.5: 887307-887308 on Build GRCh38
rs1047941989 Chr.5: 887711-887711 on Build GRCh38
rs747243709 Chr.5: 887557-887557 on Build GRCh38
rs143306265 Chr.5: 887503-887503 on Build GRCh38
rs755743518 Chr.5: 887433-887433 on Build GRCh38
rs923497704 Chr.5: 887670-887670 on Build GRCh38
rs371347859 Chr.5: 887397-887397 on Build GRCh38
rs931010609 Chr.5: 887304-887304 on Build GRCh38
rs1013893409 Chr.5: 887730-887730 on Build GRCh38
rs906155122 Chr.5: 887742-887742 on Build GRCh38
rs1051687043 Chr.5: 887426-887426 on Build GRCh38
rs775294530 Chr.5: 887382-887382 on Build GRCh38
rs745804490 Chr.5: 887484-887484 on Build GRCh38
rs575909229 Chr.5: 887615-887615 on Build GRCh38
rs753686468 Chr.5: 887479-887479 on Build GRCh38
rs943618683 Chr.5: 887317-887317 on Build GRCh38
rs541622763 Chr.5: 887650-887650 on Build GRCh38
rs1013841101 Chr.5: 887567-887567 on Build GRCh38
rs911832428 Chr.5: 887658-887658 on Build GRCh38
rs747745223 Chr.5: 887328-887328 on Build GRCh38
rs548060566 Chr.5: 887331-887331 on Build GRCh38
rs774395538 Chr.5: 887341-887341 on Build GRCh38
rs115590995 Chr.5: 887610-887610 on Build GRCh38
rs191899069 Chr.5: 887354-887354 on Build GRCh38
rs73733976 Chr.5: 887418-887418 on Build GRCh38
rs574239695 Chr.5: 887305-887305 on Build GRCh38
rs80179458 Chr.5: 887313-887313 on Build GRCh38
rs910841172 Chr.5: 887308-887308 on Build GRCh38
rs980263509 Chr.5: 887591-887591 on Build GRCh38
rs758451130 Chr.5: 887407-887407 on Build GRCh38
rs986772096 Chr.5: 887652-887652 on Build GRCh38
rs910885907 Chr.5: 887649-887649 on Build GRCh38
rs765407397 Chr.5: 887385-887385 on Build GRCh38
rs777545295 Chr.5: 887445-887445 on Build GRCh38
rs768872205 Chr.5: 887318-887318 on Build GRCh38
rs772879674 Chr.5: 887337-887337 on Build GRCh38
rs750529019 Chr.5: 887400-887400 on Build GRCh38
rs930452449 Chr.5: 887693-887693 on Build GRCh38
rs919661012 Chr.5: 887298-887298 on Build GRCh38
rs879312108 Chr.5: 887725-887725 on Build GRCh38
rs139290930 Chr.5: 887488-887488 on Build GRCh38
rs761133725 Chr.5: 887474-887478 on Build GRCh38
rs748843804 Chr.5: 887457-887457 on Build GRCh38
rs73733977 Chr.5: 887611-887611 on Build GRCh38
rs886106422 Chr.5: 887563-887563 on Build GRCh38
rs553583048 Chr.5: 887546-887546 on Build GRCh38
rs943252508 Chr.5: 887666-887666 on Build GRCh38
rs371013959 Chr.5: 887480-887480 on Build GRCh38
rs752058954 Chr.5: 887412-887412 on Build GRCh38
rs369901324 Chr.5: 887330-887330 on Build GRCh38
rs73733978 Chr.5: 887726-887726 on Build GRCh38
rs895157474 Chr.5: 887729-887729 on Build GRCh38
rs147123384 Chr.5: 887521-887521 on Build GRCh38
rs766912064 Chr.5: 887410-887410 on Build GRCh38
rs532708557 Chr.5: 887300-887300 on Build GRCh38
rs758938870 Chr.5: 887311-887311 on Build GRCh38
rs536800989 Chr.5: 887539-887539 on Build GRCh38
rs752401539 Chr.5: 887684-887684 on Build GRCh38
rs765390649 Chr.5: 887487-887488 on Build GRCh38
rs903984389 Chr.5: 887691-887691 on Build GRCh38
rs563950201 Chr.5: 887733-887733 on Build GRCh38
rs77738320 Chr.5: 887524-887524 on Build GRCh38
rs373102101 Chr.5: 887349-887349 on Build GRCh38
rs1013935176 Chr.5: 887555-887555 on Build GRCh38

View Full Product Details