Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 1334978-1335479 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLPTM1L
Entrez Gene ID
81037
Gene Name
CLPTM1 like
Gene Aliases
CRR9
UniGene
Hs.444673
Gene Chromosome Location
Chr.5: 1317744-1345070 on Build GRCh38
Transcripts
NM_030782.4 XM_011514144.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1179336 c.749G>A p.R250H Chr.5: 1335104-1335104 on Build GRCh38
3137996 c.689G>A p.R230H Chr.5: 1335164-1335164 on Build GRCh38
6546007 c.768G>A p.Q256Q Chr.5: 1335085-1335085 on Build GRCh38
5834937 c.764delT p.M255fs*21 Chr.5: 1335089-1335089 on Build GRCh38
3850154 c.724G>A p.D242N Chr.5: 1335129-1335129 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376231200 Chr.5: 1335041-1335041 on Build GRCh38
rs899880495 Chr.5: 1335259-1335259 on Build GRCh38
rs980550533 Chr.5: 1335355-1335355 on Build GRCh38
rs1018229824 Chr.5: 1335240-1335240 on Build GRCh38
rs772648892 Chr.5: 1335164-1335164 on Build GRCh38
rs563815189 Chr.5: 1335454-1335454 on Build GRCh38
rs375455299 Chr.5: 1335219-1335219 on Build GRCh38
rs942395857 Chr.5: 1335418-1335418 on Build GRCh38
rs368537107 Chr.5: 1335388-1335388 on Build GRCh38
rs936311171 Chr.5: 1335370-1335370 on Build GRCh38
rs182822447 Chr.5: 1335029-1335029 on Build GRCh38
rs765506209 Chr.5: 1335198-1335198 on Build GRCh38
rs142613582 Chr.5: 1335120-1335120 on Build GRCh38
rs1050900193 Chr.5: 1335401-1335401 on Build GRCh38
rs376537532 Chr.5: 1335006-1335006 on Build GRCh38
rs137936715 Chr.5: 1335024-1335024 on Build GRCh38
rs781446800 Chr.5: 1335216-1335217 on Build GRCh38
rs559390900 Chr.5: 1335192-1335192 on Build GRCh38
rs200411343 Chr.5: 1335153-1335153 on Build GRCh38
rs778652784 Chr.5: 1335130-1335130 on Build GRCh38
rs376982395 Chr.5: 1335111-1335111 on Build GRCh38
rs746449854 Chr.5: 1335142-1335142 on Build GRCh38
rs139544692 Chr.5: 1335361-1335361 on Build GRCh38
rs187861381 Chr.5: 1335227-1335227 on Build GRCh38
rs979678042 Chr.5: 1335366-1335366 on Build GRCh38
rs370635276 Chr.5: 1335048-1335048 on Build GRCh38
rs1020328806 Chr.5: 1335054-1335054 on Build GRCh38
rs200350933 Chr.5: 1335276-1335277 on Build GRCh38
rs747860215 Chr.5: 1335154-1335154 on Build GRCh38
rs116659965 Chr.5: 1335079-1335079 on Build GRCh38
rs34536907 Chr.5: 1335061-1335061 on Build GRCh38
rs750253978 Chr.5: 1335138-1335138 on Build GRCh38
rs149669133 Chr.5: 1335064-1335064 on Build GRCh38
rs898388014 Chr.5: 1335444-1335444 on Build GRCh38
rs150993328 Chr.5: 1335165-1335165 on Build GRCh38
rs777399989 Chr.5: 1335023-1335023 on Build GRCh38
rs199623057 Chr.5: 1335042-1335042 on Build GRCh38
rs995913129 Chr.5: 1335267-1335267 on Build GRCh38
rs749874280 Chr.5: 1335125-1335125 on Build GRCh38
rs61417568 Chr.5: 1335275-1335276 on Build GRCh38
rs750210697 Chr.5: 1335206-1335206 on Build GRCh38
rs767728138 Chr.5: 1335045-1335045 on Build GRCh38
rs564233947 Chr.5: 1335058-1335058 on Build GRCh38
rs34490357 Chr.5: 1335011-1335011 on Build GRCh38
rs774429155 Chr.5: 1335292-1335292 on Build GRCh38
rs1016847433 Chr.5: 1335234-1335234 on Build GRCh38
rs1027212827 Chr.5: 1335282-1335282 on Build GRCh38
rs193920801 Chr.5: 1335104-1335104 on Build GRCh38
rs772880482 Chr.5: 1335081-1335081 on Build GRCh38
rs766980280 Chr.5: 1335008-1335008 on Build GRCh38
rs760035535 Chr.5: 1335160-1335160 on Build GRCh38
rs980414735 Chr.5: 1335131-1335131 on Build GRCh38
rs762060593 Chr.5: 1335195-1335195 on Build GRCh38
rs763163114 Chr.5: 1335094-1335094 on Build GRCh38
rs376007679 Chr.5: 1335317-1335317 on Build GRCh38
rs772365834 Chr.5: 1335316-1335316 on Build GRCh38
rs760098618 Chr.5: 1335110-1335110 on Build GRCh38
rs190188803 Chr.5: 1335020-1335020 on Build GRCh38
rs766798575 Chr.5: 1335102-1335102 on Build GRCh38
rs767667830 Chr.5: 1335447-1335447 on Build GRCh38
rs1051588150 Chr.5: 1335448-1335448 on Build GRCh38
rs150538822 Chr.5: 1335289-1335289 on Build GRCh38
rs200650793 Chr.5: 1335047-1335047 on Build GRCh38
rs373729867 Chr.5: 1335018-1335018 on Build GRCh38
rs751547916 Chr.5: 1335222-1335222 on Build GRCh38
rs760820822 Chr.5: 1335112-1335112 on Build GRCh38
rs961050568 Chr.5: 1335334-1335334 on Build GRCh38
rs375939209 Chr.5: 1335035-1335035 on Build GRCh38
rs200514830 Chr.5: 1335098-1335098 on Build GRCh38
rs533914606 Chr.5: 1335238-1335238 on Build GRCh38
rs56966034 Chr.5: 1335353-1335353 on Build GRCh38
rs759468040 Chr.5: 1335209-1335209 on Build GRCh38
rs187613637 Chr.5: 1335017-1335017 on Build GRCh38
rs751581252 Chr.5: 1335016-1335016 on Build GRCh38
rs773447749 Chr.5: 1335038-1335038 on Build GRCh38
rs970461529 Chr.5: 1335341-1335341 on Build GRCh38
rs770758947 Chr.5: 1335025-1335025 on Build GRCh38
rs755924442 Chr.5: 1335021-1335021 on Build GRCh38
rs746817950 Chr.5: 1335082-1335082 on Build GRCh38
rs764254657 Chr.5: 1335189-1335189 on Build GRCh38
rs774951293 Chr.5: 1335105-1335105 on Build GRCh38
rs200594051 Chr.5: 1335037-1335037 on Build GRCh38
rs925549745 Chr.5: 1335375-1335375 on Build GRCh38
rs758332047 Chr.5: 1335139-1335139 on Build GRCh38
rs771598444 Chr.5: 1335034-1335034 on Build GRCh38
rs768931369 Chr.5: 1335176-1335178 on Build GRCh38
rs201214061 Chr.5: 1335043-1335043 on Build GRCh38
rs920132688 Chr.5: 1335005-1335005 on Build GRCh38
rs756950098 Chr.5: 1335129-1335129 on Build GRCh38
rs114266820 Chr.5: 1335193-1335193 on Build GRCh38
rs575085353 Chr.5: 1335362-1335362 on Build GRCh38
rs3030832 Chr.5: 1335271-1335272 on Build GRCh38
rs933702375 Chr.5: 1335397-1335397 on Build GRCh38
rs745309134 Chr.5: 1335033-1335033 on Build GRCh38
rs979687948 Chr.5: 1335106-1335106 on Build GRCh38
rs892381006 Chr.5: 1335172-1335172 on Build GRCh38
rs369713965 Chr.5: 1335415-1335415 on Build GRCh38
rs768219512 Chr.5: 1335147-1335147 on Build GRCh38
rs1038235502 Chr.5: 1335443-1335443 on Build GRCh38
rs772043717 Chr.5: 1335065-1335065 on Build GRCh38
rs988692220 Chr.5: 1335386-1335386 on Build GRCh38
rs892332585 Chr.5: 1335044-1335044 on Build GRCh38
rs375172852 Chr.5: 1335379-1335379 on Build GRCh38
rs760630498 Chr.5: 1335180-1335180 on Build GRCh38
rs145488125 Chr.5: 1335070-1335070 on Build GRCh38
rs200986888 Chr.5: 1335211-1335211 on Build GRCh38
rs535743877 Chr.5: 1335335-1335335 on Build GRCh38

View Full Product Details