Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 31313228-31313731 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH6
Entrez Gene ID
1004
Gene Name
cadherin 6
Gene Aliases
CAD6, KCAD
UniGene
Hs.124776
Gene Chromosome Location
Chr.5: 31193655-31329146 on Build GRCh38
Transcripts
NM_004932.3 XM_011513921.2 XM_017008910.1 XM_017008911.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4594421 c.1322_1343del22 p.F441fs*1 Chr.5: 31313386-31313407 on Build GRCh38
4597991 c.1281G>A p.M427I Chr.5: 31313345-31313345 on Build GRCh38
234526 c.1329G>A p.S443S Chr.5: 31313393-31313393 on Build GRCh38
3615336 c.1313G>A p.G438D Chr.5: 31313377-31313377 on Build GRCh38
1436980 c.1328C>T p.S443L Chr.5: 31313392-31313392 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142200699 Chr.5: 31313393-31313393 on Build GRCh38
rs538951609 Chr.5: 31313525-31313525 on Build GRCh38
rs959204020 Chr.5: 31313703-31313703 on Build GRCh38
rs768002288 Chr.5: 31313390-31313390 on Build GRCh38
rs367617909 Chr.5: 31313407-31313407 on Build GRCh38
rs552471855 Chr.5: 31313568-31313568 on Build GRCh38
rs1054407974 Chr.5: 31313698-31313698 on Build GRCh38
rs774760334 Chr.5: 31313381-31313381 on Build GRCh38
rs779613584 Chr.5: 31313333-31313333 on Build GRCh38
rs927543463 Chr.5: 31313362-31313362 on Build GRCh38
rs763482229 Chr.5: 31313376-31313376 on Build GRCh38
rs746694742 Chr.5: 31313360-31313360 on Build GRCh38
rs201412636 Chr.5: 31313491-31313491 on Build GRCh38
rs986379823 Chr.5: 31313429-31313429 on Build GRCh38
rs534777505 Chr.5: 31313682-31313682 on Build GRCh38
rs774935348 Chr.5: 31313478-31313478 on Build GRCh38
rs901204157 Chr.5: 31313573-31313573 on Build GRCh38
rs371463771 Chr.5: 31313368-31313368 on Build GRCh38
rs1025805178 Chr.5: 31313262-31313262 on Build GRCh38
rs556963406 Chr.5: 31313308-31313308 on Build GRCh38
rs771406443 Chr.5: 31313380-31313380 on Build GRCh38
rs750364235 Chr.5: 31313423-31313423 on Build GRCh38
rs757216983 Chr.5: 31313413-31313413 on Build GRCh38
rs375136266 Chr.5: 31313489-31313489 on Build GRCh38
rs768230312 Chr.5: 31313465-31313465 on Build GRCh38
rs1045517022 Chr.5: 31313653-31313653 on Build GRCh38
rs916945882 Chr.5: 31313378-31313378 on Build GRCh38
rs764214082 Chr.5: 31313394-31313394 on Build GRCh38
rs752985511 Chr.5: 31313277-31313277 on Build GRCh38
rs778775598 Chr.5: 31313417-31313417 on Build GRCh38
rs565885347 Chr.5: 31313600-31313600 on Build GRCh38
rs957646178 Chr.5: 31313398-31313398 on Build GRCh38
rs776760199 Chr.5: 31313495-31313495 on Build GRCh38
rs994235008 Chr.5: 31313625-31313625 on Build GRCh38
rs752932258 Chr.5: 31313392-31313392 on Build GRCh38
rs958359089 Chr.5: 31313363-31313363 on Build GRCh38
rs1025752752 Chr.5: 31313637-31313637 on Build GRCh38
rs776053051 Chr.5: 31313374-31313374 on Build GRCh38
rs370528050 Chr.5: 31313474-31313474 on Build GRCh38
rs34885550 Chr.5: 31313273-31313274 on Build GRCh38
rs998881310 Chr.5: 31313660-31313660 on Build GRCh38
rs762414512 Chr.5: 31313278-31313281 on Build GRCh38
rs952822108 Chr.5: 31313642-31313642 on Build GRCh38
rs373772247 Chr.5: 31313384-31313384 on Build GRCh38
rs1034562034 Chr.5: 31313686-31313686 on Build GRCh38
rs180792041 Chr.5: 31313482-31313482 on Build GRCh38
rs780746375 Chr.5: 31313473-31313473 on Build GRCh38
rs745730316 Chr.5: 31313304-31313304 on Build GRCh38
rs753763360 Chr.5: 31313288-31313288 on Build GRCh38
rs748303694 Chr.5: 31313530-31313530 on Build GRCh38
rs966245834 Chr.5: 31313438-31313438 on Build GRCh38
rs772518328 Chr.5: 31313483-31313483 on Build GRCh38
rs778739695 Chr.5: 31313303-31313303 on Build GRCh38
rs952880904 Chr.5: 31313298-31313298 on Build GRCh38
rs771700462 Chr.5: 31313329-31313329 on Build GRCh38
rs777748756 Chr.5: 31313280-31313280 on Build GRCh38
rs746641388 Chr.5: 31313445-31313445 on Build GRCh38
rs138335545 Chr.5: 31313433-31313433 on Build GRCh38
rs757192976 Chr.5: 31313299-31313299 on Build GRCh38
rs569654135 Chr.5: 31313524-31313524 on Build GRCh38
rs764292015 Chr.5: 31313492-31313492 on Build GRCh38
rs949915496 Chr.5: 31313646-31313646 on Build GRCh38
rs375114720 Chr.5: 31313468-31313468 on Build GRCh38
rs372686442 Chr.5: 31313500-31313500 on Build GRCh38
rs776298187 Chr.5: 31313435-31313435 on Build GRCh38
rs756201117 Chr.5: 31313279-31313279 on Build GRCh38
rs1006135309 Chr.5: 31313659-31313659 on Build GRCh38

View Full Product Details