Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 37121378-37121880 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C5orf42
Entrez Gene ID
65250
Gene Name
chromosome 5 open reading frame 42
Gene Aliases
Hug, JBTS17, OFD6
UniGene
Hs.586199
Gene Chromosome Location
Chr.5: 37063928-37249428 on Build GRCh38
Transcripts
NM_023073.3 XM_005248345.3 XM_005248346.3 XM_005248347.3 XM_005248349.3 XM_005248350.3 XM_006714491.3 XM_011514085.2 XM_011514086.2 XM_011514087.2 XM_011514088.2 XM_011514089.2 XM_011514090.2 XM_011514094.2 XM_017009760.1 XM_017009761.1 XM_017009762.1 XM_017009763.1 XM_017009764.1 XM_017009765.1 XM_017009766.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2688144 c.5595C>T p.Y1865Y Chr.5: 37121739-37121739 on Build GRCh38
2688134 c.5680C>T p.Q1894* Chr.5: 37121654-37121654 on Build GRCh38
4628429 c.8949A>G p.L2983L Chr.5: 37121691-37121691 on Build GRCh38
2688146 c.5592G>A p.A1864A Chr.5: 37121742-37121742 on Build GRCh38
4667822 c.8882G>A p.R2961H Chr.5: 37121758-37121758 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs557372169 Chr.5: 37121647-37121647 on Build GRCh38
rs749855192 Chr.5: 37121829-37121829 on Build GRCh38
rs780529334 Chr.5: 37121706-37121709 on Build GRCh38
rs944160435 Chr.5: 37121688-37121688 on Build GRCh38
rs974002627 Chr.5: 37121405-37121405 on Build GRCh38
rs1006171379 Chr.5: 37121733-37121733 on Build GRCh38
rs760027584 Chr.5: 37121633-37121633 on Build GRCh38
rs200358545 Chr.5: 37121771-37121771 on Build GRCh38
rs1011838329 Chr.5: 37121487-37121487 on Build GRCh38
rs758829030 Chr.5: 37121645-37121645 on Build GRCh38
rs768143928 Chr.5: 37121646-37121646 on Build GRCh38
rs777181922 Chr.5: 37121617-37121617 on Build GRCh38
rs547548409 Chr.5: 37121454-37121454 on Build GRCh38
rs960770953 Chr.5: 37121560-37121562 on Build GRCh38
rs771321110 Chr.5: 37121722-37121722 on Build GRCh38
rs10491422 Chr.5: 37121548-37121548 on Build GRCh38
rs755235404 Chr.5: 37121717-37121718 on Build GRCh38
rs969240792 Chr.5: 37121583-37121583 on Build GRCh38
rs752727888 Chr.5: 37121775-37121775 on Build GRCh38
rs753593935 Chr.5: 37121767-37121767 on Build GRCh38
rs573500656 Chr.5: 37121727-37121727 on Build GRCh38
rs767189816 Chr.5: 37121656-37121656 on Build GRCh38
rs771249284 Chr.5: 37121600-37121600 on Build GRCh38
rs370799496 Chr.5: 37121580-37121580 on Build GRCh38
rs794727691 Chr.5: 37121717-37121717 on Build GRCh38
rs16903511 Chr.5: 37121664-37121664 on Build GRCh38
rs942338818 Chr.5: 37121497-37121497 on Build GRCh38
rs1041919795 Chr.5: 37121643-37121643 on Build GRCh38
rs781525957 Chr.5: 37121762-37121763 on Build GRCh38
rs748606873 Chr.5: 37121483-37121483 on Build GRCh38
rs141014620 Chr.5: 37121709-37121709 on Build GRCh38
rs377142277 Chr.5: 37121739-37121739 on Build GRCh38
rs148832999 Chr.5: 37121541-37121541 on Build GRCh38
rs973974455 Chr.5: 37121599-37121599 on Build GRCh38
rs781573320 Chr.5: 37121807-37121807 on Build GRCh38
rs780517679 Chr.5: 37121828-37121828 on Build GRCh38
rs79995491 Chr.5: 37121415-37121415 on Build GRCh38
rs548661330 Chr.5: 37121490-37121490 on Build GRCh38
rs758537775 Chr.5: 37121777-37121777 on Build GRCh38
rs555079361 Chr.5: 37121715-37121715 on Build GRCh38
rs1030734277 Chr.5: 37121555-37121555 on Build GRCh38
rs770043266 Chr.5: 37121827-37121828 on Build GRCh38
rs150242262 Chr.5: 37121756-37121756 on Build GRCh38
rs761407466 Chr.5: 37121652-37121652 on Build GRCh38
rs751814670 Chr.5: 37121711-37121717 on Build GRCh38
rs748261423 Chr.5: 37121797-37121800 on Build GRCh38
rs766147646 Chr.5: 37121665-37121665 on Build GRCh38
rs747363351 Chr.5: 37121586-37121586 on Build GRCh38
rs752850962 Chr.5: 37121703-37121703 on Build GRCh38
rs1030945020 Chr.5: 37121763-37121763 on Build GRCh38
rs143453963 Chr.5: 37121764-37121764 on Build GRCh38
rs748760500 Chr.5: 37121472-37121472 on Build GRCh38
rs750972473 Chr.5: 37121616-37121621 on Build GRCh38
rs1023889958 Chr.5: 37121571-37121571 on Build GRCh38
rs751736082 Chr.5: 37121800-37121800 on Build GRCh38
rs921440177 Chr.5: 37121730-37121730 on Build GRCh38
rs930032982 Chr.5: 37121428-37121428 on Build GRCh38
rs1013415541 Chr.5: 37121561-37121561 on Build GRCh38
rs747253172 Chr.5: 37121721-37121721 on Build GRCh38
rs779085289 Chr.5: 37121689-37121689 on Build GRCh38
rs746251684 Chr.5: 37121812-37121812 on Build GRCh38
rs374088464 Chr.5: 37121670-37121670 on Build GRCh38
rs781761071 Chr.5: 37121451-37121451 on Build GRCh38
rs1034379733 Chr.5: 37121702-37121702 on Build GRCh38
rs910912824 Chr.5: 37121491-37121491 on Build GRCh38
rs567149383 Chr.5: 37121500-37121500 on Build GRCh38
rs540797695 Chr.5: 37121738-37121738 on Build GRCh38
rs754882012 Chr.5: 37121676-37121676 on Build GRCh38
rs778065504 Chr.5: 37121788-37121788 on Build GRCh38
rs138862600 Chr.5: 37121759-37121759 on Build GRCh38
rs769430882 Chr.5: 37121486-37121486 on Build GRCh38
rs370851364 Chr.5: 37121742-37121742 on Build GRCh38
rs962271195 Chr.5: 37121462-37121462 on Build GRCh38
rs750116851 Chr.5: 37121658-37121658 on Build GRCh38
rs771622959 Chr.5: 37121758-37121758 on Build GRCh38
rs918360406 Chr.5: 37121477-37121477 on Build GRCh38
rs893007797 Chr.5: 37121546-37121546 on Build GRCh38
rs534480328 Chr.5: 37121534-37121534 on Build GRCh38
rs769109400 Chr.5: 37121427-37121427 on Build GRCh38
rs760313941 Chr.5: 37121762-37121762 on Build GRCh38
rs764899843 Chr.5: 37121844-37121844 on Build GRCh38
rs368643781 Chr.5: 37121805-37121805 on Build GRCh38
rs1047160952 Chr.5: 37121441-37121441 on Build GRCh38
rs919902868 Chr.5: 37121630-37121630 on Build GRCh38
rs773810601 Chr.5: 37121745-37121745 on Build GRCh38
rs756511467 Chr.5: 37121813-37121813 on Build GRCh38
rs538658773 Chr.5: 37121579-37121579 on Build GRCh38

View Full Product Details