Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 40852232-40852737 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CARD6
Entrez Gene ID
84674
Gene Name
caspase recruitment domain family member 6
Gene Aliases
CINCIN1
UniGene
Hs.200242
Gene Chromosome Location
Chr.5: 40841190-40855354 on Build GRCh38
Transcripts
NM_032587.3 XM_017009989.1 XM_017009990.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1437569 c.973T>A p.L325I Chr.5: 40852305-40852305 on Build GRCh38
1068194 c.966A>G p.P322P Chr.5: 40852298-40852298 on Build GRCh38
1437570 c.1264G>A p.G422R Chr.5: 40852596-40852596 on Build GRCh38
1068196 c.1254C>A p.I418I Chr.5: 40852586-40852586 on Build GRCh38
5000416 c.1042A>G p.K348E Chr.5: 40852374-40852374 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764329594 Chr.5: 40852285-40852285 on Build GRCh38
rs531378160 Chr.5: 40852630-40852630 on Build GRCh38
rs780800033 Chr.5: 40852497-40852498 on Build GRCh38
rs773167124 Chr.5: 40852610-40852610 on Build GRCh38
rs113915437 Chr.5: 40852463-40852463 on Build GRCh38
rs758933185 Chr.5: 40852544-40852544 on Build GRCh38
rs552200656 Chr.5: 40852531-40852532 on Build GRCh38
rs767810450 Chr.5: 40852391-40852391 on Build GRCh38
rs538870423 Chr.5: 40852681-40852681 on Build GRCh38
rs762964993 Chr.5: 40852284-40852284 on Build GRCh38
rs766607266 Chr.5: 40852614-40852614 on Build GRCh38
rs745567681 Chr.5: 40852441-40852441 on Build GRCh38
rs150754394 Chr.5: 40852269-40852269 on Build GRCh38
rs757201919 Chr.5: 40852710-40852711 on Build GRCh38
rs766519414 Chr.5: 40852515-40852515 on Build GRCh38
rs892815827 Chr.5: 40852696-40852696 on Build GRCh38
rs757505639 Chr.5: 40852296-40852296 on Build GRCh38
rs759199449 Chr.5: 40852367-40852367 on Build GRCh38
rs3812030 Chr.5: 40852516-40852516 on Build GRCh38
rs143022216 Chr.5: 40852672-40852672 on Build GRCh38
rs753198481 Chr.5: 40852530-40852530 on Build GRCh38
rs200590550 Chr.5: 40852351-40852351 on Build GRCh38
rs376800897 Chr.5: 40852447-40852447 on Build GRCh38
rs763376155 Chr.5: 40852677-40852677 on Build GRCh38
rs764606208 Chr.5: 40852678-40852678 on Build GRCh38
rs576851671 Chr.5: 40852406-40852406 on Build GRCh38
rs772684643 Chr.5: 40852259-40852259 on Build GRCh38
rs267600629 Chr.5: 40852457-40852457 on Build GRCh38
rs779331588 Chr.5: 40852421-40852421 on Build GRCh38
rs767540928 Chr.5: 40852482-40852482 on Build GRCh38
rs745318819 Chr.5: 40852559-40852559 on Build GRCh38
rs755646040 Chr.5: 40852693-40852693 on Build GRCh38
rs139145302 Chr.5: 40852275-40852275 on Build GRCh38
rs774977188 Chr.5: 40852629-40852630 on Build GRCh38
rs759432156 Chr.5: 40852401-40852401 on Build GRCh38
rs755283154 Chr.5: 40852517-40852517 on Build GRCh38
rs538146189 Chr.5: 40852422-40852422 on Build GRCh38
rs750821489 Chr.5: 40852311-40852311 on Build GRCh38
rs371334015 Chr.5: 40852395-40852395 on Build GRCh38
rs143505234 Chr.5: 40852578-40852578 on Build GRCh38
rs1032935410 Chr.5: 40852588-40852588 on Build GRCh38
rs755846640 Chr.5: 40852560-40852560 on Build GRCh38
rs775414145 Chr.5: 40852372-40852372 on Build GRCh38
rs779679586 Chr.5: 40852703-40852703 on Build GRCh38
rs751687948 Chr.5: 40852292-40852292 on Build GRCh38
rs761750479 Chr.5: 40852390-40852390 on Build GRCh38
rs377732886 Chr.5: 40852507-40852507 on Build GRCh38
rs756761924 Chr.5: 40852434-40852434 on Build GRCh38
rs777329484 Chr.5: 40852345-40852345 on Build GRCh38
rs151076474 Chr.5: 40852612-40852612 on Build GRCh38
rs1049025549 Chr.5: 40852673-40852673 on Build GRCh38
rs369845824 Chr.5: 40852477-40852477 on Build GRCh38
rs139946405 Chr.5: 40852576-40852576 on Build GRCh38
rs749841915 Chr.5: 40852339-40852339 on Build GRCh38
rs754194377 Chr.5: 40852414-40852414 on Build GRCh38
rs113160122 Chr.5: 40852455-40852455 on Build GRCh38
rs886728420 Chr.5: 40852694-40852694 on Build GRCh38
rs770480969 Chr.5: 40852266-40852266 on Build GRCh38
rs897118756 Chr.5: 40852584-40852584 on Build GRCh38
rs760375788 Chr.5: 40852264-40852264 on Build GRCh38
rs776882109 Chr.5: 40852617-40852617 on Build GRCh38
rs769134907 Chr.5: 40852344-40852344 on Build GRCh38
rs749924535 Chr.5: 40852689-40852689 on Build GRCh38
rs566975527 Chr.5: 40852348-40852348 on Build GRCh38
rs183932779 Chr.5: 40852506-40852506 on Build GRCh38
rs569969060 Chr.5: 40852402-40852402 on Build GRCh38
rs3812031 Chr.5: 40852609-40852609 on Build GRCh38
rs765651097 Chr.5: 40852658-40852658 on Build GRCh38
rs78772032 Chr.5: 40852435-40852435 on Build GRCh38
rs572093435 Chr.5: 40852605-40852605 on Build GRCh38
rs765559234 Chr.5: 40852519-40852519 on Build GRCh38
rs769528277 Chr.5: 40852371-40852371 on Build GRCh38
rs781214387 Chr.5: 40852302-40852302 on Build GRCh38
rs369718011 Chr.5: 40852270-40852270 on Build GRCh38
rs764058090 Chr.5: 40852384-40852384 on Build GRCh38
rs747695580 Chr.5: 40852499-40852500 on Build GRCh38
rs957621194 Chr.5: 40852362-40852362 on Build GRCh38
rs780364291 Chr.5: 40852338-40852338 on Build GRCh38
rs749127157 Chr.5: 40852589-40852589 on Build GRCh38
rs148937373 Chr.5: 40852608-40852608 on Build GRCh38
rs561968147 Chr.5: 40852707-40852707 on Build GRCh38
rs761778578 Chr.5: 40852481-40852481 on Build GRCh38
rs373307405 Chr.5: 40852443-40852443 on Build GRCh38
rs780637265 Chr.5: 40852550-40852550 on Build GRCh38
rs928722893 Chr.5: 40852461-40852461 on Build GRCh38
rs558509923 Chr.5: 40852375-40852375 on Build GRCh38
rs188934 Chr.5: 40852349-40852349 on Build GRCh38
rs927375687 Chr.5: 40852667-40852667 on Build GRCh38
rs980605750 Chr.5: 40852659-40852659 on Build GRCh38
rs150175347 Chr.5: 40852599-40852599 on Build GRCh38
rs776373819 Chr.5: 40852357-40852357 on Build GRCh38
rs199835164 Chr.5: 40852708-40852708 on Build GRCh38
rs374736711 Chr.5: 40852419-40852419 on Build GRCh38

View Full Product Details