Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 41925156-41925648 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO4
Entrez Gene ID
26272
Gene Name
F-box protein 4
Gene Aliases
FBX4
UniGene
Hs.165575
Gene Chromosome Location
Chr.5: 41925249-42040230 on Build GRCh38
Transcripts
NM_001297437.1 NM_012176.2 NM_033484.2 XM_006714470.3 XM_011514026.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7367933 c.103T>G p.W35G Chr.5: 41925412-41925412 on Build GRCh38
5636610 c.172A>C p.T58P Chr.5: 41925481-41925481 on Build GRCh38
6365392 c.29C>A p.T10K Chr.5: 41925338-41925338 on Build GRCh38
7212258 c.70G>C p.E24Q Chr.5: 41925379-41925379 on Build GRCh38
6406037 c.158A>G p.D53G Chr.5: 41925467-41925467 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs546100978 Chr.5: 41925174-41925174 on Build GRCh38
rs2231914 Chr.5: 41925224-41925224 on Build GRCh38
rs1027893002 Chr.5: 41925578-41925578 on Build GRCh38
rs200832091 Chr.5: 41925536-41925536 on Build GRCh38
rs560779006 Chr.5: 41925186-41925186 on Build GRCh38
rs921911712 Chr.5: 41925184-41925184 on Build GRCh38
rs887966919 Chr.5: 41925517-41925517 on Build GRCh38
rs953348934 Chr.5: 41925618-41925618 on Build GRCh38
rs113955528 Chr.5: 41925243-41925243 on Build GRCh38
rs2231917 Chr.5: 41925333-41925333 on Build GRCh38
rs758711689 Chr.5: 41925213-41925213 on Build GRCh38
rs1053099322 Chr.5: 41925424-41925424 on Build GRCh38
rs774709021 Chr.5: 41925280-41925280 on Build GRCh38
rs376237718 Chr.5: 41925443-41925443 on Build GRCh38
rs1043639734 Chr.5: 41925380-41925380 on Build GRCh38
rs923834573 Chr.5: 41925411-41925411 on Build GRCh38
rs925093503 Chr.5: 41925419-41925419 on Build GRCh38
rs895873973 Chr.5: 41925238-41925238 on Build GRCh38
rs762437813 Chr.5: 41925331-41925331 on Build GRCh38
rs2231915 Chr.5: 41925254-41925254 on Build GRCh38
rs1005940946 Chr.5: 41925337-41925337 on Build GRCh38
rs997557508 Chr.5: 41925366-41925366 on Build GRCh38
rs567898431 Chr.5: 41925336-41925336 on Build GRCh38
rs961693227 Chr.5: 41925343-41925343 on Build GRCh38
rs1014714146 Chr.5: 41925455-41925455 on Build GRCh38
rs560742737 Chr.5: 41925594-41925594 on Build GRCh38
rs935908769 Chr.5: 41925609-41925609 on Build GRCh38
rs2231916 Chr.5: 41925329-41925329 on Build GRCh38
rs1001742180 Chr.5: 41925301-41925301 on Build GRCh38
rs953416559 Chr.5: 41925408-41925408 on Build GRCh38
rs1034927120 Chr.5: 41925324-41925324 on Build GRCh38
rs891815004 Chr.5: 41925256-41925256 on Build GRCh38
rs375482456 Chr.5: 41925377-41925377 on Build GRCh38
rs928940264 Chr.5: 41925503-41925503 on Build GRCh38
rs542749708 Chr.5: 41925582-41925582 on Build GRCh38
rs995024495 Chr.5: 41925269-41925269 on Build GRCh38
rs2231918 Chr.5: 41925531-41925531 on Build GRCh38
rs950962061 Chr.5: 41925603-41925603 on Build GRCh38
rs550564556 Chr.5: 41925233-41925233 on Build GRCh38
rs113745031 Chr.5: 41925339-41925339 on Build GRCh38
rs1053215565 Chr.5: 41925235-41925235 on Build GRCh38
rs976045591 Chr.5: 41925206-41925206 on Build GRCh38
rs897562178 Chr.5: 41925566-41925566 on Build GRCh38
rs781607246 Chr.5: 41925586-41925586 on Build GRCh38
rs568828861 Chr.5: 41925246-41925246 on Build GRCh38
rs946014306 Chr.5: 41925262-41925262 on Build GRCh38
rs751320866 Chr.5: 41925444-41925444 on Build GRCh38
rs763827758 Chr.5: 41925431-41925431 on Build GRCh38
rs564396919 Chr.5: 41925198-41925198 on Build GRCh38
rs937578493 Chr.5: 41925507-41925507 on Build GRCh38
rs950412272 Chr.5: 41925283-41925283 on Build GRCh38
rs957828560 Chr.5: 41925426-41925426 on Build GRCh38
rs946075051 Chr.5: 41925483-41925483 on Build GRCh38
rs757777710 Chr.5: 41925454-41925454 on Build GRCh38
rs907412997 Chr.5: 41925290-41925290 on Build GRCh38
rs961663260 Chr.5: 41925199-41925199 on Build GRCh38
rs921727198 Chr.5: 41925343-41925344 on Build GRCh38
rs545041200 Chr.5: 41925512-41925512 on Build GRCh38
rs765044998 Chr.5: 41925359-41925359 on Build GRCh38
rs1046136011 Chr.5: 41925490-41925490 on Build GRCh38
rs911349898 Chr.5: 41925245-41925245 on Build GRCh38
rs528356901 Chr.5: 41925188-41925188 on Build GRCh38
rs757232639 Chr.5: 41925446-41925446 on Build GRCh38
rs553344441 Chr.5: 41925374-41925374 on Build GRCh38
rs997231958 Chr.5: 41925590-41925590 on Build GRCh38
rs764724367 Chr.5: 41925414-41925414 on Build GRCh38
rs936051060 Chr.5: 41925252-41925252 on Build GRCh38
rs746013720 Chr.5: 41925497-41925497 on Build GRCh38
rs531968042 Chr.5: 41925227-41925227 on Build GRCh38
rs376927920 Chr.5: 41925286-41925286 on Build GRCh38
rs923142229 Chr.5: 41925207-41925207 on Build GRCh38

View Full Product Details