Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 54978505-54979045 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESM1
Entrez Gene ID
11082
Gene Name
endothelial cell specific molecule 1
Gene Aliases
endocan
UniGene
Hs.129944
Gene Chromosome Location
Chr.5: 54977867-54985586 on Build GRCh38
Transcripts
NM_001135604.1 NM_007036.4

SNP Detail

SNP ID Chromosome Location
rs934981973 Chr.5: 54978948-54978948 on Build GRCh38
rs1030511849 Chr.5: 54978838-54978839 on Build GRCh38
rs933503028 Chr.5: 54979002-54979002 on Build GRCh38
rs1006852702 Chr.5: 54978695-54978695 on Build GRCh38
rs573849639 Chr.5: 54978599-54978599 on Build GRCh38
rs992072366 Chr.5: 54978555-54978555 on Build GRCh38
rs955031164 Chr.5: 54978839-54978839 on Build GRCh38
rs916496291 Chr.5: 54978560-54978560 on Build GRCh38
rs143086419 Chr.5: 54978666-54978666 on Build GRCh38
rs924979877 Chr.5: 54978922-54978922 on Build GRCh38
rs865988327 Chr.5: 54978800-54978800 on Build GRCh38
rs149051390 Chr.5: 54978596-54978596 on Build GRCh38
rs960568153 Chr.5: 54978763-54978763 on Build GRCh38
rs757564921 Chr.5: 54978646-54978646 on Build GRCh38
rs186850443 Chr.5: 54978627-54978627 on Build GRCh38
rs904225790 Chr.5: 54978798-54978798 on Build GRCh38
rs779541596 Chr.5: 54978833-54978834 on Build GRCh38
rs59091910 Chr.5: 54978844-54978845 on Build GRCh38
rs948011215 Chr.5: 54978868-54978868 on Build GRCh38
rs201449172 Chr.5: 54978829-54978833 on Build GRCh38
rs901573739 Chr.5: 54978976-54978976 on Build GRCh38
rs905192671 Chr.5: 54978808-54978808 on Build GRCh38
rs1056950258 Chr.5: 54979001-54979001 on Build GRCh38
rs200684666 Chr.5: 54978833-54978833 on Build GRCh38
rs148346079 Chr.5: 54978806-54978806 on Build GRCh38
rs997680644 Chr.5: 54978654-54978654 on Build GRCh38
rs988367774 Chr.5: 54978835-54978835 on Build GRCh38
rs929245875 Chr.5: 54978918-54978918 on Build GRCh38
rs572434254 Chr.5: 54978537-54978537 on Build GRCh38
rs1006478042 Chr.5: 54979023-54979023 on Build GRCh38
rs559831352 Chr.5: 54978910-54978910 on Build GRCh38
rs956944506 Chr.5: 54978804-54978804 on Build GRCh38
rs143241193 Chr.5: 54978858-54978858 on Build GRCh38
rs35235179 Chr.5: 54978828-54978829 on Build GRCh38
rs929191934 Chr.5: 54978673-54978673 on Build GRCh38
rs532989765 Chr.5: 54978824-54978824 on Build GRCh38
rs542912728 Chr.5: 54978957-54978957 on Build GRCh38
rs988315241 Chr.5: 54978551-54978551 on Build GRCh38
rs191724249 Chr.5: 54978803-54978803 on Build GRCh38
rs182617929 Chr.5: 54978569-54978569 on Build GRCh38
rs919221099 Chr.5: 54978912-54978912 on Build GRCh38
rs1052149554 Chr.5: 54978617-54978617 on Build GRCh38
rs889258349 Chr.5: 54979020-54979020 on Build GRCh38
rs903493014 Chr.5: 54978585-54978585 on Build GRCh38
rs10557410 Chr.5: 54978840-54978844 on Build GRCh38
rs147113188 Chr.5: 54978955-54978955 on Build GRCh38
rs878913459 Chr.5: 54978829-54978829 on Build GRCh38
rs1016287583 Chr.5: 54978696-54978696 on Build GRCh38
rs962317092 Chr.5: 54978703-54978703 on Build GRCh38
rs767347760 Chr.5: 54978674-54978674 on Build GRCh38
rs1047439 Chr.5: 54978848-54978848 on Build GRCh38
rs1037621961 Chr.5: 54978626-54978626 on Build GRCh38
rs751034981 Chr.5: 54978637-54978637 on Build GRCh38
rs138587249 Chr.5: 54979010-54979010 on Build GRCh38
rs910625669 Chr.5: 54978850-54978850 on Build GRCh38
rs113720845 Chr.5: 54978643-54978643 on Build GRCh38
rs1035730856 Chr.5: 54978747-54978747 on Build GRCh38
rs1043386102 Chr.5: 54978576-54978576 on Build GRCh38
rs942008440 Chr.5: 54978618-54978618 on Build GRCh38
rs879063716 Chr.5: 54978826-54978826 on Build GRCh38
rs956890010 Chr.5: 54978532-54978532 on Build GRCh38
rs769288467 Chr.5: 54978894-54978894 on Build GRCh38
rs972183982 Chr.5: 54978716-54978716 on Build GRCh38

View Full Product Details