Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 54979336-54979850 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESM1
Entrez Gene ID
11082
Gene Name
endothelial cell specific molecule 1
Gene Aliases
endocan
UniGene
Hs.129944
Gene Chromosome Location
Chr.5: 54977867-54985586 on Build GRCh38
Transcripts
NM_001135604.1 NM_007036.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5403613 c.531G>A p.M177I Chr.5: 54979356-54979356 on Build GRCh38
4498611 c.523C>T p.P175S Chr.5: 54979364-54979364 on Build GRCh38
1068768 c.458A>T p.D153V Chr.5: 54979429-54979429 on Build GRCh38
1068767 c.490G>T p.E164* Chr.5: 54979397-54979397 on Build GRCh38
6332850 c.550C>A p.R184S Chr.5: 54979337-54979337 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761450922 Chr.5: 54979775-54979775 on Build GRCh38
rs576725927 Chr.5: 54979391-54979393 on Build GRCh38
rs537064807 Chr.5: 54979611-54979611 on Build GRCh38
rs769525291 Chr.5: 54979607-54979607 on Build GRCh38
rs569922861 Chr.5: 54979576-54979576 on Build GRCh38
rs112350718 Chr.5: 54979371-54979371 on Build GRCh38
rs555999817 Chr.5: 54979627-54979627 on Build GRCh38
rs572240917 Chr.5: 54979816-54979816 on Build GRCh38
rs919405200 Chr.5: 54979579-54979579 on Build GRCh38
rs148664278 Chr.5: 54979663-54979663 on Build GRCh38
rs1030247609 Chr.5: 54979750-54979750 on Build GRCh38
rs12517586 Chr.5: 54979518-54979518 on Build GRCh38
rs1032925637 Chr.5: 54979777-54979777 on Build GRCh38
rs986185181 Chr.5: 54979793-54979793 on Build GRCh38
rs370994211 Chr.5: 54979474-54979474 on Build GRCh38
rs933722726 Chr.5: 54979630-54979630 on Build GRCh38
rs762980287 Chr.5: 54979404-54979404 on Build GRCh38
rs200811457 Chr.5: 54979449-54979449 on Build GRCh38
rs773897568 Chr.5: 54979716-54979716 on Build GRCh38
rs756307716 Chr.5: 54979443-54979443 on Build GRCh38
rs904591347 Chr.5: 54979689-54979689 on Build GRCh38
rs193192856 Chr.5: 54979461-54979461 on Build GRCh38
rs963471781 Chr.5: 54979825-54979825 on Build GRCh38
rs1009496491 Chr.5: 54979749-54979749 on Build GRCh38
rs1039276126 Chr.5: 54979675-54979675 on Build GRCh38
rs768851686 Chr.5: 54979364-54979364 on Build GRCh38
rs368314976 Chr.5: 54979391-54979391 on Build GRCh38
rs1000254501 Chr.5: 54979692-54979692 on Build GRCh38
rs970069834 Chr.5: 54979754-54979754 on Build GRCh38
rs372901161 Chr.5: 54979425-54979425 on Build GRCh38
rs988650996 Chr.5: 54979812-54979812 on Build GRCh38
rs1057281688 Chr.5: 54979623-54979623 on Build GRCh38
rs748615805 Chr.5: 54979452-54979452 on Build GRCh38
rs555074677 Chr.5: 54979535-54979535 on Build GRCh38
rs1032287744 Chr.5: 54979424-54979424 on Build GRCh38
rs767918445 Chr.5: 54979426-54979426 on Build GRCh38
rs1026405091 Chr.5: 54979715-54979715 on Build GRCh38
rs774807270 Chr.5: 54979368-54979368 on Build GRCh38
rs775179767 Chr.5: 54979642-54979642 on Build GRCh38
rs1051205054 Chr.5: 54979636-54979636 on Build GRCh38
rs138338667 Chr.5: 54979390-54979390 on Build GRCh38
rs994912813 Chr.5: 54979422-54979422 on Build GRCh38
rs867211480 Chr.5: 54979811-54979811 on Build GRCh38
rs750495600 Chr.5: 54979427-54979427 on Build GRCh38
rs113805621 Chr.5: 54979554-54979554 on Build GRCh38
rs956486151 Chr.5: 54979745-54979745 on Build GRCh38
rs530429117 Chr.5: 54979548-54979549 on Build GRCh38
rs942236318 Chr.5: 54979557-54979557 on Build GRCh38
rs143424877 Chr.5: 54979680-54979680 on Build GRCh38
rs929579763 Chr.5: 54979600-54979600 on Build GRCh38
rs770631310 Chr.5: 54979370-54979370 on Build GRCh38
rs531355804 Chr.5: 54979362-54979362 on Build GRCh38
rs950760667 Chr.5: 54979429-54979429 on Build GRCh38
rs1032607527 Chr.5: 54979738-54979738 on Build GRCh38
rs571977543 Chr.5: 54979445-54979445 on Build GRCh38
rs375895378 Chr.5: 54979447-54979447 on Build GRCh38
rs375557331 Chr.5: 54979807-54979807 on Build GRCh38
rs143914015 Chr.5: 54979415-54979415 on Build GRCh38
rs550324980 Chr.5: 54979398-54979398 on Build GRCh38
rs762739188 Chr.5: 54979470-54979470 on Build GRCh38
rs1052895633 Chr.5: 54979584-54979584 on Build GRCh38
rs985913757 Chr.5: 54979508-54979508 on Build GRCh38
rs185200944 Chr.5: 54979683-54979683 on Build GRCh38
rs374654852 Chr.5: 54979448-54979448 on Build GRCh38
rs763731061 Chr.5: 54979405-54979405 on Build GRCh38
rs912978743 Chr.5: 54979613-54979613 on Build GRCh38
rs190435385 Chr.5: 54979818-54979818 on Build GRCh38
rs775072851 Chr.5: 54979463-54979463 on Build GRCh38
rs780871606 Chr.5: 54979460-54979460 on Build GRCh38
rs770543699 Chr.5: 54979457-54979457 on Build GRCh38

View Full Product Details