Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 55746055-55746563 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX4
Entrez Gene ID
54514
Gene Name
DEAD-box helicase 4
Gene Aliases
VASA
UniGene
Hs.223581
Gene Chromosome Location
Chr.5: 55737994-55817146 on Build GRCh38
Transcripts
NM_001142549.1 NM_001166533.1 NM_024415.2 XM_011543495.2 XM_011543497.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6428628 c.92G>T p.G31V Chr.5: 55746186-55746186 on Build GRCh38
5749231 c.122C>T p.S41L Chr.5: 55746216-55746216 on Build GRCh38
5403647 c.83G>A p.G28E Chr.5: 55746177-55746177 on Build GRCh38
5975630 c.111T>C p.T37T Chr.5: 55746205-55746205 on Build GRCh38
233926 c.70-9T>G p.? Chr.5: 55746155-55746155 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs66846694 Chr.5: 55746115-55746115 on Build GRCh38
rs141308424 Chr.5: 55746205-55746205 on Build GRCh38
rs532526521 Chr.5: 55746114-55746114 on Build GRCh38
rs992678315 Chr.5: 55746518-55746518 on Build GRCh38
rs141231413 Chr.5: 55746389-55746389 on Build GRCh38
rs1012418666 Chr.5: 55746309-55746309 on Build GRCh38
rs756857832 Chr.5: 55746139-55746140 on Build GRCh38
rs777552854 Chr.5: 55746497-55746498 on Build GRCh38
rs376529344 Chr.5: 55746240-55746240 on Build GRCh38
rs745321927 Chr.5: 55746233-55746233 on Build GRCh38
rs748413698 Chr.5: 55746171-55746171 on Build GRCh38
rs761754206 Chr.5: 55746273-55746273 on Build GRCh38
rs747949150 Chr.5: 55746489-55746489 on Build GRCh38
rs1007227916 Chr.5: 55746361-55746361 on Build GRCh38
rs777288263 Chr.5: 55746200-55746200 on Build GRCh38
rs888880906 Chr.5: 55746360-55746360 on Build GRCh38
rs144225002 Chr.5: 55746119-55746119 on Build GRCh38
rs192699620 Chr.5: 55746424-55746424 on Build GRCh38
rs776774010 Chr.5: 55746536-55746536 on Build GRCh38
rs552617330 Chr.5: 55746165-55746165 on Build GRCh38
rs775432520 Chr.5: 55746249-55746249 on Build GRCh38
rs1016343816 Chr.5: 55746365-55746366 on Build GRCh38
rs73127426 Chr.5: 55746335-55746335 on Build GRCh38
rs147762531 Chr.5: 55746334-55746334 on Build GRCh38
rs548259830 Chr.5: 55746441-55746441 on Build GRCh38
rs554874546 Chr.5: 55746532-55746532 on Build GRCh38
rs751608066 Chr.5: 55746148-55746148 on Build GRCh38
rs759393265 Chr.5: 55746234-55746234 on Build GRCh38
rs771635664 Chr.5: 55746185-55746185 on Build GRCh38
rs565167705 Chr.5: 55746083-55746083 on Build GRCh38
rs146565989 Chr.5: 55746183-55746183 on Build GRCh38
rs770871592 Chr.5: 55746219-55746219 on Build GRCh38
rs751695480 Chr.5: 55746270-55746270 on Build GRCh38
rs753028554 Chr.5: 55746124-55746124 on Build GRCh38
rs1022581373 Chr.5: 55746318-55746318 on Build GRCh38
rs769316385 Chr.5: 55746236-55746236 on Build GRCh38
rs901368507 Chr.5: 55746287-55746287 on Build GRCh38
rs750761578 Chr.5: 55746153-55746153 on Build GRCh38
rs776759889 Chr.5: 55746228-55746228 on Build GRCh38
rs199859572 Chr.5: 55746264-55746264 on Build GRCh38
rs371211865 Chr.5: 55746508-55746509 on Build GRCh38
rs959552369 Chr.5: 55746297-55746297 on Build GRCh38
rs35230239 Chr.5: 55746366-55746366 on Build GRCh38
rs562412646 Chr.5: 55746355-55746355 on Build GRCh38
rs537061129 Chr.5: 55746494-55746495 on Build GRCh38
rs753586401 Chr.5: 55746140-55746146 on Build GRCh38
rs779693434 Chr.5: 55746102-55746102 on Build GRCh38
rs778530167 Chr.5: 55746177-55746177 on Build GRCh38
rs992940475 Chr.5: 55746173-55746173 on Build GRCh38
rs200278733 Chr.5: 55746217-55746217 on Build GRCh38
rs777812202 Chr.5: 55746265-55746265 on Build GRCh38
rs778492206 Chr.5: 55746143-55746146 on Build GRCh38
rs778942657 Chr.5: 55746150-55746150 on Build GRCh38
rs993854616 Chr.5: 55746392-55746392 on Build GRCh38
rs184171613 Chr.5: 55746137-55746137 on Build GRCh38
rs897240055 Chr.5: 55746232-55746232 on Build GRCh38
rs977790470 Chr.5: 55746357-55746357 on Build GRCh38
rs187345413 Chr.5: 55746151-55746151 on Build GRCh38
rs923581653 Chr.5: 55746372-55746372 on Build GRCh38
rs868584994 Chr.5: 55746209-55746209 on Build GRCh38
rs755498553 Chr.5: 55746495-55746497 on Build GRCh38
rs998292292 Chr.5: 55746329-55746329 on Build GRCh38
rs570941314 Chr.5: 55746351-55746351 on Build GRCh38
rs756525934 Chr.5: 55746168-55746168 on Build GRCh38

View Full Product Details