Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 55766614-55767109 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX4
Entrez Gene ID
54514
Gene Name
DEAD-box helicase 4
Gene Aliases
VASA
UniGene
Hs.223581
Gene Chromosome Location
Chr.5: 55737994-55817146 on Build GRCh38
Transcripts
NM_001142549.1 NM_001166533.1 NM_001166534.1 NM_024415.2 XM_011543495.2 XM_011543497.2

SNP Detail

SNP ID Chromosome Location
rs1039138207 Chr.5: 55767010-55767010 on Build GRCh38
rs186381998 Chr.5: 55766792-55766792 on Build GRCh38
rs528423840 Chr.5: 55766801-55766801 on Build GRCh38
rs143970375 Chr.5: 55766808-55766808 on Build GRCh38
rs921616755 Chr.5: 55767018-55767018 on Build GRCh38
rs777351847 Chr.5: 55766990-55766990 on Build GRCh38
rs967042492 Chr.5: 55766764-55766764 on Build GRCh38
rs1036345541 Chr.5: 55766942-55766942 on Build GRCh38
rs913416948 Chr.5: 55766688-55766688 on Build GRCh38
rs35248526 Chr.5: 55766928-55766928 on Build GRCh38
rs894628459 Chr.5: 55766716-55766716 on Build GRCh38
rs1036420668 Chr.5: 55766675-55766675 on Build GRCh38
rs10035707 Chr.5: 55766695-55766695 on Build GRCh38
rs967955580 Chr.5: 55766989-55766989 on Build GRCh38
rs796822283 Chr.5: 55766932-55766932 on Build GRCh38
rs13181170 Chr.5: 55766936-55766936 on Build GRCh38
rs566118415 Chr.5: 55767016-55767016 on Build GRCh38
rs529095926 Chr.5: 55766863-55766863 on Build GRCh38
rs761235026 Chr.5: 55767067-55767067 on Build GRCh38
rs369956247 Chr.5: 55766894-55766894 on Build GRCh38
rs376374723 Chr.5: 55766809-55766809 on Build GRCh38
rs1048975435 Chr.5: 55767064-55767064 on Build GRCh38
rs946172165 Chr.5: 55766689-55766689 on Build GRCh38
rs534264585 Chr.5: 55767019-55767019 on Build GRCh38
rs928614317 Chr.5: 55766988-55766988 on Build GRCh38
rs190877084 Chr.5: 55766700-55766700 on Build GRCh38
rs774845268 Chr.5: 55767015-55767015 on Build GRCh38
rs999088225 Chr.5: 55767086-55767086 on Build GRCh38
rs1021846720 Chr.5: 55766753-55766753 on Build GRCh38
rs776703812 Chr.5: 55767076-55767077 on Build GRCh38
rs539132636 Chr.5: 55766958-55766958 on Build GRCh38
rs551384843 Chr.5: 55767002-55767002 on Build GRCh38
rs545398727 Chr.5: 55766785-55766785 on Build GRCh38
rs903410642 Chr.5: 55767078-55767078 on Build GRCh38
rs182632683 Chr.5: 55766720-55766720 on Build GRCh38
rs944401975 Chr.5: 55766712-55766712 on Build GRCh38
rs12652234 Chr.5: 55767087-55767087 on Build GRCh38
rs998492594 Chr.5: 55766771-55766771 on Build GRCh38
rs754627852 Chr.5: 55766680-55766681 on Build GRCh38
rs984012081 Chr.5: 55766914-55766914 on Build GRCh38
rs555639428 Chr.5: 55767044-55767044 on Build GRCh38
rs911667465 Chr.5: 55766998-55766998 on Build GRCh38
rs563001270 Chr.5: 55766715-55766715 on Build GRCh38
rs776011709 Chr.5: 55766954-55766954 on Build GRCh38
rs894646137 Chr.5: 55766994-55766994 on Build GRCh38
rs191587813 Chr.5: 55766819-55766819 on Build GRCh38
rs746657230 Chr.5: 55767037-55767037 on Build GRCh38
rs1056280049 Chr.5: 55766991-55766991 on Build GRCh38
rs965180144 Chr.5: 55766806-55766806 on Build GRCh38
rs907262608 Chr.5: 55766678-55766678 on Build GRCh38
rs1043340753 Chr.5: 55767052-55767052 on Build GRCh38
rs900449728 Chr.5: 55766721-55766722 on Build GRCh38
rs890176919 Chr.5: 55766697-55766697 on Build GRCh38
rs1027499166 Chr.5: 55766791-55766791 on Build GRCh38
rs971503519 Chr.5: 55766845-55766845 on Build GRCh38

View Full Product Details