Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 96675419-96675921 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAST
Entrez Gene ID
831
Gene Name
calpastatin
Gene Aliases
BS-17, PLACK
UniGene
Hs.436186
Gene Chromosome Location
Chr.5: 96662037-96774683 on Build GRCh38
Transcripts
NM_001042440.3 NM_001190442.1 XM_006714696.3 XM_006714697.3 XM_006714698.3 XM_006714699.3 XM_006714700.3 XM_006714701.3 XM_006714702.3 XM_006714703.3 XM_006714704.3 XM_006714705.3 XM_006714706.3 XM_011543654.2 XM_011543655.2 XM_011543656.2 XM_011543657.2 XM_017009911.1 XM_017009912.1 XM_017009913.1 XM_017009914.1 XM_017009915.1 XM_017009916.1 XM_017009917.1 XM_017009918.1 XM_017009919.1 XM_017009920.1 XM_017009921.1 XM_017009922.1 XM_017009923.1 XM_017009924.1 XM_017009925.1 XM_017009926.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3618874 c.92C>A p.T31N Chr.5: 96675555-96675555 on Build GRCh38
5611890 c.131C>T p.S44L Chr.5: 96675594-96675594 on Build GRCh38
6172370 c.123G>C p.K41N Chr.5: 96675586-96675586 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs996963286 Chr.5: 96675661-96675661 on Build GRCh38
rs756670389 Chr.5: 96675521-96675521 on Build GRCh38
rs565081258 Chr.5: 96675891-96675891 on Build GRCh38
rs754311845 Chr.5: 96675609-96675609 on Build GRCh38
rs956065421 Chr.5: 96675768-96675768 on Build GRCh38
rs73774372 Chr.5: 96675706-96675706 on Build GRCh38
rs998794091 Chr.5: 96675695-96675695 on Build GRCh38
rs866404737 Chr.5: 96675857-96675857 on Build GRCh38
rs752061522 Chr.5: 96675569-96675571 on Build GRCh38
rs758356777 Chr.5: 96675643-96675643 on Build GRCh38
rs980218375 Chr.5: 96675475-96675475 on Build GRCh38
rs750625444 Chr.5: 96675599-96675599 on Build GRCh38
rs759506173 Chr.5: 96675573-96675573 on Build GRCh38
rs376842478 Chr.5: 96675571-96675571 on Build GRCh38
rs544446230 Chr.5: 96675839-96675839 on Build GRCh38
rs757850347 Chr.5: 96675595-96675595 on Build GRCh38
rs771808417 Chr.5: 96675561-96675561 on Build GRCh38
rs767682372 Chr.5: 96675595-96675595 on Build GRCh38
rs181141746 Chr.5: 96675536-96675536 on Build GRCh38
rs1041248906 Chr.5: 96675624-96675624 on Build GRCh38
rs750749746 Chr.5: 96675502-96675502 on Build GRCh38
rs536094736 Chr.5: 96675772-96675772 on Build GRCh38
rs574816726 Chr.5: 96675577-96675577 on Build GRCh38
rs917358653 Chr.5: 96675773-96675774 on Build GRCh38
rs920554404 Chr.5: 96675501-96675501 on Build GRCh38
rs576848325 Chr.5: 96675803-96675803 on Build GRCh38
rs763376823 Chr.5: 96675529-96675529 on Build GRCh38
rs779579585 Chr.5: 96675734-96675734 on Build GRCh38
rs920707656 Chr.5: 96675840-96675840 on Build GRCh38
rs776200753 Chr.5: 96675871-96675871 on Build GRCh38
rs746669665 Chr.5: 96675638-96675638 on Build GRCh38
rs143938202 Chr.5: 96675774-96675774 on Build GRCh38
rs778505100 Chr.5: 96675555-96675555 on Build GRCh38
rs867545611 Chr.5: 96675778-96675778 on Build GRCh38
rs555852975 Chr.5: 96675819-96675819 on Build GRCh38
rs535717874 Chr.5: 96675601-96675601 on Build GRCh38
rs751838122 Chr.5: 96675491-96675491 on Build GRCh38
rs1048329985 Chr.5: 96675546-96675546 on Build GRCh38
rs535238735 Chr.5: 96675858-96675858 on Build GRCh38
rs757958682 Chr.5: 96675623-96675623 on Build GRCh38
rs556319858 Chr.5: 96675556-96675556 on Build GRCh38
rs572583933 Chr.5: 96675890-96675890 on Build GRCh38
rs965562284 Chr.5: 96675691-96675691 on Build GRCh38
rs73774371 Chr.5: 96675698-96675698 on Build GRCh38
rs905146396 Chr.5: 96675503-96675503 on Build GRCh38
rs764583175 Chr.5: 96675548-96675549 on Build GRCh38
rs752204947 Chr.5: 96675622-96675622 on Build GRCh38
rs778596786 Chr.5: 96675620-96675620 on Build GRCh38
rs752910556 Chr.5: 96675527-96675527 on Build GRCh38
rs774351513 Chr.5: 96675582-96675582 on Build GRCh38
rs754501776 Chr.5: 96675541-96675541 on Build GRCh38
rs978997193 Chr.5: 96675750-96675750 on Build GRCh38
rs796625580 Chr.5: 96675454-96675454 on Build GRCh38
rs777608906 Chr.5: 96675632-96675632 on Build GRCh38
rs888094041 Chr.5: 96675845-96675845 on Build GRCh38
rs12522566 Chr.5: 96675447-96675447 on Build GRCh38
rs777733239 Chr.5: 96675564-96675564 on Build GRCh38
rs755443924 Chr.5: 96675615-96675615 on Build GRCh38
rs1031863469 Chr.5: 96675755-96675755 on Build GRCh38
rs746828115 Chr.5: 96675565-96675565 on Build GRCh38
rs930613437 Chr.5: 96675846-96675846 on Build GRCh38
rs773703549 Chr.5: 96675647-96675647 on Build GRCh38
rs762055986 Chr.5: 96675588-96675588 on Build GRCh38
rs1015373803 Chr.5: 96675676-96675676 on Build GRCh38
rs780461725 Chr.5: 96675523-96675523 on Build GRCh38
rs926482473 Chr.5: 96675780-96675780 on Build GRCh38

View Full Product Details