Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 103096437-103096941 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIN1
Entrez Gene ID
54826
Gene Name
gypsy retrotransposon integrase 1
Gene Aliases
GIN-1, TGIN1, ZH2C2
UniGene
Hs.24088
Gene Chromosome Location
Chr.5: 103086000-103120149 on Build GRCh38
Transcripts
NM_001317954.1 NM_017676.2 NR_134299.1 XM_005272028.3 XM_017009599.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7330562 c.1228delG p.V410fs*3 Chr.5: 103096607-103096607 on Build GRCh38
1567190 c.1209C>A p.V403V Chr.5: 103096626-103096626 on Build GRCh38
1541367 c.1212G>A p.L404L Chr.5: 103096623-103096623 on Build GRCh38
370892 c.1204G>A p.A402T Chr.5: 103096631-103096631 on Build GRCh38
7226002 c.1186A>C p.I396L Chr.5: 103096649-103096649 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782722391 Chr.5: 103096685-103096685 on Build GRCh38
rs201966023 Chr.5: 103096833-103096833 on Build GRCh38
rs75209620 Chr.5: 103096494-103096494 on Build GRCh38
rs782528931 Chr.5: 103096814-103096814 on Build GRCh38
rs781925703 Chr.5: 103096497-103096497 on Build GRCh38
rs113924545 Chr.5: 103096800-103096800 on Build GRCh38
rs782796335 Chr.5: 103096727-103096727 on Build GRCh38
rs782412815 Chr.5: 103096834-103096834 on Build GRCh38
rs782640571 Chr.5: 103096815-103096815 on Build GRCh38
rs949369656 Chr.5: 103096527-103096527 on Build GRCh38
rs782731191 Chr.5: 103096533-103096533 on Build GRCh38
rs530042521 Chr.5: 103096662-103096662 on Build GRCh38
rs782578811 Chr.5: 103096616-103096616 on Build GRCh38
rs199707690 Chr.5: 103096651-103096651 on Build GRCh38
rs74391316 Chr.5: 103096673-103096673 on Build GRCh38
rs965565538 Chr.5: 103096805-103096805 on Build GRCh38
rs781815767 Chr.5: 103096543-103096543 on Build GRCh38
rs782534926 Chr.5: 103096575-103096575 on Build GRCh38
rs377378151 Chr.5: 103096465-103096465 on Build GRCh38
rs753391143 Chr.5: 103096761-103096761 on Build GRCh38
rs369908324 Chr.5: 103096690-103096690 on Build GRCh38
rs969355018 Chr.5: 103096871-103096871 on Build GRCh38
rs376587037 Chr.5: 103096658-103096658 on Build GRCh38
rs1036943521 Chr.5: 103096568-103096568 on Build GRCh38
rs782697665 Chr.5: 103096560-103096560 on Build GRCh38
rs782755512 Chr.5: 103096890-103096890 on Build GRCh38
rs765869524 Chr.5: 103096730-103096730 on Build GRCh38
rs782028018 Chr.5: 103096639-103096639 on Build GRCh38
rs782133161 Chr.5: 103096864-103096864 on Build GRCh38
rs782172246 Chr.5: 103096514-103096514 on Build GRCh38
rs565710282 Chr.5: 103096628-103096628 on Build GRCh38
rs782225893 Chr.5: 103096601-103096601 on Build GRCh38
rs200967173 Chr.5: 103096813-103096813 on Build GRCh38
rs782801620 Chr.5: 103096518-103096518 on Build GRCh38
rs782644119 Chr.5: 103096585-103096585 on Build GRCh38
rs782096229 Chr.5: 103096710-103096710 on Build GRCh38
rs781807062 Chr.5: 103096763-103096763 on Build GRCh38
rs782446898 Chr.5: 103096552-103096552 on Build GRCh38
rs782225594 Chr.5: 103096819-103096819 on Build GRCh38
rs782004536 Chr.5: 103096519-103096519 on Build GRCh38
rs782363848 Chr.5: 103096657-103096657 on Build GRCh38
rs754528315 Chr.5: 103096787-103096787 on Build GRCh38
rs778728014 Chr.5: 103096798-103096798 on Build GRCh38
rs374632190 Chr.5: 103096760-103096760 on Build GRCh38
rs371484594 Chr.5: 103096635-103096635 on Build GRCh38
rs782334728 Chr.5: 103096824-103096824 on Build GRCh38
rs782434947 Chr.5: 103096783-103096783 on Build GRCh38
rs781874430 Chr.5: 103096754-103096754 on Build GRCh38
rs573856435 Chr.5: 103096472-103096472 on Build GRCh38
rs782572366 Chr.5: 103096830-103096830 on Build GRCh38
rs782485549 Chr.5: 103096607-103096607 on Build GRCh38
rs782107369 Chr.5: 103096523-103096523 on Build GRCh38
rs782189444 Chr.5: 103096617-103096617 on Build GRCh38
rs782014083 Chr.5: 103096851-103096851 on Build GRCh38
rs782163658 Chr.5: 103096707-103096707 on Build GRCh38
rs781903264 Chr.5: 103096566-103096566 on Build GRCh38
rs782800384 Chr.5: 103096766-103096767 on Build GRCh38
rs782694815 Chr.5: 103096791-103096791 on Build GRCh38

View Full Product Details