Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 113001226-113001764 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCP2
Entrez Gene ID
167227
Gene Name
decapping mRNA 2
Gene Aliases
NUDT20
UniGene
Hs.443875
Gene Chromosome Location
Chr.5: 112976702-113022195 on Build GRCh38
Transcripts
NM_001242377.1 NM_152624.5 NR_038352.1 XM_017009163.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4676575 c.777G>A p.T259T Chr.5: 113001645-113001645 on Build GRCh38
4676574 c.672A>T p.K224N Chr.5: 113001443-113001443 on Build GRCh38
5816044 c.652C>A p.L218I Chr.5: 113001423-113001423 on Build GRCh38
4552927 c.575G>A p.R192K Chr.5: 113001226-113001226 on Build GRCh38
6529578 c.577G>C p.E193Q Chr.5: 113001228-113001228 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs188694255 Chr.5: 113001473-113001473 on Build GRCh38
rs760057225 Chr.5: 113001373-113001373 on Build GRCh38
rs758099700 Chr.5: 113001284-113001284 on Build GRCh38
rs200973942 Chr.5: 113001478-113001478 on Build GRCh38
rs749940019 Chr.5: 113001509-113001509 on Build GRCh38
rs747940211 Chr.5: 113001533-113001533 on Build GRCh38
rs764554877 Chr.5: 113001393-113001393 on Build GRCh38
rs754931764 Chr.5: 113001527-113001527 on Build GRCh38
rs780717459 Chr.5: 113001413-113001413 on Build GRCh38
rs766793855 Chr.5: 113001268-113001268 on Build GRCh38
rs751195051 Chr.5: 113001401-113001401 on Build GRCh38
rs772270550 Chr.5: 113001337-113001337 on Build GRCh38
rs370598360 Chr.5: 113001535-113001535 on Build GRCh38
rs760892590 Chr.5: 113001341-113001341 on Build GRCh38
rs776069879 Chr.5: 113001497-113001497 on Build GRCh38
rs148079095 Chr.5: 113001647-113001647 on Build GRCh38
rs991328600 Chr.5: 113001656-113001656 on Build GRCh38
rs770329753 Chr.5: 113001494-113001494 on Build GRCh38
rs752656460 Chr.5: 113001714-113001714 on Build GRCh38
rs370253943 Chr.5: 113001644-113001644 on Build GRCh38
rs989440611 Chr.5: 113001690-113001690 on Build GRCh38
rs373950381 Chr.5: 113001633-113001633 on Build GRCh38
rs777625022 Chr.5: 113001288-113001288 on Build GRCh38
rs377670849 Chr.5: 113001522-113001522 on Build GRCh38
rs773650442 Chr.5: 113001694-113001694 on Build GRCh38
rs760803146 Chr.5: 113001258-113001258 on Build GRCh38
rs756878494 Chr.5: 113001406-113001406 on Build GRCh38
rs771097845 Chr.5: 113001480-113001480 on Build GRCh38
rs753719085 Chr.5: 113001518-113001518 on Build GRCh38
rs766068517 Chr.5: 113001709-113001709 on Build GRCh38
rs756434131 Chr.5: 113001648-113001648 on Build GRCh38
rs764731248 Chr.5: 113001724-113001724 on Build GRCh38
rs901561034 Chr.5: 113001443-113001443 on Build GRCh38
rs759592279 Chr.5: 113001491-113001491 on Build GRCh38
rs752000414 Chr.5: 113001501-113001501 on Build GRCh38
rs913261069 Chr.5: 113001705-113001705 on Build GRCh38
rs781065228 Chr.5: 113001383-113001383 on Build GRCh38
rs753096232 Chr.5: 113001376-113001376 on Build GRCh38
rs778889207 Chr.5: 113001558-113001558 on Build GRCh38
rs759112678 Chr.5: 113001720-113001720 on Build GRCh38
rs760388579 Chr.5: 113001620-113001620 on Build GRCh38
rs781335428 Chr.5: 113001545-113001545 on Build GRCh38
rs373545100 Chr.5: 113001329-113001329 on Build GRCh38
rs376171167 Chr.5: 113001635-113001635 on Build GRCh38
rs946516933 Chr.5: 113001658-113001658 on Build GRCh38
rs755455187 Chr.5: 113001277-113001277 on Build GRCh38
rs754305101 Chr.5: 113001269-113001269 on Build GRCh38
rs143656592 Chr.5: 113001624-113001624 on Build GRCh38
rs755647535 Chr.5: 113001510-113001510 on Build GRCh38
rs148251063 Chr.5: 113001642-113001642 on Build GRCh38
rs573813285 Chr.5: 113001547-113001547 on Build GRCh38
rs112761672 Chr.5: 113001408-113001408 on Build GRCh38
rs947357650 Chr.5: 113001713-113001713 on Build GRCh38
rs889639685 Chr.5: 113001328-113001328 on Build GRCh38
rs35369053 Chr.5: 113001487-113001488 on Build GRCh38
rs749977813 Chr.5: 113001618-113001618 on Build GRCh38
rs376632643 Chr.5: 113001371-113001371 on Build GRCh38
rs780246693 Chr.5: 113001655-113001655 on Build GRCh38
rs756058426 Chr.5: 113001420-113001420 on Build GRCh38
rs765099711 Chr.5: 113001600-113001600 on Build GRCh38
rs756717478 Chr.5: 113001309-113001309 on Build GRCh38
rs758896820 Chr.5: 113001392-113001392 on Build GRCh38
rs778646654 Chr.5: 113001530-113001530 on Build GRCh38
rs745414532 Chr.5: 113001419-113001419 on Build GRCh38
rs766424495 Chr.5: 113001358-113001358 on Build GRCh38
rs762146807 Chr.5: 113001601-113001601 on Build GRCh38
rs769653680 Chr.5: 113001318-113001318 on Build GRCh38
rs767198913 Chr.5: 113001612-113001612 on Build GRCh38
rs745612382 Chr.5: 113001552-113001552 on Build GRCh38
rs202191634 Chr.5: 113001308-113001308 on Build GRCh38
rs115165195 Chr.5: 113001283-113001283 on Build GRCh38
rs775238310 Chr.5: 113001331-113001331 on Build GRCh38
rs934983913 Chr.5: 113001386-113001386 on Build GRCh38
rs1006675140 Chr.5: 113001414-113001414 on Build GRCh38
rs776103405 Chr.5: 113001714-113001714 on Build GRCh38
rs1043109801 Chr.5: 113001290-113001290 on Build GRCh38
rs778877529 Chr.5: 113001639-113001639 on Build GRCh38
rs772267282 Chr.5: 113001692-113001692 on Build GRCh38
rs767126196 Chr.5: 113001508-113001508 on Build GRCh38
rs777634554 Chr.5: 113001645-113001645 on Build GRCh38
rs902457072 Chr.5: 113001316-113001316 on Build GRCh38
rs776748142 Chr.5: 113001487-113001487 on Build GRCh38
rs555415112 Chr.5: 113001703-113001703 on Build GRCh38
rs749134044 Chr.5: 113001335-113001335 on Build GRCh38
rs774791530 Chr.5: 113001684-113001684 on Build GRCh38
rs778693558 Chr.5: 113001475-113001475 on Build GRCh38
rs780026913 Chr.5: 113001433-113001433 on Build GRCh38
rs748697305 Chr.5: 113001686-113001686 on Build GRCh38
rs1019523383 Chr.5: 113001436-113001436 on Build GRCh38
rs765710158 Chr.5: 113001375-113001375 on Build GRCh38
rs763511982 Chr.5: 113001561-113001561 on Build GRCh38
rs370145343 Chr.5: 113001310-113001310 on Build GRCh38
rs775732586 Chr.5: 113001554-113001554 on Build GRCh38
rs373708284 Chr.5: 113001657-113001657 on Build GRCh38
rs765596511 Chr.5: 113001279-113001279 on Build GRCh38
rs773246802 Chr.5: 113001628-113001628 on Build GRCh38
rs901096660 Chr.5: 113001493-113001493 on Build GRCh38
rs374071934 Chr.5: 113001507-113001507 on Build GRCh38
rs745766156 Chr.5: 113001311-113001311 on Build GRCh38
rs371167098 Chr.5: 113001725-113001725 on Build GRCh38
rs779544088 Chr.5: 113001514-113001514 on Build GRCh38
rs983898490 Chr.5: 113001526-113001527 on Build GRCh38
rs769932954 Chr.5: 113001553-113001553 on Build GRCh38
rs369984345 Chr.5: 113001442-113001442 on Build GRCh38
rs763563359 Chr.5: 113001500-113001500 on Build GRCh38
rs112399801 Chr.5: 113001670-113001670 on Build GRCh38
rs769330450 Chr.5: 113001566-113001566 on Build GRCh38

View Full Product Details