Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 113010619-113011143 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCP2
Entrez Gene ID
167227
Gene Name
decapping mRNA 2
Gene Aliases
NUDT20
UniGene
Hs.443875
Gene Chromosome Location
Chr.5: 112976702-113022195 on Build GRCh38
Transcripts
NM_001242377.1 NM_152624.5 NR_038352.1 XM_017009163.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1619329 c.1084G>T p.D362Y Chr.5: 113010792-113010792 on Build GRCh38
6640983 c.1059G>T p.K353N Chr.5: 113010767-113010767 on Build GRCh38
4159364 c.1099G>T p.D367Y Chr.5: 113010807-113010807 on Build GRCh38
6640981 c.1055A>G p.E352G Chr.5: 113010763-113010763 on Build GRCh38
7113148 c.1075A>G p.K359E Chr.5: 113010783-113010783 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765071962 Chr.5: 113010733-113010734 on Build GRCh38
rs998627084 Chr.5: 113011083-113011083 on Build GRCh38
rs754195128 Chr.5: 113010828-113010828 on Build GRCh38
rs909305117 Chr.5: 113010835-113010835 on Build GRCh38
rs767590721 Chr.5: 113010815-113010815 on Build GRCh38
rs1049800028 Chr.5: 113011095-113011095 on Build GRCh38
rs916351954 Chr.5: 113010944-113010944 on Build GRCh38
rs766730813 Chr.5: 113010824-113010824 on Build GRCh38
rs948930722 Chr.5: 113010674-113010674 on Build GRCh38
rs199826215 Chr.5: 113010743-113010743 on Build GRCh38
rs559258662 Chr.5: 113010733-113010733 on Build GRCh38
rs548674061 Chr.5: 113010735-113010736 on Build GRCh38
rs575408274 Chr.5: 113011009-113011009 on Build GRCh38
rs58980024 Chr.5: 113010735-113010735 on Build GRCh38
rs577088385 Chr.5: 113010668-113010668 on Build GRCh38
rs201023531 Chr.5: 113010720-113010721 on Build GRCh38
rs111240107 Chr.5: 113010733-113010733 on Build GRCh38
rs983962318 Chr.5: 113010889-113010889 on Build GRCh38
rs571556045 Chr.5: 113010663-113010663 on Build GRCh38
rs778305010 Chr.5: 113010734-113010735 on Build GRCh38
rs1006197594 Chr.5: 113011058-113011058 on Build GRCh38
rs750887122 Chr.5: 113010819-113010819 on Build GRCh38
rs1053220997 Chr.5: 113010652-113010652 on Build GRCh38
rs998213023 Chr.5: 113011034-113011034 on Build GRCh38
rs200824296 Chr.5: 113010737-113010737 on Build GRCh38
rs748926231 Chr.5: 113010739-113010739 on Build GRCh38
rs751570801 Chr.5: 113010736-113010738 on Build GRCh38
rs4705747 Chr.5: 113010940-113010940 on Build GRCh38
rs566139624 Chr.5: 113011028-113011028 on Build GRCh38
rs760093190 Chr.5: 113010733-113010735 on Build GRCh38
rs759309056 Chr.5: 113010734-113010737 on Build GRCh38
rs538848879 Chr.5: 113011013-113011014 on Build GRCh38
rs961300707 Chr.5: 113010915-113010915 on Build GRCh38
rs369747094 Chr.5: 113010848-113010848 on Build GRCh38
rs769052033 Chr.5: 113010719-113010720 on Build GRCh38
rs4705748 Chr.5: 113010941-113010941 on Build GRCh38
rs771390109 Chr.5: 113010720-113010723 on Build GRCh38
rs770710039 Chr.5: 113010778-113010778 on Build GRCh38
rs1053803904 Chr.5: 113011054-113011054 on Build GRCh38
rs1016820053 Chr.5: 113010910-113010910 on Build GRCh38
rs200465444 Chr.5: 113010749-113010749 on Build GRCh38
rs774398410 Chr.5: 113010789-113010789 on Build GRCh38
rs764345575 Chr.5: 113010785-113010785 on Build GRCh38
rs756987629 Chr.5: 113011050-113011050 on Build GRCh38
rs1043209982 Chr.5: 113010971-113010971 on Build GRCh38
rs768067516 Chr.5: 113010731-113010731 on Build GRCh38
rs776321450 Chr.5: 113010780-113010780 on Build GRCh38
rs550220199 Chr.5: 113010914-113010914 on Build GRCh38
rs180788798 Chr.5: 113011033-113011033 on Build GRCh38
rs752075643 Chr.5: 113010736-113010736 on Build GRCh38
rs61417979 Chr.5: 113010748-113010749 on Build GRCh38
rs763198678 Chr.5: 113010781-113010781 on Build GRCh38
rs374411821 Chr.5: 113010884-113010884 on Build GRCh38
rs757737184 Chr.5: 113010721-113010721 on Build GRCh38
rs554206356 Chr.5: 113011056-113011056 on Build GRCh38
rs1011774152 Chr.5: 113010665-113010665 on Build GRCh38
rs541348324 Chr.5: 113010719-113010719 on Build GRCh38
rs950741776 Chr.5: 113010791-113010791 on Build GRCh38
rs200950345 Chr.5: 113010841-113010841 on Build GRCh38
rs369850559 Chr.5: 113010776-113010776 on Build GRCh38
rs571874479 Chr.5: 113010924-113010924 on Build GRCh38
rs781669152 Chr.5: 113010917-113010917 on Build GRCh38
rs753508254 Chr.5: 113010734-113010736 on Build GRCh38
rs750309648 Chr.5: 113010722-113010723 on Build GRCh38
rs202051920 Chr.5: 113010750-113010750 on Build GRCh38
rs781377721 Chr.5: 113010711-113010712 on Build GRCh38
rs201721169 Chr.5: 113010822-113010822 on Build GRCh38
rs753421327 Chr.5: 113010982-113010982 on Build GRCh38
rs767401950 Chr.5: 113010736-113010737 on Build GRCh38
rs200180306 Chr.5: 113010718-113010719 on Build GRCh38
rs368095981 Chr.5: 113010859-113010859 on Build GRCh38
rs772123316 Chr.5: 113010751-113010751 on Build GRCh38
rs948857120 Chr.5: 113010954-113010954 on Build GRCh38
rs765185790 Chr.5: 113010716-113010719 on Build GRCh38
rs780300005 Chr.5: 113010738-113010738 on Build GRCh38
rs773215415 Chr.5: 113010757-113010757 on Build GRCh38
rs889875623 Chr.5: 113011097-113011097 on Build GRCh38
rs796621277 Chr.5: 113010736-113010737 on Build GRCh38
rs758288883 Chr.5: 113010736-113010736 on Build GRCh38
rs761945428 Chr.5: 113010811-113010811 on Build GRCh38
rs76293534 Chr.5: 113010735-113010735 on Build GRCh38

View Full Product Details