Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 113013242-113013746 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCP2
Entrez Gene ID
167227
Gene Name
decapping mRNA 2
Gene Aliases
NUDT20
UniGene
Hs.443875
Gene Chromosome Location
Chr.5: 112976702-113022195 on Build GRCh38
Transcripts
NM_001242377.1 NM_152624.5 NR_038352.1 XM_017009163.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4159365 c.1152T>C p.A384A Chr.5: 113013373-113013373 on Build GRCh38
7143680 c.1228G>A p.D410N Chr.5: 113013449-113013449 on Build GRCh38
5986967 c.1147C>T p.H383Y Chr.5: 113013368-113013368 on Build GRCh38
5451785 c.1193C>T p.P398L Chr.5: 113013414-113013414 on Build GRCh38
1059683 c.1151C>A p.A384D Chr.5: 113013372-113013372 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761214078 Chr.5: 113013334-113013334 on Build GRCh38
rs1008966392 Chr.5: 113013685-113013685 on Build GRCh38
rs561739767 Chr.5: 113013620-113013620 on Build GRCh38
rs773199862 Chr.5: 113013328-113013328 on Build GRCh38
rs199970082 Chr.5: 113013483-113013483 on Build GRCh38
rs979852627 Chr.5: 113013609-113013609 on Build GRCh38
rs138329876 Chr.5: 113013388-113013388 on Build GRCh38
rs370977840 Chr.5: 113013390-113013390 on Build GRCh38
rs774492120 Chr.5: 113013307-113013307 on Build GRCh38
rs771466472 Chr.5: 113013395-113013395 on Build GRCh38
rs772858074 Chr.5: 113013626-113013626 on Build GRCh38
rs1053957318 Chr.5: 113013641-113013641 on Build GRCh38
rs766899009 Chr.5: 113013498-113013498 on Build GRCh38
rs79000546 Chr.5: 113013385-113013385 on Build GRCh38
rs751073360 Chr.5: 113013415-113013415 on Build GRCh38
rs923599018 Chr.5: 113013593-113013593 on Build GRCh38
rs187263544 Chr.5: 113013718-113013718 on Build GRCh38
rs749631981 Chr.5: 113013534-113013537 on Build GRCh38
rs961468052 Chr.5: 113013396-113013396 on Build GRCh38
rs745807991 Chr.5: 113013374-113013374 on Build GRCh38
rs750093070 Chr.5: 113013683-113013683 on Build GRCh38
rs144451239 Chr.5: 113013461-113013461 on Build GRCh38
rs148286686 Chr.5: 113013283-113013283 on Build GRCh38
rs764522478 Chr.5: 113013493-113013493 on Build GRCh38
rs760084673 Chr.5: 113013480-113013480 on Build GRCh38
rs754218987 Chr.5: 113013338-113013338 on Build GRCh38
rs773964328 Chr.5: 113013443-113013443 on Build GRCh38
rs770809820 Chr.5: 113013279-113013279 on Build GRCh38
rs777927133 Chr.5: 113013470-113013470 on Build GRCh38
rs184122046 Chr.5: 113013397-113013397 on Build GRCh38
rs765655977 Chr.5: 113013403-113013403 on Build GRCh38
rs945358728 Chr.5: 113013662-113013662 on Build GRCh38
rs752347811 Chr.5: 113013358-113013358 on Build GRCh38
rs763708424 Chr.5: 113013414-113013414 on Build GRCh38
rs150445267 Chr.5: 113013386-113013386 on Build GRCh38
rs760008039 Chr.5: 113013561-113013561 on Build GRCh38
rs528679648 Chr.5: 113013445-113013445 on Build GRCh38
rs1010016943 Chr.5: 113013347-113013347 on Build GRCh38
rs13357043 Chr.5: 113013597-113013597 on Build GRCh38
rs761044827 Chr.5: 113013675-113013675 on Build GRCh38
rs756013447 Chr.5: 113013512-113013512 on Build GRCh38
rs777306575 Chr.5: 113013363-113013363 on Build GRCh38
rs374619019 Chr.5: 113013501-113013501 on Build GRCh38
rs770034347 Chr.5: 113013486-113013488 on Build GRCh38
rs766365644 Chr.5: 113013505-113013505 on Build GRCh38
rs767539520 Chr.5: 113013314-113013314 on Build GRCh38
rs190878661 Chr.5: 113013703-113013703 on Build GRCh38
rs747157076 Chr.5: 113013524-113013524 on Build GRCh38
rs369596242 Chr.5: 113013419-113013419 on Build GRCh38
rs558208044 Chr.5: 113013579-113013579 on Build GRCh38
rs200524232 Chr.5: 113013288-113013288 on Build GRCh38
rs900327466 Chr.5: 113013700-113013700 on Build GRCh38
rs745704541 Chr.5: 113013289-113013289 on Build GRCh38
rs182138466 Chr.5: 113013713-113013713 on Build GRCh38
rs758154928 Chr.5: 113013362-113013362 on Build GRCh38
rs756653389 Chr.5: 113013434-113013434 on Build GRCh38
rs774463566 Chr.5: 113013402-113013402 on Build GRCh38
rs866714126 Chr.5: 113013669-113013669 on Build GRCh38
rs185047058 Chr.5: 113013520-113013520 on Build GRCh38
rs775214517 Chr.5: 113013350-113013350 on Build GRCh38
rs4705749 Chr.5: 113013682-113013682 on Build GRCh38
rs749134380 Chr.5: 113013465-113013465 on Build GRCh38
rs17135261 Chr.5: 113013710-113013710 on Build GRCh38
rs780770495 Chr.5: 113013442-113013442 on Build GRCh38
rs760045840 Chr.5: 113013345-113013345 on Build GRCh38
rs756023141 Chr.5: 113013503-113013503 on Build GRCh38
rs991022718 Chr.5: 113013547-113013547 on Build GRCh38
rs762039402 Chr.5: 113013311-113013311 on Build GRCh38
rs770864919 Chr.5: 113013479-113013479 on Build GRCh38
rs769523264 Chr.5: 113013381-113013381 on Build GRCh38
rs569141365 Chr.5: 113013369-113013369 on Build GRCh38
rs761551459 Chr.5: 113013496-113013496 on Build GRCh38
rs1042442038 Chr.5: 113013352-113013352 on Build GRCh38
rs772328965 Chr.5: 113013469-113013469 on Build GRCh38
rs746925040 Chr.5: 113013278-113013278 on Build GRCh38
rs1039175127 Chr.5: 113013692-113013692 on Build GRCh38
rs773815539 Chr.5: 113013495-113013495 on Build GRCh38
rs568235501 Chr.5: 113013433-113013433 on Build GRCh38
rs747262797 Chr.5: 113013476-113013476 on Build GRCh38
rs149633478 Chr.5: 113013412-113013412 on Build GRCh38
rs146621442 Chr.5: 113013346-113013346 on Build GRCh38
rs376212582 Chr.5: 113013522-113013522 on Build GRCh38
rs540122362 Chr.5: 113013564-113013564 on Build GRCh38
rs997041224 Chr.5: 113013674-113013674 on Build GRCh38
rs551417149 Chr.5: 113013384-113013384 on Build GRCh38
rs1042077579 Chr.5: 113013589-113013589 on Build GRCh38
rs779850722 Chr.5: 113013382-113013382 on Build GRCh38
rs756079360 Chr.5: 113013457-113013457 on Build GRCh38
rs915487875 Chr.5: 113013643-113013643 on Build GRCh38
rs983523645 Chr.5: 113013595-113013595 on Build GRCh38
rs9326869 Chr.5: 113013373-113013373 on Build GRCh38
rs6892784 Chr.5: 113013600-113013600 on Build GRCh38
rs780329897 Chr.5: 113013516-113013517 on Build GRCh38
rs181023764 Chr.5: 113013453-113013453 on Build GRCh38
rs373156882 Chr.5: 113013301-113013301 on Build GRCh38
rs917859639 Chr.5: 113013548-113013548 on Build GRCh38
rs893804970 Chr.5: 113013306-113013306 on Build GRCh38
rs139117940 Chr.5: 113013367-113013367 on Build GRCh38
rs753629894 Chr.5: 113013509-113013509 on Build GRCh38
rs763477420 Chr.5: 113013488-113013488 on Build GRCh38

View Full Product Details