Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 115993350-115993864 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LVRN
Entrez Gene ID
206338
Gene Name
laeverin
Gene Aliases
APQ, AQPEP, TAQPEP
UniGene
Hs.556880
Gene Chromosome Location
Chr.5: 115962440-116027602 on Build GRCh38
Transcripts
NM_173800.4 XM_011543263.2 XM_011543264.2 XM_017009215.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7323934 c.1291T>C p.W431R Chr.5: 115993771-115993771 on Build GRCh38
3849658 c.1322C>T p.A441V Chr.5: 115993802-115993802 on Build GRCh38
5481913 c.1374+4A>G p.? Chr.5: 115993858-115993858 on Build GRCh38
4640612 c.1345A>T p.I449F Chr.5: 115993825-115993825 on Build GRCh38
4585336 c.1346T>G p.I449S Chr.5: 115993826-115993826 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749530311 Chr.5: 115993815-115993815 on Build GRCh38
rs774576393 Chr.5: 115993764-115993764 on Build GRCh38
rs143028971 Chr.5: 115993416-115993416 on Build GRCh38
rs372959461 Chr.5: 115993741-115993741 on Build GRCh38
rs758335122 Chr.5: 115993791-115993791 on Build GRCh38
rs778527550 Chr.5: 115993698-115993698 on Build GRCh38
rs953531137 Chr.5: 115993394-115993394 on Build GRCh38
rs189111760 Chr.5: 115993428-115993428 on Build GRCh38
rs752445666 Chr.5: 115993446-115993446 on Build GRCh38
rs909905044 Chr.5: 115993557-115993557 on Build GRCh38
rs750815707 Chr.5: 115993769-115993769 on Build GRCh38
rs186396448 Chr.5: 115993632-115993632 on Build GRCh38
rs769967045 Chr.5: 115993714-115993714 on Build GRCh38
rs772887506 Chr.5: 115993710-115993710 on Build GRCh38
rs965729232 Chr.5: 115993504-115993504 on Build GRCh38
rs1009463380 Chr.5: 115993397-115993397 on Build GRCh38
rs201368771 Chr.5: 115993790-115993790 on Build GRCh38
rs565492669 Chr.5: 115993653-115993653 on Build GRCh38
rs576335243 Chr.5: 115993623-115993623 on Build GRCh38
rs763059241 Chr.5: 115993727-115993727 on Build GRCh38
rs1043640822 Chr.5: 115993699-115993699 on Build GRCh38
rs939385605 Chr.5: 115993625-115993625 on Build GRCh38
rs369618913 Chr.5: 115993761-115993761 on Build GRCh38
rs776368407 Chr.5: 115993805-115993805 on Build GRCh38
rs10478268 Chr.5: 115993596-115993596 on Build GRCh38
rs542909128 Chr.5: 115993490-115993490 on Build GRCh38
rs1033653978 Chr.5: 115993830-115993830 on Build GRCh38
rs756481757 Chr.5: 115993776-115993776 on Build GRCh38
rs751769551 Chr.5: 115993801-115993801 on Build GRCh38
rs887264793 Chr.5: 115993767-115993767 on Build GRCh38
rs573724440 Chr.5: 115993468-115993468 on Build GRCh38
rs913716423 Chr.5: 115993660-115993660 on Build GRCh38
rs1045962162 Chr.5: 115993487-115993487 on Build GRCh38
rs1011210337 Chr.5: 115993551-115993551 on Build GRCh38
rs145831521 Chr.5: 115993832-115993832 on Build GRCh38
rs903556555 Chr.5: 115993708-115993708 on Build GRCh38
rs373564784 Chr.5: 115993737-115993737 on Build GRCh38
rs926699986 Chr.5: 115993624-115993624 on Build GRCh38
rs894357719 Chr.5: 115993480-115993480 on Build GRCh38
rs770726349 Chr.5: 115993803-115993803 on Build GRCh38
rs751586483 Chr.5: 115993749-115993749 on Build GRCh38
rs369931696 Chr.5: 115993782-115993782 on Build GRCh38
rs768908849 Chr.5: 115993680-115993680 on Build GRCh38
rs912012667 Chr.5: 115993410-115993410 on Build GRCh38
rs945314689 Chr.5: 115993682-115993682 on Build GRCh38
rs555862000 Chr.5: 115993717-115993717 on Build GRCh38
rs746827971 Chr.5: 115993802-115993802 on Build GRCh38
rs951650313 Chr.5: 115993534-115993534 on Build GRCh38
rs1012786396 Chr.5: 115993481-115993481 on Build GRCh38
rs986699587 Chr.5: 115993404-115993404 on Build GRCh38
rs200401368 Chr.5: 115993745-115993745 on Build GRCh38
rs1029757422 Chr.5: 115993517-115993517 on Build GRCh38
rs1036387105 Chr.5: 115993436-115993436 on Build GRCh38
rs1048973875 Chr.5: 115993479-115993479 on Build GRCh38
rs1346478 Chr.5: 115993553-115993553 on Build GRCh38
rs373046941 Chr.5: 115993780-115993780 on Build GRCh38
rs553863237 Chr.5: 115993445-115993445 on Build GRCh38
rs763593930 Chr.5: 115993513-115993513 on Build GRCh38
rs748120208 Chr.5: 115993704-115993704 on Build GRCh38
rs532873623 Chr.5: 115993683-115993683 on Build GRCh38
rs1029809812 Chr.5: 115993762-115993762 on Build GRCh38
rs775665794 Chr.5: 115993715-115993715 on Build GRCh38
rs1049044387 Chr.5: 115993707-115993707 on Build GRCh38
rs778580648 Chr.5: 115993784-115993784 on Build GRCh38
rs371832728 Chr.5: 115993713-115993713 on Build GRCh38
rs181456850 Chr.5: 115993577-115993577 on Build GRCh38
rs934478783 Chr.5: 115993771-115993771 on Build GRCh38
rs962736687 Chr.5: 115993587-115993587 on Build GRCh38
rs777751245 Chr.5: 115993792-115993792 on Build GRCh38
rs972569863 Chr.5: 115993650-115993650 on Build GRCh38
rs865894827 Chr.5: 115993475-115993475 on Build GRCh38
rs999031013 Chr.5: 115993491-115993491 on Build GRCh38
rs747594884 Chr.5: 115993785-115993785 on Build GRCh38

View Full Product Details