Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 142875004-142875504 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP26
Entrez Gene ID
23092
Gene Name
Rho GTPase activating protein 26
Gene Aliases
GRAF, GRAF1, OPHN1L, OPHN1L1
UniGene
Hs.654668
Gene Chromosome Location
Chr.5: 142770370-143229007 on Build GRCh38
Transcripts
NM_001135608.1 NM_015071.4 XM_005268398.4 XM_005268399.4 XM_005268400.4 XM_005268402.4 XM_006714774.3 XM_011537610.2 XM_011537611.2 XM_011537612.2 XM_017009247.1 XM_017009248.1 XM_017009249.1 XM_017009250.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3611846 c.309G>A p.R103R Chr.5: 142875168-142875168 on Build GRCh38
7419056 c.308G>A p.R103Q Chr.5: 142875167-142875167 on Build GRCh38
5912184 c.251-4C>T p.? Chr.5: 142875106-142875106 on Build GRCh38
6295058 c.312+8G>A p.? Chr.5: 142875179-142875179 on Build GRCh38
7216828 c.255A>G p.R85R Chr.5: 142875114-142875114 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774671486 Chr.5: 142875132-142875132 on Build GRCh38
rs765897153 Chr.5: 142875200-142875200 on Build GRCh38
rs972916434 Chr.5: 142875397-142875397 on Build GRCh38
rs75621483 Chr.5: 142875418-142875418 on Build GRCh38
rs562548632 Chr.5: 142875029-142875029 on Build GRCh38
rs1044445703 Chr.5: 142875415-142875415 on Build GRCh38
rs920658997 Chr.5: 142875348-142875348 on Build GRCh38
rs944596215 Chr.5: 142875393-142875393 on Build GRCh38
rs370435398 Chr.5: 142875061-142875061 on Build GRCh38
rs1001262927 Chr.5: 142875299-142875299 on Build GRCh38
rs762486970 Chr.5: 142875190-142875190 on Build GRCh38
rs1056889597 Chr.5: 142875032-142875032 on Build GRCh38
rs1040326332 Chr.5: 142875034-142875035 on Build GRCh38
rs759537245 Chr.5: 142875064-142875065 on Build GRCh38
rs546855011 Chr.5: 142875469-142875469 on Build GRCh38
rs750334908 Chr.5: 142875185-142875185 on Build GRCh38
rs1047837203 Chr.5: 142875368-142875368 on Build GRCh38
rs753046222 Chr.5: 142875214-142875214 on Build GRCh38
rs770917906 Chr.5: 142875126-142875126 on Build GRCh38
rs917503890 Chr.5: 142875286-142875286 on Build GRCh38
rs927017409 Chr.5: 142875436-142875436 on Build GRCh38
rs1028361339 Chr.5: 142875317-142875317 on Build GRCh38
rs754522956 Chr.5: 142875371-142875371 on Build GRCh38
rs986844191 Chr.5: 142875363-142875363 on Build GRCh38
rs73286481 Chr.5: 142875333-142875333 on Build GRCh38
rs1032241284 Chr.5: 142875220-142875220 on Build GRCh38
rs761574373 Chr.5: 142875167-142875167 on Build GRCh38
rs571277033 Chr.5: 142875411-142875411 on Build GRCh38
rs533006130 Chr.5: 142875466-142875466 on Build GRCh38
rs920152821 Chr.5: 142875409-142875409 on Build GRCh38
rs758493286 Chr.5: 142875289-142875289 on Build GRCh38
rs751119809 Chr.5: 142875208-142875208 on Build GRCh38
rs900128198 Chr.5: 142875059-142875059 on Build GRCh38
rs778977295 Chr.5: 142875181-142875181 on Build GRCh38
rs745723600 Chr.5: 142875213-142875213 on Build GRCh38
rs995849580 Chr.5: 142875082-142875082 on Build GRCh38
rs961510484 Chr.5: 142875372-142875372 on Build GRCh38
rs370338871 Chr.5: 142875093-142875093 on Build GRCh38
rs529207979 Chr.5: 142875473-142875473 on Build GRCh38
rs568105184 Chr.5: 142875424-142875424 on Build GRCh38
rs1022024368 Chr.5: 142875101-142875101 on Build GRCh38
rs772467491 Chr.5: 142875156-142875156 on Build GRCh38
rs776566350 Chr.5: 142875161-142875161 on Build GRCh38
rs587778050 Chr.5: 142875166-142875166 on Build GRCh38
rs956264307 Chr.5: 142875223-142875223 on Build GRCh38
rs968689520 Chr.5: 142875292-142875292 on Build GRCh38
rs999304668 Chr.5: 142875201-142875201 on Build GRCh38
rs754593346 Chr.5: 142875210-142875210 on Build GRCh38
rs185200 Chr.5: 142875114-142875114 on Build GRCh38
rs1043394447 Chr.5: 142875426-142875426 on Build GRCh38
rs373317587 Chr.5: 142875135-142875135 on Build GRCh38
rs566614540 Chr.5: 142875470-142875470 on Build GRCh38
rs993483834 Chr.5: 142875276-142875276 on Build GRCh38
rs545142202 Chr.5: 142875298-142875298 on Build GRCh38

View Full Product Details