Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 142879295-142879805 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP26
Entrez Gene ID
23092
Gene Name
Rho GTPase activating protein 26
Gene Aliases
GRAF, GRAF1, OPHN1L, OPHN1L1
UniGene
Hs.654668
Gene Chromosome Location
Chr.5: 142770370-143229007 on Build GRCh38
Transcripts
NM_001135608.1 NM_015071.4 XM_005268398.4 XM_005268399.4 XM_005268400.4 XM_005268402.4 XM_006714774.3 XM_011537610.2 XM_011537611.2 XM_011537612.2 XM_017009247.1 XM_017009248.1 XM_017009249.1 XM_017009250.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6414759 c.361A>T p.K121* Chr.5: 142879422-142879422 on Build GRCh38
3611847 c.372C>T p.I124I Chr.5: 142879433-142879433 on Build GRCh38
1732238 c.359G>A p.R120Q Chr.5: 142879420-142879420 on Build GRCh38
6603327 c.318G>T p.E106D Chr.5: 142879379-142879379 on Build GRCh38
215964 c.358C>T p.R120* Chr.5: 142879419-142879419 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1012494924 Chr.5: 142879629-142879629 on Build GRCh38
rs956403684 Chr.5: 142879620-142879620 on Build GRCh38
rs746016092 Chr.5: 142879370-142879370 on Build GRCh38
rs539120690 Chr.5: 142879638-142879638 on Build GRCh38
rs990505669 Chr.5: 142879580-142879580 on Build GRCh38
rs758979435 Chr.5: 142879486-142879486 on Build GRCh38
rs752884065 Chr.5: 142879468-142879468 on Build GRCh38
rs1025708652 Chr.5: 142879744-142879744 on Build GRCh38
rs756459324 Chr.5: 142879430-142879430 on Build GRCh38
rs764384167 Chr.5: 142879432-142879432 on Build GRCh38
rs939499183 Chr.5: 142879609-142879609 on Build GRCh38
rs758560091 Chr.5: 142879444-142879444 on Build GRCh38
rs911081109 Chr.5: 142879551-142879551 on Build GRCh38
rs775871768 Chr.5: 142879374-142879374 on Build GRCh38
rs761604734 Chr.5: 142879378-142879378 on Build GRCh38
rs370969753 Chr.5: 142879450-142879450 on Build GRCh38
rs987484113 Chr.5: 142879653-142879653 on Build GRCh38
rs906792231 Chr.5: 142879751-142879751 on Build GRCh38
rs772422132 Chr.5: 142879433-142879433 on Build GRCh38
rs770990322 Chr.5: 142879362-142879362 on Build GRCh38
rs144661167 Chr.5: 142879388-142879388 on Build GRCh38
rs989603759 Chr.5: 142879621-142879621 on Build GRCh38
rs982210141 Chr.5: 142879368-142879368 on Build GRCh38
rs768277972 Chr.5: 142879434-142879434 on Build GRCh38
rs556542164 Chr.5: 142879679-142879679 on Build GRCh38
rs989325434 Chr.5: 142879424-142879424 on Build GRCh38
rs762777887 Chr.5: 142879389-142879389 on Build GRCh38
rs922498707 Chr.5: 142879651-142879651 on Build GRCh38
rs914239793 Chr.5: 142879582-142879582 on Build GRCh38
rs899304696 Chr.5: 142879536-142879536 on Build GRCh38
rs745887563 Chr.5: 142879440-142879440 on Build GRCh38
rs948211620 Chr.5: 142879730-142879730 on Build GRCh38
rs945706970 Chr.5: 142879585-142879585 on Build GRCh38
rs773845372 Chr.5: 142879402-142879402 on Build GRCh38
rs766051196 Chr.5: 142879666-142879666 on Build GRCh38
rs377365899 Chr.5: 142879753-142879753 on Build GRCh38
rs886515962 Chr.5: 142879514-142879514 on Build GRCh38
rs772852766 Chr.5: 142879466-142879466 on Build GRCh38
rs551013311 Chr.5: 142879421-142879421 on Build GRCh38
rs765458919 Chr.5: 142879466-142879467 on Build GRCh38
rs768971885 Chr.5: 142879463-142879463 on Build GRCh38
rs779626045 Chr.5: 142879437-142879437 on Build GRCh38
rs902651243 Chr.5: 142879375-142879375 on Build GRCh38
rs574079741 Chr.5: 142879636-142879636 on Build GRCh38
rs113632964 Chr.5: 142879396-142879396 on Build GRCh38
rs1056542145 Chr.5: 142879613-142879613 on Build GRCh38
rs557420971 Chr.5: 142879643-142879643 on Build GRCh38
rs775968945 Chr.5: 142879484-142879484 on Build GRCh38
rs374637673 Chr.5: 142879481-142879481 on Build GRCh38
rs749788598 Chr.5: 142879359-142879359 on Build GRCh38
rs1012381861 Chr.5: 142879545-142879545 on Build GRCh38
rs1037793302 Chr.5: 142879521-142879521 on Build GRCh38
rs762953809 Chr.5: 142879652-142879652 on Build GRCh38
rs116101547 Chr.5: 142879367-142879367 on Build GRCh38
rs27779 Chr.5: 142879518-142879518 on Build GRCh38
rs1052226524 Chr.5: 142879506-142879506 on Build GRCh38
rs368528114 Chr.5: 142879414-142879414 on Build GRCh38
rs928400706 Chr.5: 142879604-142879604 on Build GRCh38
rs140225755 Chr.5: 142879731-142879731 on Build GRCh38
rs772414264 Chr.5: 142879371-142879371 on Build GRCh38
rs189174774 Chr.5: 142879756-142879756 on Build GRCh38
rs1008344629 Chr.5: 142879758-142879758 on Build GRCh38
rs1037720200 Chr.5: 142879687-142879687 on Build GRCh38
rs111510621 Chr.5: 142879578-142879578 on Build GRCh38
rs756445857 Chr.5: 142879327-142879327 on Build GRCh38
rs769509954 Chr.5: 142879381-142879381 on Build GRCh38
rs534915757 Chr.5: 142879588-142879588 on Build GRCh38
rs1012939271 Chr.5: 142879438-142879438 on Build GRCh38
rs145809831 Chr.5: 142879614-142879614 on Build GRCh38
rs975266123 Chr.5: 142879647-142879647 on Build GRCh38
rs184602803 Chr.5: 142879547-142879547 on Build GRCh38
rs752295227 Chr.5: 142879427-142879427 on Build GRCh38
rs747220418 Chr.5: 142879461-142879461 on Build GRCh38
rs772966511 Chr.5: 142879616-142879616 on Build GRCh38
rs760932069 Chr.5: 142879505-142879505 on Build GRCh38
rs945807926 Chr.5: 142879346-142879346 on Build GRCh38
rs899273616 Chr.5: 142879717-142879717 on Build GRCh38
rs749173856 Chr.5: 142879469-142879469 on Build GRCh38
rs778273591 Chr.5: 142879342-142879342 on Build GRCh38
rs963850185 Chr.5: 142879646-142879646 on Build GRCh38
rs530958843 Chr.5: 142879406-142879406 on Build GRCh38
rs371207175 Chr.5: 142879522-142879522 on Build GRCh38
rs766345092 Chr.5: 142879401-142879401 on Build GRCh38

View Full Product Details