Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 149505357-149505872 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSNK1A1
Entrez Gene ID
1452
Gene Name
casein kinase 1 alpha 1
Gene Aliases
CK1, CK1a, CKIa, HEL-S-77p, HLCDGP1, PRO2975
UniGene
Hs.529862
Gene Chromosome Location
Chr.5: 149495894-149551552 on Build GRCh38
Transcripts
NM_001025105.2 NM_001271741.1 NM_001271742.1 NM_001892.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7140857 c.951C>G p.N317K Chr.5: 149505586-149505586 on Build GRCh38
5628327 c.1005A>G p.K335K Chr.5: 149505448-149505448 on Build GRCh38
3211571 c.1084A>G p.M362V Chr.5: 149505453-149505453 on Build GRCh38
1202493 c.959A>G p.Y320C Chr.5: 149505578-149505578 on Build GRCh38
1486483 c.930C>G p.A310A Chr.5: 149505523-149505523 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201722541 Chr.5: 149505405-149505405 on Build GRCh38
rs769704267 Chr.5: 149505616-149505617 on Build GRCh38
rs992722848 Chr.5: 149505695-149505695 on Build GRCh38
rs777754880 Chr.5: 149505501-149505501 on Build GRCh38
rs201877962 Chr.5: 149505580-149505580 on Build GRCh38
rs772749741 Chr.5: 149505408-149505408 on Build GRCh38
rs908531332 Chr.5: 149505797-149505797 on Build GRCh38
rs992359462 Chr.5: 149505416-149505416 on Build GRCh38
rs1023305617 Chr.5: 149505704-149505704 on Build GRCh38
rs983309687 Chr.5: 149505768-149505768 on Build GRCh38
rs3733847 Chr.5: 149505809-149505809 on Build GRCh38
rs900098767 Chr.5: 149505811-149505811 on Build GRCh38
rs757065353 Chr.5: 149505775-149505775 on Build GRCh38
rs776778335 Chr.5: 149505402-149505402 on Build GRCh38
rs904732789 Chr.5: 149505838-149505838 on Build GRCh38
rs779281905 Chr.5: 149505539-149505539 on Build GRCh38
rs756620717 Chr.5: 149505491-149505491 on Build GRCh38
rs899963001 Chr.5: 149505384-149505384 on Build GRCh38
rs762534270 Chr.5: 149505632-149505632 on Build GRCh38
rs751550938 Chr.5: 149505568-149505568 on Build GRCh38
rs757227847 Chr.5: 149505535-149505535 on Build GRCh38
rs748138854 Chr.5: 149505605-149505605 on Build GRCh38
rs944039959 Chr.5: 149505799-149505799 on Build GRCh38
rs369743923 Chr.5: 149505401-149505401 on Build GRCh38
rs755338170 Chr.5: 149505448-149505448 on Build GRCh38
rs999037646 Chr.5: 149505489-149505489 on Build GRCh38
rs542273175 Chr.5: 149505727-149505727 on Build GRCh38
rs973222059 Chr.5: 149505740-149505740 on Build GRCh38
rs930905756 Chr.5: 149505592-149505592 on Build GRCh38
rs750623940 Chr.5: 149505816-149505816 on Build GRCh38
rs998609796 Chr.5: 149505845-149505845 on Build GRCh38
rs760560736 Chr.5: 149505398-149505398 on Build GRCh38
rs920864643 Chr.5: 149505490-149505490 on Build GRCh38
rs765736453 Chr.5: 149505637-149505637 on Build GRCh38
rs931378666 Chr.5: 149505751-149505751 on Build GRCh38
rs969838471 Chr.5: 149505733-149505733 on Build GRCh38
rs761830529 Chr.5: 149505403-149505403 on Build GRCh38
rs920457724 Chr.5: 149505741-149505741 on Build GRCh38
rs769804898 Chr.5: 149505613-149505613 on Build GRCh38
rs1040861947 Chr.5: 149505773-149505773 on Build GRCh38
rs767860830 Chr.5: 149505453-149505453 on Build GRCh38
rs749416274 Chr.5: 149505532-149505532 on Build GRCh38
rs960010632 Chr.5: 149505653-149505653 on Build GRCh38
rs939691084 Chr.5: 149505436-149505436 on Build GRCh38
rs1043232069 Chr.5: 149505828-149505828 on Build GRCh38
rs55868099 Chr.5: 149505443-149505443 on Build GRCh38
rs374179351 Chr.5: 149505841-149505841 on Build GRCh38
rs772977210 Chr.5: 149505625-149505625 on Build GRCh38
rs753259198 Chr.5: 149505454-149505454 on Build GRCh38
rs766074358 Chr.5: 149505422-149505422 on Build GRCh38
rs201948041 Chr.5: 149505418-149505418 on Build GRCh38
rs776547952 Chr.5: 149505604-149505604 on Build GRCh38

View Full Product Details