Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 149506883-149507432 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSNK1A1
Entrez Gene ID
1452
Gene Name
casein kinase 1 alpha 1
Gene Aliases
CK1, CK1a, CKIa, HEL-S-77p, HLCDGP1, PRO2975
UniGene
Hs.529862
Gene Chromosome Location
Chr.5: 149495894-149551552 on Build GRCh38
Transcripts
NM_001025105.2 NM_001271741.1 NM_001271742.1 NM_001892.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6636700 c.791G>A p.R264H Chr.5: 149507093-149507093 on Build GRCh38
4675132 c.846C>T p.R282R Chr.5: 149507038-149507038 on Build GRCh38
4425888 c.780_781insTTA p.L260_N261insL Chr.5: 149507103-149507104 on Build GRCh38
4772430 c.844C>T p.R282C Chr.5: 149507040-149507040 on Build GRCh38
7245078 c.862T>C p.L288L Chr.5: 149507106-149507106 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749186897 Chr.5: 149507151-149507155 on Build GRCh38
rs758665547 Chr.5: 149507073-149507073 on Build GRCh38
rs371662153 Chr.5: 149507038-149507038 on Build GRCh38
rs756394368 Chr.5: 149506998-149506998 on Build GRCh38
rs778932699 Chr.5: 149507303-149507303 on Build GRCh38
rs754949334 Chr.5: 149507255-149507255 on Build GRCh38
rs865943998 Chr.5: 149507114-149507114 on Build GRCh38
rs113728568 Chr.5: 149507325-149507325 on Build GRCh38
rs575187826 Chr.5: 149507224-149507224 on Build GRCh38
rs779124439 Chr.5: 149507051-149507051 on Build GRCh38
rs996026398 Chr.5: 149507251-149507251 on Build GRCh38
rs959534212 Chr.5: 149507250-149507250 on Build GRCh38
rs889859473 Chr.5: 149507260-149507260 on Build GRCh38
rs370326610 Chr.5: 149507143-149507143 on Build GRCh38
rs886147836 Chr.5: 149507017-149507017 on Build GRCh38
rs115256922 Chr.5: 149506963-149506963 on Build GRCh38
rs931240036 Chr.5: 149506944-149506944 on Build GRCh38
rs1048242772 Chr.5: 149506950-149506950 on Build GRCh38
rs375067462 Chr.5: 149507088-149507088 on Build GRCh38
rs780385622 Chr.5: 149507083-149507083 on Build GRCh38
rs771743238 Chr.5: 149507182-149507182 on Build GRCh38
rs377042926 Chr.5: 149507000-149507000 on Build GRCh38
rs1024562868 Chr.5: 149507342-149507342 on Build GRCh38
rs540274924 Chr.5: 149507376-149507376 on Build GRCh38
rs971509704 Chr.5: 149507353-149507353 on Build GRCh38
rs868439164 Chr.5: 149507070-149507070 on Build GRCh38
rs1006607208 Chr.5: 149506958-149506958 on Build GRCh38
rs1001106591 Chr.5: 149507184-149507184 on Build GRCh38
rs554969230 Chr.5: 149507228-149507228 on Build GRCh38
rs1048694273 Chr.5: 149507007-149507007 on Build GRCh38
rs556833761 Chr.5: 149507153-149507153 on Build GRCh38
rs1009925262 Chr.5: 149506965-149506965 on Build GRCh38
rs188200739 Chr.5: 149507108-149507108 on Build GRCh38
rs149551454 Chr.5: 149507311-149507311 on Build GRCh38
rs927491052 Chr.5: 149507390-149507390 on Build GRCh38
rs749174716 Chr.5: 149507136-149507136 on Build GRCh38
rs746143349 Chr.5: 149507068-149507068 on Build GRCh38
rs972739294 Chr.5: 149507374-149507374 on Build GRCh38
rs1000302879 Chr.5: 149507275-149507275 on Build GRCh38
rs753027853 Chr.5: 149506988-149506988 on Build GRCh38
rs868542733 Chr.5: 149507304-149507304 on Build GRCh38
rs770530263 Chr.5: 149507147-149507147 on Build GRCh38
rs1033423929 Chr.5: 149507281-149507281 on Build GRCh38
rs775564113 Chr.5: 149507151-149507162 on Build GRCh38
rs573223971 Chr.5: 149507402-149507402 on Build GRCh38
rs769413851 Chr.5: 149507113-149507113 on Build GRCh38
rs759225086 Chr.5: 149507161-149507161 on Build GRCh38
rs3756505 Chr.5: 149506960-149506960 on Build GRCh38
rs374430739 Chr.5: 149506980-149506980 on Build GRCh38
rs930394316 Chr.5: 149507392-149507392 on Build GRCh38
rs773202370 Chr.5: 149507134-149507134 on Build GRCh38
rs558786773 Chr.5: 149506936-149506936 on Build GRCh38
rs367802378 Chr.5: 149506918-149506918 on Build GRCh38
rs760440923 Chr.5: 149506951-149506951 on Build GRCh38

View Full Product Details