Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 159917377-159917916 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADRA1B
Entrez Gene ID
147
Gene Name
adrenoceptor alpha 1B
Gene Aliases
ADRA1, ALPHA1BAR
UniGene
Hs.368632
Gene Chromosome Location
Chr.5: 159916733-159989205 on Build GRCh38
Transcripts
NM_000679.3 XM_005265818.3 XM_005265819.2 XM_006714821.3 XM_011534435.1 XM_011534437.2 XM_011534438.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4625501 c.788A>G p.D263G Chr.5: 159917693-159917693 on Build GRCh38
736975 c.593C>T p.T198I Chr.5: 159917498-159917498 on Build GRCh38
4553048 c.579G>A p.K193K Chr.5: 159917484-159917484 on Build GRCh38
4561122 c.870G>A p.K290K Chr.5: 159917775-159917775 on Build GRCh38
5596229 c.480G>A p.R160R Chr.5: 159917385-159917385 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866336763 Chr.5: 159917595-159917595 on Build GRCh38
rs200205839 Chr.5: 159917404-159917404 on Build GRCh38
rs745591182 Chr.5: 159917491-159917491 on Build GRCh38
rs1001010195 Chr.5: 159917614-159917614 on Build GRCh38
rs776503343 Chr.5: 159917695-159917695 on Build GRCh38
rs778570104 Chr.5: 159917625-159917625 on Build GRCh38
rs375325263 Chr.5: 159917508-159917508 on Build GRCh38
rs767897511 Chr.5: 159917567-159917567 on Build GRCh38
rs773653936 Chr.5: 159917860-159917860 on Build GRCh38
rs375482491 Chr.5: 159917416-159917416 on Build GRCh38
rs747013713 Chr.5: 159917827-159917844 on Build GRCh38
rs567109309 Chr.5: 159917486-159917486 on Build GRCh38
rs202097956 Chr.5: 159917788-159917788 on Build GRCh38
rs193266765 Chr.5: 159917712-159917712 on Build GRCh38
rs768628373 Chr.5: 159917689-159917689 on Build GRCh38
rs767751800 Chr.5: 159917730-159917730 on Build GRCh38
rs764414699 Chr.5: 159917875-159917875 on Build GRCh38
rs8192449 Chr.5: 159917886-159917886 on Build GRCh38
rs775757641 Chr.5: 159917851-159917851 on Build GRCh38
rs776642038 Chr.5: 159917864-159917864 on Build GRCh38
rs774512757 Chr.5: 159917719-159917719 on Build GRCh38
rs766562864 Chr.5: 159917460-159917460 on Build GRCh38
rs538840783 Chr.5: 159917526-159917526 on Build GRCh38
rs772006674 Chr.5: 159917493-159917493 on Build GRCh38
rs771917339 Chr.5: 159917580-159917581 on Build GRCh38
rs760728132 Chr.5: 159917506-159917506 on Build GRCh38
rs762712089 Chr.5: 159917476-159917476 on Build GRCh38
rs758073042 Chr.5: 159917789-159917789 on Build GRCh38
rs138161802 Chr.5: 159917616-159917616 on Build GRCh38
rs770934468 Chr.5: 159917718-159917718 on Build GRCh38
rs569874100 Chr.5: 159917860-159917860 on Build GRCh38
rs371948194 Chr.5: 159917677-159917677 on Build GRCh38
rs772799249 Chr.5: 159917397-159917397 on Build GRCh38
rs767558240 Chr.5: 159917437-159917437 on Build GRCh38
rs996097042 Chr.5: 159917670-159917670 on Build GRCh38
rs756589246 Chr.5: 159917579-159917579 on Build GRCh38
rs772191822 Chr.5: 159917787-159917787 on Build GRCh38
rs748796552 Chr.5: 159917478-159917478 on Build GRCh38
rs909145193 Chr.5: 159917572-159917572 on Build GRCh38
rs771955277 Chr.5: 159917638-159917638 on Build GRCh38
rs375270240 Chr.5: 159917741-159917741 on Build GRCh38
rs764385052 Chr.5: 159917523-159917525 on Build GRCh38
rs150394007 Chr.5: 159917578-159917578 on Build GRCh38
rs748673379 Chr.5: 159917468-159917468 on Build GRCh38
rs574447886 Chr.5: 159917765-159917765 on Build GRCh38
rs745539683 Chr.5: 159917636-159917636 on Build GRCh38
rs149628499 Chr.5: 159917790-159917790 on Build GRCh38
rs770527826 Chr.5: 159917407-159917407 on Build GRCh38
rs764657630 Chr.5: 159917745-159917745 on Build GRCh38
rs369120935 Chr.5: 159917592-159917592 on Build GRCh38
rs542410229 Chr.5: 159917475-159917475 on Build GRCh38
rs775520347 Chr.5: 159917499-159917499 on Build GRCh38
rs761926773 Chr.5: 159917441-159917441 on Build GRCh38
rs759717495 Chr.5: 159917728-159917728 on Build GRCh38
rs777527602 Chr.5: 159917706-159917708 on Build GRCh38
rs3729604 Chr.5: 159917454-159917454 on Build GRCh38
rs746852984 Chr.5: 159917688-159917688 on Build GRCh38
rs781171713 Chr.5: 159917791-159917791 on Build GRCh38
rs367829656 Chr.5: 159917449-159917449 on Build GRCh38
rs778325967 Chr.5: 159917772-159917772 on Build GRCh38
rs781618796 Chr.5: 159917466-159917466 on Build GRCh38
rs750573642 Chr.5: 159917453-159917453 on Build GRCh38
rs761297353 Chr.5: 159917428-159917428 on Build GRCh38
rs753003514 Chr.5: 159917735-159917735 on Build GRCh38
rs766624495 Chr.5: 159917547-159917547 on Build GRCh38
rs774367999 Chr.5: 159917806-159917806 on Build GRCh38
rs554286300 Chr.5: 159917859-159917859 on Build GRCh38
rs146057113 Chr.5: 159917406-159917406 on Build GRCh38
rs3730284 Chr.5: 159917439-159917439 on Build GRCh38
rs552863026 Chr.5: 159917556-159917556 on Build GRCh38
rs751740593 Chr.5: 159917555-159917555 on Build GRCh38
rs537149780 Chr.5: 159917698-159917698 on Build GRCh38
rs763003987 Chr.5: 159917545-159917545 on Build GRCh38
rs1003066551 Chr.5: 159917645-159917645 on Build GRCh38
rs748079207 Chr.5: 159917805-159917805 on Build GRCh38
rs371731539 Chr.5: 159917463-159917463 on Build GRCh38
rs756724973 Chr.5: 159917618-159917618 on Build GRCh38
rs937614786 Chr.5: 159917481-159917481 on Build GRCh38
rs201944859 Chr.5: 159917771-159917771 on Build GRCh38
rs779966041 Chr.5: 159917661-159917661 on Build GRCh38
rs751944506 Chr.5: 159917462-159917462 on Build GRCh38
rs759306305 Chr.5: 159917431-159917431 on Build GRCh38
rs942105612 Chr.5: 159917435-159917435 on Build GRCh38
rs576994119 Chr.5: 159917850-159917850 on Build GRCh38
rs1031620021 Chr.5: 159917849-159917849 on Build GRCh38
rs973874534 Chr.5: 159917842-159917842 on Build GRCh38
rs754360695 Chr.5: 159917746-159917746 on Build GRCh38
rs766237542 Chr.5: 159917587-159917587 on Build GRCh38

View Full Product Details