Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 160399845-160400356 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZBED8
Entrez Gene ID
63920
Gene Name
zinc finger BED-type containing 8
Gene Aliases
Buster3, C5orf54
UniGene
Hs.529464
Gene Chromosome Location
Chr.5: 160393148-160400097 on Build GRCh38
Transcripts
NM_001303251.1 NM_022090.4

SNP Detail

SNP ID Chromosome Location
rs1037627735 Chr.5: 160400082-160400082 on Build GRCh38
rs148031634 Chr.5: 160400007-160400007 on Build GRCh38
rs746810058 Chr.5: 160400255-160400255 on Build GRCh38
rs371134019 Chr.5: 160399947-160399947 on Build GRCh38
rs565512577 Chr.5: 160400043-160400043 on Build GRCh38
rs895803860 Chr.5: 160400141-160400141 on Build GRCh38
rs1026348856 Chr.5: 160400137-160400137 on Build GRCh38
rs533157038 Chr.5: 160399969-160399969 on Build GRCh38
rs563297098 Chr.5: 160400284-160400284 on Build GRCh38
rs534835692 Chr.5: 160400061-160400061 on Build GRCh38
rs556956532 Chr.5: 160400102-160400102 on Build GRCh38
rs943130786 Chr.5: 160400280-160400280 on Build GRCh38
rs1012939711 Chr.5: 160399869-160399869 on Build GRCh38
rs764927887 Chr.5: 160400034-160400034 on Build GRCh38
rs907291382 Chr.5: 160400020-160400020 on Build GRCh38
rs550015979 Chr.5: 160400235-160400235 on Build GRCh38
rs909425009 Chr.5: 160400135-160400135 on Build GRCh38
rs778334103 Chr.5: 160400158-160400158 on Build GRCh38
rs1007771707 Chr.5: 160400081-160400081 on Build GRCh38
rs913592491 Chr.5: 160400219-160400219 on Build GRCh38
rs936350438 Chr.5: 160400039-160400039 on Build GRCh38
rs1051580689 Chr.5: 160399942-160399942 on Build GRCh38
rs969210224 Chr.5: 160399901-160399901 on Build GRCh38
rs888972806 Chr.5: 160400079-160400079 on Build GRCh38
rs963419571 Chr.5: 160399991-160399991 on Build GRCh38
rs779743566 Chr.5: 160400202-160400202 on Build GRCh38
rs1023443388 Chr.5: 160400173-160400173 on Build GRCh38
rs1044374907 Chr.5: 160400186-160400186 on Build GRCh38
rs562389530 Chr.5: 160400175-160400175 on Build GRCh38
rs1022974583 Chr.5: 160399873-160399873 on Build GRCh38
rs776737537 Chr.5: 160400299-160400299 on Build GRCh38
rs993407732 Chr.5: 160400131-160400131 on Build GRCh38
rs181548895 Chr.5: 160400187-160400187 on Build GRCh38
rs997216882 Chr.5: 160399921-160399921 on Build GRCh38
rs374520415 Chr.5: 160400015-160400015 on Build GRCh38
rs960147237 Chr.5: 160399999-160399999 on Build GRCh38
rs1052086820 Chr.5: 160400041-160400041 on Build GRCh38
rs985897260 Chr.5: 160399978-160399978 on Build GRCh38
rs987190578 Chr.5: 160400272-160400272 on Build GRCh38
rs564619509 Chr.5: 160399965-160399965 on Build GRCh38
rs956679629 Chr.5: 160400030-160400030 on Build GRCh38
rs1014367276 Chr.5: 160400167-160400167 on Build GRCh38
rs766622781 Chr.5: 160399907-160399907 on Build GRCh38
rs756784666 Chr.5: 160400117-160400117 on Build GRCh38
rs34018399 Chr.5: 160400173-160400174 on Build GRCh38
rs1007271613 Chr.5: 160400218-160400218 on Build GRCh38
rs142087524 Chr.5: 160400057-160400057 on Build GRCh38
rs1035249389 Chr.5: 160400021-160400021 on Build GRCh38
rs397750249 Chr.5: 160400181-160400182 on Build GRCh38
rs559787162 Chr.5: 160400172-160400173 on Build GRCh38
rs910264329 Chr.5: 160399986-160399986 on Build GRCh38
rs1037973331 Chr.5: 160400283-160400283 on Build GRCh38
rs910256749 Chr.5: 160400277-160400277 on Build GRCh38
rs1016567801 Chr.5: 160399977-160399977 on Build GRCh38
rs146326216 Chr.5: 160400195-160400195 on Build GRCh38
rs573685080 Chr.5: 160400125-160400125 on Build GRCh38
rs932964226 Chr.5: 160400237-160400237 on Build GRCh38
rs970257924 Chr.5: 160400184-160400184 on Build GRCh38
rs1022641624 Chr.5: 160400212-160400212 on Build GRCh38
rs755277184 Chr.5: 160400008-160400008 on Build GRCh38
rs751931254 Chr.5: 160400006-160400006 on Build GRCh38
rs536636957 Chr.5: 160400078-160400078 on Build GRCh38
rs200605286 Chr.5: 160400172-160400172 on Build GRCh38
rs190750191 Chr.5: 160399964-160399964 on Build GRCh38
rs753163186 Chr.5: 160400031-160400031 on Build GRCh38
rs910337784 Chr.5: 160400059-160400059 on Build GRCh38
rs542493154 Chr.5: 160400129-160400129 on Build GRCh38
rs577166096 Chr.5: 160400314-160400314 on Build GRCh38
rs944864075 Chr.5: 160399988-160399988 on Build GRCh38
rs397762167 Chr.5: 160400182-160400183 on Build GRCh38
rs985759833 Chr.5: 160400055-160400055 on Build GRCh38
rs956833574 Chr.5: 160399949-160399949 on Build GRCh38
rs61337580 Chr.5: 160399992-160399992 on Build GRCh38
rs185864555 Chr.5: 160400200-160400200 on Build GRCh38
rs557553688 Chr.5: 160400070-160400070 on Build GRCh38
rs17409777 Chr.5: 160400201-160400201 on Build GRCh38
rs577698109 Chr.5: 160400074-160400074 on Build GRCh38
rs112272406 Chr.5: 160400018-160400018 on Build GRCh38
rs74952614 Chr.5: 160400214-160400214 on Build GRCh38
rs768495026 Chr.5: 160400286-160400286 on Build GRCh38
rs967711664 Chr.5: 160399966-160399966 on Build GRCh38
rs899116364 Chr.5: 160400111-160400111 on Build GRCh38
rs569607082 Chr.5: 160399994-160399994 on Build GRCh38

View Full Product Details