Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 178204417-178204907 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNRNPAB
Entrez Gene ID
3182
Gene Name
heterogeneous nuclear ribonucleoprotein A/B
Gene Aliases
ABBP1, HNRPAB
UniGene
Hs.591731
Gene Chromosome Location
Chr.5: 178204507-178211183 on Build GRCh38
Transcripts
NM_004499.3 NM_031266.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7430495 c.5C>T p.S2L Chr.5: 178204842-178204842 on Build GRCh38
5662887 c.65C>T p.A22V Chr.5: 178204902-178204902 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs896540147 Chr.5: 178204512-178204512 on Build GRCh38
rs1037635112 Chr.5: 178204695-178204695 on Build GRCh38
rs180935005 Chr.5: 178204659-178204659 on Build GRCh38
rs1034029450 Chr.5: 178204625-178204625 on Build GRCh38
rs1048182208 Chr.5: 178204522-178204522 on Build GRCh38
rs890652008 Chr.5: 178204600-178204600 on Build GRCh38
rs988860026 Chr.5: 178204490-178204490 on Build GRCh38
rs534795122 Chr.5: 178204553-178204553 on Build GRCh38
rs1016369680 Chr.5: 178204469-178204469 on Build GRCh38
rs143881773 Chr.5: 178204555-178204555 on Build GRCh38
rs771824167 Chr.5: 178204523-178204523 on Build GRCh38
rs971050298 Chr.5: 178204821-178204821 on Build GRCh38
rs968408464 Chr.5: 178204545-178204545 on Build GRCh38
rs1006510542 Chr.5: 178204615-178204615 on Build GRCh38
rs912910352 Chr.5: 178204491-178204491 on Build GRCh38
rs996259365 Chr.5: 178204534-178204534 on Build GRCh38
rs997540669 Chr.5: 178204599-178204601 on Build GRCh38
rs1013686937 Chr.5: 178204520-178204520 on Build GRCh38
rs886432420 Chr.5: 178204564-178204564 on Build GRCh38
rs1030902651 Chr.5: 178204581-178204581 on Build GRCh38
rs532826080 Chr.5: 178204747-178204747 on Build GRCh38
rs541221472 Chr.5: 178204661-178204661 on Build GRCh38
rs922977052 Chr.5: 178204487-178204487 on Build GRCh38
rs937406551 Chr.5: 178204758-178204769 on Build GRCh38
rs763662785 Chr.5: 178204603-178204603 on Build GRCh38
rs774991812 Chr.5: 178204550-178204550 on Build GRCh38
rs900737725 Chr.5: 178204572-178204572 on Build GRCh38
rs954485854 Chr.5: 178204489-178204489 on Build GRCh38
rs1041799912 Chr.5: 178204590-178204590 on Build GRCh38
rs937124595 Chr.5: 178204503-178204503 on Build GRCh38
rs537307687 Chr.5: 178204494-178204494 on Build GRCh38
rs1024190028 Chr.5: 178204784-178204784 on Build GRCh38
rs550955362 Chr.5: 178204455-178204455 on Build GRCh38
rs993043927 Chr.5: 178204558-178204558 on Build GRCh38
rs1043017527 Chr.5: 178204493-178204493 on Build GRCh38
rs992784941 Chr.5: 178204650-178204650 on Build GRCh38
rs886466814 Chr.5: 178204524-178204524 on Build GRCh38
rs1020351228 Chr.5: 178204541-178204541 on Build GRCh38
rs1034000610 Chr.5: 178204737-178204737 on Build GRCh38
rs146897728 Chr.5: 178204559-178204559 on Build GRCh38
rs1037666316 Chr.5: 178204569-178204569 on Build GRCh38
rs370637878 Chr.5: 178204849-178204849 on Build GRCh38
rs138283723 Chr.5: 178204516-178204516 on Build GRCh38
rs1006426950 Chr.5: 178204526-178204526 on Build GRCh38
rs918460940 Chr.5: 178204475-178204475 on Build GRCh38
rs974455113 Chr.5: 178204482-178204482 on Build GRCh38
rs958601123 Chr.5: 178204640-178204640 on Build GRCh38
rs1001958837 Chr.5: 178204510-178204510 on Build GRCh38
rs901913687 Chr.5: 178204591-178204591 on Build GRCh38
rs964886582 Chr.5: 178204473-178204473 on Build GRCh38
rs1013965814 Chr.5: 178204449-178204449 on Build GRCh38
rs927104639 Chr.5: 178204501-178204501 on Build GRCh38
rs978432739 Chr.5: 178204549-178204549 on Build GRCh38
rs55653122 Chr.5: 178204467-178204467 on Build GRCh38
rs909519053 Chr.5: 178204465-178204465 on Build GRCh38
rs904245688 Chr.5: 178204722-178204722 on Build GRCh38
rs1034480910 Chr.5: 178204885-178204885 on Build GRCh38
rs555398723 Chr.5: 178204508-178204508 on Build GRCh38
rs889290581 Chr.5: 178204772-178204772 on Build GRCh38
rs907923775 Chr.5: 178204687-178204687 on Build GRCh38
rs947273332 Chr.5: 178204492-178204492 on Build GRCh38
rs765862424 Chr.5: 178204803-178204803 on Build GRCh38
rs999977012 Chr.5: 178204735-178204735 on Build GRCh38
rs1056968697 Chr.5: 178204504-178204504 on Build GRCh38
rs946071000 Chr.5: 178204736-178204736 on Build GRCh38
rs753208075 Chr.5: 178204611-178204611 on Build GRCh38
rs954583127 Chr.5: 178204538-178204538 on Build GRCh38
rs1026335234 Chr.5: 178204451-178204451 on Build GRCh38
rs985432064 Chr.5: 178204554-178204554 on Build GRCh38
rs551192335 Chr.5: 178204768-178204768 on Build GRCh38
rs1019999684 Chr.5: 178204613-178204613 on Build GRCh38
rs988605958 Chr.5: 178204539-178204539 on Build GRCh38
rs766667790 Chr.5: 178204547-178204547 on Build GRCh38
rs900539488 Chr.5: 178204702-178204702 on Build GRCh38
rs868515393 Chr.5: 178204704-178204704 on Build GRCh38
rs978401482 Chr.5: 178204617-178204617 on Build GRCh38
rs941999934 Chr.5: 178204568-178204568 on Build GRCh38
rs760825764 Chr.5: 178204527-178204527 on Build GRCh38
rs1030515262 Chr.5: 178204536-178204536 on Build GRCh38
rs949825642 Chr.5: 178204476-178204476 on Build GRCh38
rs559559756 Chr.5: 178204662-178204662 on Build GRCh38
rs1025349789 Chr.5: 178204812-178204812 on Build GRCh38
rs1054778376 Chr.5: 178204565-178204565 on Build GRCh38
rs942135774 Chr.5: 178204688-178204688 on Build GRCh38
rs968460350 Chr.5: 178204616-178204616 on Build GRCh38
rs900769036 Chr.5: 178204532-178204532 on Build GRCh38
rs1041927154 Chr.5: 178204718-178204718 on Build GRCh38
rs188993620 Chr.5: 178204557-178204557 on Build GRCh38
rs866477157 Chr.5: 178204561-178204561 on Build GRCh38
rs1045685923 Chr.5: 178204480-178204480 on Build GRCh38
rs11546684 Chr.5: 178204566-178204566 on Build GRCh38
rs1052062613 Chr.5: 178204710-178204710 on Build GRCh38
rs951376178 Chr.5: 178204799-178204799 on Build GRCh38
rs951006791 Chr.5: 178204452-178204452 on Build GRCh38
rs922155748 Chr.5: 178204450-178204450 on Build GRCh38
rs576380451 Chr.5: 178204627-178204627 on Build GRCh38
rs370707939 Chr.5: 178204563-178204563 on Build GRCh38
rs983028925 Chr.5: 178204663-178204663 on Build GRCh38
rs776858195 Chr.5: 178204556-178204556 on Build GRCh38
rs901836805 Chr.5: 178204753-178204753 on Build GRCh38
rs996455643 Chr.5: 178204578-178204578 on Build GRCh38
rs562226717 Chr.5: 178204655-178204655 on Build GRCh38
rs890610906 Chr.5: 178204713-178204713 on Build GRCh38
rs774144926 Chr.5: 178204445-178204445 on Build GRCh38
rs17080447 Chr.5: 178204782-178204782 on Build GRCh38
rs1016503615 Chr.5: 178204530-178204530 on Build GRCh38
rs1010365892 Chr.5: 178204716-178204716 on Build GRCh38

View Full Product Details