Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150806551-150807045 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTSK
Entrez Gene ID
1513
Gene Name
cathepsin K
Gene Aliases
CTS02, CTSO, CTSO1, CTSO2, PKND, PYCD
UniGene
Hs.632466
Gene Chromosome Location
Chr.1: 150796208-150808441 on Build GRCh38
Transcripts
NM_000396.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
675526 c.225G>T p.L75L Chr.1: 150806758-150806758 on Build GRCh38
6637813 c.2T>A p.M1K Chr.1: 150806804-150806804 on Build GRCh38
289673 c.177-9C>A p.? Chr.1: 150806815-150806815 on Build GRCh38
3801769 c.96C>A p.T32T Chr.1: 150806710-150806710 on Build GRCh38
5843572 c.187C>T p.L63F Chr.1: 150806796-150806796 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776595418 Chr.1: 150806741-150806741 on Build GRCh38
rs778368118 Chr.1: 150806803-150806803 on Build GRCh38
rs746596138 Chr.1: 150806727-150806727 on Build GRCh38
rs958032340 Chr.1: 150806864-150806864 on Build GRCh38
rs145128083 Chr.1: 150806742-150806742 on Build GRCh38
rs188977420 Chr.1: 150806582-150806582 on Build GRCh38
rs938267905 Chr.1: 150806765-150806765 on Build GRCh38
rs751636691 Chr.1: 150806829-150806829 on Build GRCh38
rs1057517252 Chr.1: 150806780-150806780 on Build GRCh38
rs1055444697 Chr.1: 150806770-150806770 on Build GRCh38
rs756695909 Chr.1: 150806799-150806799 on Build GRCh38
rs202055627 Chr.1: 150806798-150806798 on Build GRCh38
rs763148656 Chr.1: 150806701-150806703 on Build GRCh38
rs371464900 Chr.1: 150806794-150806794 on Build GRCh38
rs779796688 Chr.1: 150806810-150806810 on Build GRCh38
rs751721868 Chr.1: 150806653-150806653 on Build GRCh38
rs139004720 Chr.1: 150806776-150806776 on Build GRCh38
rs148868110 Chr.1: 150806619-150806619 on Build GRCh38
rs901409428 Chr.1: 150807006-150807009 on Build GRCh38
rs267598007 Chr.1: 150806709-150806709 on Build GRCh38
rs59080032 Chr.1: 150807025-150807026 on Build GRCh38
rs587717509 Chr.1: 150806901-150806901 on Build GRCh38
rs1048095760 Chr.1: 150807025-150807025 on Build GRCh38
rs1057517279 Chr.1: 150806685-150806685 on Build GRCh38
rs780707867 Chr.1: 150806833-150806833 on Build GRCh38
rs111262867 Chr.1: 150807001-150807001 on Build GRCh38
rs138360770 Chr.1: 150806784-150806784 on Build GRCh38
rs146635750 Chr.1: 150807005-150807006 on Build GRCh38
rs143503946 Chr.1: 150806986-150806986 on Build GRCh38
rs376008346 Chr.1: 150806797-150806797 on Build GRCh38
rs201147412 Chr.1: 150806636-150806636 on Build GRCh38
rs757556509 Chr.1: 150806809-150806809 on Build GRCh38
rs886044631 Chr.1: 150806716-150806716 on Build GRCh38
rs777031474 Chr.1: 150806580-150806580 on Build GRCh38
rs1028244850 Chr.1: 150806841-150806841 on Build GRCh38
rs755011972 Chr.1: 150806665-150806665 on Build GRCh38
rs889764876 Chr.1: 150806969-150806969 on Build GRCh38
rs587662312 Chr.1: 150806866-150806866 on Build GRCh38
rs587650413 Chr.1: 150806946-150806946 on Build GRCh38
rs931348037 Chr.1: 150806957-150806957 on Build GRCh38
rs879450713 Chr.1: 150807004-150807005 on Build GRCh38
rs774989958 Chr.1: 150806724-150806724 on Build GRCh38
rs201938306 Chr.1: 150806667-150806667 on Build GRCh38
rs377383882 Chr.1: 150806664-150806664 on Build GRCh38
rs989977569 Chr.1: 150806616-150806616 on Build GRCh38
rs913967365 Chr.1: 150806902-150806902 on Build GRCh38
rs747549150 Chr.1: 150806640-150806640 on Build GRCh38
rs1048622635 Chr.1: 150806787-150806787 on Build GRCh38
rs10648774 Chr.1: 150807005-150807005 on Build GRCh38
rs1057516790 Chr.1: 150806758-150806758 on Build GRCh38
rs878961164 Chr.1: 150806696-150806696 on Build GRCh38
rs12746973 Chr.1: 150806925-150806925 on Build GRCh38
rs989534619 Chr.1: 150806893-150806893 on Build GRCh38
rs587623935 Chr.1: 150806631-150806631 on Build GRCh38
rs766465430 Chr.1: 150806811-150806813 on Build GRCh38
rs192522598 Chr.1: 150806980-150806980 on Build GRCh38
rs1048435976 Chr.1: 150806959-150806959 on Build GRCh38
rs200480574 Chr.1: 150806815-150806815 on Build GRCh38
rs768245146 Chr.1: 150806816-150806816 on Build GRCh38
rs754855152 Chr.1: 150806853-150806853 on Build GRCh38
rs1020217112 Chr.1: 150806633-150806633 on Build GRCh38
rs750595350 Chr.1: 150806687-150806689 on Build GRCh38
rs879490870 Chr.1: 150806602-150806602 on Build GRCh38
rs201051635 Chr.1: 150806785-150806785 on Build GRCh38
rs938216810 Chr.1: 150806948-150806948 on Build GRCh38
rs770382502 Chr.1: 150806919-150806922 on Build GRCh38
rs369459638 Chr.1: 150806689-150806689 on Build GRCh38
rs756640761 Chr.1: 150806676-150806676 on Build GRCh38
rs754286434 Chr.1: 150806806-150806806 on Build GRCh38
rs755476938 Chr.1: 150806833-150806834 on Build GRCh38
rs201086925 Chr.1: 150806790-150806790 on Build GRCh38
rs71659432 Chr.1: 150807024-150807025 on Build GRCh38
rs966847865 Chr.1: 150806912-150806912 on Build GRCh38
rs771768736 Chr.1: 150806691-150806691 on Build GRCh38
rs901317598 Chr.1: 150806661-150806661 on Build GRCh38

View Full Product Details