Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 5368471-5368965 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FARS2
Entrez Gene ID
10667
Gene Name
phenylalanyl-tRNA synthetase 2, mitochondrial
Gene Aliases
COXPD14, FARS1, HSPC320, PheRS
UniGene
Hs.484547
Gene Chromosome Location
Chr.6: 5261001-5771592 on Build GRCh38
Transcripts
NM_001318872.1 NM_006567.4 XM_005248812.3 XM_006714966.2 XM_011514247.2 XM_011514248.2 XM_011514249.1 XM_011514251.2 XM_017010186.1 XM_017010187.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4884152 c.384C>G p.S128R Chr.6: 5368954-5368954 on Build GRCh38
6547063 c.256C>T p.L86L Chr.6: 5368826-5368826 on Build GRCh38
4913232 c.273G>A p.E91E Chr.6: 5368843-5368843 on Build GRCh38
6471309 c.358G>T p.V120F Chr.6: 5368928-5368928 on Build GRCh38
4850977 c.18C>G p.L6L Chr.6: 5368588-5368588 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200731335 Chr.6: 5368914-5368914 on Build GRCh38
rs372239507 Chr.6: 5368526-5368526 on Build GRCh38
rs747659489 Chr.6: 5368665-5368665 on Build GRCh38
rs201710092 Chr.6: 5368709-5368709 on Build GRCh38
rs964653311 Chr.6: 5368756-5368756 on Build GRCh38
rs143959504 Chr.6: 5368837-5368837 on Build GRCh38
rs777130213 Chr.6: 5368801-5368801 on Build GRCh38
rs774652318 Chr.6: 5368562-5368562 on Build GRCh38
rs113155624 Chr.6: 5368672-5368672 on Build GRCh38
rs754644366 Chr.6: 5368656-5368656 on Build GRCh38
rs537850422 Chr.6: 5368650-5368650 on Build GRCh38
rs773187294 Chr.6: 5368829-5368829 on Build GRCh38
rs746431585 Chr.6: 5368676-5368676 on Build GRCh38
rs1057519095 Chr.6: 5368931-5368931 on Build GRCh38
rs780467389 Chr.6: 5368754-5368754 on Build GRCh38
rs551537911 Chr.6: 5368932-5368932 on Build GRCh38
rs201693261 Chr.6: 5368633-5368633 on Build GRCh38
rs748418895 Chr.6: 5368822-5368822 on Build GRCh38
rs750482903 Chr.6: 5368713-5368713 on Build GRCh38
rs748813583 Chr.6: 5368783-5368783 on Build GRCh38
rs759889873 Chr.6: 5368911-5368911 on Build GRCh38
rs368352168 Chr.6: 5368773-5368773 on Build GRCh38
rs758852317 Chr.6: 5368785-5368785 on Build GRCh38
rs201410497 Chr.6: 5368858-5368858 on Build GRCh38
rs566589950 Chr.6: 5368942-5368942 on Build GRCh38
rs34382405 Chr.6: 5368740-5368740 on Build GRCh38
rs775771055 Chr.6: 5368687-5368687 on Build GRCh38
rs759961408 Chr.6: 5368574-5368574 on Build GRCh38
rs573996374 Chr.6: 5368640-5368640 on Build GRCh38
rs780570475 Chr.6: 5368791-5368791 on Build GRCh38
rs762847777 Chr.6: 5368724-5368724 on Build GRCh38
rs369489810 Chr.6: 5368589-5368589 on Build GRCh38
rs140589763 Chr.6: 5368888-5368888 on Build GRCh38
rs139618748 Chr.6: 5368894-5368894 on Build GRCh38
rs761074033 Chr.6: 5368690-5368690 on Build GRCh38
rs764390927 Chr.6: 5368591-5368591 on Build GRCh38
rs1000968507 Chr.6: 5368681-5368681 on Build GRCh38
rs556195502 Chr.6: 5368661-5368661 on Build GRCh38
rs765226644 Chr.6: 5368606-5368606 on Build GRCh38
rs780728442 Chr.6: 5368531-5368531 on Build GRCh38
rs761946912 Chr.6: 5368599-5368599 on Build GRCh38
rs772379847 Chr.6: 5368576-5368576 on Build GRCh38
rs200380429 Chr.6: 5368578-5368578 on Build GRCh38
rs41302853 Chr.6: 5368909-5368909 on Build GRCh38
rs769085279 Chr.6: 5368798-5368798 on Build GRCh38
rs915798435 Chr.6: 5368931-5368931 on Build GRCh38
rs369145259 Chr.6: 5368902-5368902 on Build GRCh38
rs766227186 Chr.6: 5368843-5368843 on Build GRCh38
rs776929836 Chr.6: 5368699-5368699 on Build GRCh38
rs755776049 Chr.6: 5368882-5368882 on Build GRCh38
rs766289702 Chr.6: 5368737-5368737 on Build GRCh38
rs375178804 Chr.6: 5368796-5368796 on Build GRCh38
rs1000610059 Chr.6: 5368809-5368809 on Build GRCh38
rs752460037 Chr.6: 5368878-5368878 on Build GRCh38
rs750384202 Chr.6: 5368608-5368608 on Build GRCh38
rs749337994 Chr.6: 5368929-5368929 on Build GRCh38
rs73718082 Chr.6: 5368753-5368753 on Build GRCh38
rs374531453 Chr.6: 5368772-5368772 on Build GRCh38
rs894420718 Chr.6: 5368642-5368642 on Build GRCh38
rs148568494 Chr.6: 5368671-5368671 on Build GRCh38
rs774214415 Chr.6: 5368857-5368857 on Build GRCh38
rs886961566 Chr.6: 5368571-5368571 on Build GRCh38
rs748474162 Chr.6: 5368907-5368907 on Build GRCh38
rs931069133 Chr.6: 5368550-5368550 on Build GRCh38
rs771455141 Chr.6: 5368536-5368536 on Build GRCh38
rs113667689 Chr.6: 5368580-5368580 on Build GRCh38
rs769031929 Chr.6: 5368698-5368698 on Build GRCh38
rs770035560 Chr.6: 5368823-5368823 on Build GRCh38
rs377298153 Chr.6: 5368834-5368834 on Build GRCh38
rs146423499 Chr.6: 5368870-5368870 on Build GRCh38
rs747604331 Chr.6: 5368535-5368535 on Build GRCh38
rs781723007 Chr.6: 5368903-5368903 on Build GRCh38
rs367691523 Chr.6: 5368794-5368794 on Build GRCh38
rs371681314 Chr.6: 5368838-5368838 on Build GRCh38
rs111573636 Chr.6: 5368577-5368577 on Build GRCh38
rs374957295 Chr.6: 5368893-5368893 on Build GRCh38
rs753993545 Chr.6: 5368598-5368598 on Build GRCh38
rs982485775 Chr.6: 5368880-5368880 on Build GRCh38
rs961872269 Chr.6: 5368897-5368897 on Build GRCh38
rs772604608 Chr.6: 5368678-5368678 on Build GRCh38

View Full Product Details