Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 6195663-6196196 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F13A1
Entrez Gene ID
2162
Gene Name
coagulation factor XIII A chain
Gene Aliases
F13A
UniGene
Hs.335513
Gene Chromosome Location
Chr.6: 6144078-6320691 on Build GRCh38
Transcripts
NM_000129.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3629115 c.1226G>A p.R409Q Chr.6: 6195876-6195876 on Build GRCh38
280915 c.1298T>C p.F433S Chr.6: 6195804-6195804 on Build GRCh38
5543872 c.1220T>A p.M407K Chr.6: 6195882-6195882 on Build GRCh38
5638552 c.1250C>G p.A417G Chr.6: 6195852-6195852 on Build GRCh38
5871001 c.1235C>T p.P412L Chr.6: 6195867-6195867 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748366519 Chr.6: 6195989-6195989 on Build GRCh38
rs904804163 Chr.6: 6196095-6196095 on Build GRCh38
rs749063236 Chr.6: 6195754-6195754 on Build GRCh38
rs1010968955 Chr.6: 6195924-6195924 on Build GRCh38
rs532879106 Chr.6: 6195876-6195876 on Build GRCh38
rs757186297 Chr.6: 6195890-6195890 on Build GRCh38
rs138036988 Chr.6: 6195885-6195885 on Build GRCh38
rs879675898 Chr.6: 6195908-6195909 on Build GRCh38
rs752387839 Chr.6: 6195862-6195863 on Build GRCh38
rs1010692675 Chr.6: 6196035-6196035 on Build GRCh38
rs996342854 Chr.6: 6196134-6196134 on Build GRCh38
rs891763465 Chr.6: 6195983-6195983 on Build GRCh38
rs758807616 Chr.6: 6195690-6195690 on Build GRCh38
rs143513902 Chr.6: 6195816-6195816 on Build GRCh38
rs547446984 Chr.6: 6195716-6195716 on Build GRCh38
rs745509233 Chr.6: 6195771-6195771 on Build GRCh38
rs547712318 Chr.6: 6195817-6195817 on Build GRCh38
rs967020277 Chr.6: 6196125-6196125 on Build GRCh38
rs554857731 Chr.6: 6195955-6195955 on Build GRCh38
rs931153830 Chr.6: 6195846-6195846 on Build GRCh38
rs537528503 Chr.6: 6196136-6196136 on Build GRCh38
rs777460600 Chr.6: 6195753-6195753 on Build GRCh38
rs771765208 Chr.6: 6195778-6195778 on Build GRCh38
rs778750323 Chr.6: 6195769-6195769 on Build GRCh38
rs756028102 Chr.6: 6195884-6195884 on Build GRCh38
rs1037980724 Chr.6: 6195814-6195814 on Build GRCh38
rs977989666 Chr.6: 6196171-6196171 on Build GRCh38
rs139636536 Chr.6: 6195865-6195865 on Build GRCh38
rs183526980 Chr.6: 6196099-6196099 on Build GRCh38
rs763420902 Chr.6: 6195842-6195842 on Build GRCh38
rs768025872 Chr.6: 6195927-6195927 on Build GRCh38
rs756017371 Chr.6: 6195752-6195752 on Build GRCh38
rs533747347 Chr.6: 6195704-6195704 on Build GRCh38
rs1021870144 Chr.6: 6196057-6196057 on Build GRCh38
rs943322791 Chr.6: 6195798-6195798 on Build GRCh38
rs919874553 Chr.6: 6195836-6195836 on Build GRCh38
rs558694591 Chr.6: 6196068-6196068 on Build GRCh38
rs894835557 Chr.6: 6195915-6195915 on Build GRCh38
rs121913070 Chr.6: 6195859-6195859 on Build GRCh38
rs78204282 Chr.6: 6195896-6195896 on Build GRCh38
rs752593769 Chr.6: 6195746-6195746 on Build GRCh38
rs5979 Chr.6: 6195763-6195763 on Build GRCh38
rs371619552 Chr.6: 6195831-6195831 on Build GRCh38
rs768572894 Chr.6: 6195700-6195700 on Build GRCh38
rs5978 Chr.6: 6195908-6195908 on Build GRCh38
rs770291227 Chr.6: 6195841-6195841 on Build GRCh38
rs200317911 Chr.6: 6195922-6195922 on Build GRCh38
rs764800678 Chr.6: 6195777-6195778 on Build GRCh38
rs542717136 Chr.6: 6195701-6195701 on Build GRCh38
rs766697040 Chr.6: 6195860-6195860 on Build GRCh38
rs374616246 Chr.6: 6195901-6195901 on Build GRCh38
rs1022239357 Chr.6: 6196094-6196094 on Build GRCh38
rs550185106 Chr.6: 6195725-6195725 on Build GRCh38
rs899399406 Chr.6: 6195824-6195824 on Build GRCh38
rs752786253 Chr.6: 6195877-6195877 on Build GRCh38
rs759699312 Chr.6: 6195866-6195866 on Build GRCh38
rs531703955 Chr.6: 6195709-6195709 on Build GRCh38
rs5977 Chr.6: 6195909-6195909 on Build GRCh38
rs2274391 Chr.6: 6195693-6195693 on Build GRCh38
rs910562646 Chr.6: 6195715-6195715 on Build GRCh38
rs34625784 Chr.6: 6195908-6195909 on Build GRCh38
rs756388735 Chr.6: 6196004-6196004 on Build GRCh38
rs753688775 Chr.6: 6195888-6195888 on Build GRCh38

View Full Product Details