Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 11713468-11713974 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADTRP
Entrez Gene ID
84830
Gene Name
androgen dependent TFPI regulating protein
Gene Aliases
AIG1L, C6orf105, dJ413H6.1
UniGene
Hs.126409
Gene Chromosome Location
Chr.6: 11713523-11779628 on Build GRCh38
Transcripts
NM_001143948.1 NM_032744.3 XM_005249454.3 XM_011514956.1 XM_017011377.1

SNP Detail

SNP ID Chromosome Location
rs1013325016 Chr.6: 11713856-11713856 on Build GRCh38
rs906100416 Chr.6: 11713825-11713825 on Build GRCh38
rs904297330 Chr.6: 11713948-11713948 on Build GRCh38
rs183883573 Chr.6: 11713684-11713684 on Build GRCh38
rs548944286 Chr.6: 11713846-11713846 on Build GRCh38
rs972774690 Chr.6: 11713733-11713733 on Build GRCh38
rs139878588 Chr.6: 11713896-11713896 on Build GRCh38
rs895848065 Chr.6: 11713807-11713807 on Build GRCh38
rs920017389 Chr.6: 11713754-11713754 on Build GRCh38
rs191582343 Chr.6: 11713683-11713683 on Build GRCh38
rs760869113 Chr.6: 11713638-11713638 on Build GRCh38
rs943027384 Chr.6: 11713549-11713549 on Build GRCh38
rs1042757121 Chr.6: 11713923-11713923 on Build GRCh38
rs1009910738 Chr.6: 11713921-11713921 on Build GRCh38
rs757072071 Chr.6: 11713908-11713908 on Build GRCh38
rs931440946 Chr.6: 11713767-11713767 on Build GRCh38
rs934749523 Chr.6: 11713761-11713761 on Build GRCh38
rs974558460 Chr.6: 11713552-11713552 on Build GRCh38
rs986807609 Chr.6: 11713639-11713639 on Build GRCh38
rs368192274 Chr.6: 11713698-11713698 on Build GRCh38
rs569055900 Chr.6: 11713880-11713880 on Build GRCh38
rs1044603969 Chr.6: 11713798-11713798 on Build GRCh38
rs752774984 Chr.6: 11713606-11713606 on Build GRCh38
rs1043622229 Chr.6: 11713743-11713743 on Build GRCh38
rs34508693 Chr.6: 11713904-11713904 on Build GRCh38
rs912589764 Chr.6: 11713661-11713661 on Build GRCh38
rs535492080 Chr.6: 11713799-11713799 on Build GRCh38
rs768147604 Chr.6: 11713584-11713584 on Build GRCh38
rs911583376 Chr.6: 11713539-11713539 on Build GRCh38
rs1028841819 Chr.6: 11713567-11713567 on Build GRCh38
rs187945502 Chr.6: 11713781-11713781 on Build GRCh38
rs546552537 Chr.6: 11713724-11713724 on Build GRCh38
rs1056796313 Chr.6: 11713875-11713875 on Build GRCh38
rs527330484 Chr.6: 11713740-11713741 on Build GRCh38
rs938966144 Chr.6: 11713858-11713858 on Build GRCh38
rs968487485 Chr.6: 11713529-11713529 on Build GRCh38

View Full Product Details