Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 11769843-11770372 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADTRP
Entrez Gene ID
84830
Gene Name
androgen dependent TFPI regulating protein
Gene Aliases
AIG1L, C6orf105, dJ413H6.1
UniGene
Hs.126409
Gene Chromosome Location
Chr.6: 11713523-11779628 on Build GRCh38
Transcripts
NM_001143948.1 NM_032744.3 XM_005249454.3 XM_011514956.1 XM_011514958.2 XM_011514959.1 XM_017011377.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3697469 c.162C>T p.N54N Chr.6: 11770076-11770076 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs537601940 Chr.6: 11770131-11770131 on Build GRCh38
rs751375131 Chr.6: 11769997-11769997 on Build GRCh38
rs773408218 Chr.6: 11770090-11770090 on Build GRCh38
rs766885913 Chr.6: 11769975-11769975 on Build GRCh38
rs146305853 Chr.6: 11770213-11770213 on Build GRCh38
rs768036918 Chr.6: 11769958-11769958 on Build GRCh38
rs1055921668 Chr.6: 11770161-11770161 on Build GRCh38
rs775110997 Chr.6: 11770110-11770110 on Build GRCh38
rs9380513 Chr.6: 11769870-11769870 on Build GRCh38
rs760713300 Chr.6: 11770122-11770122 on Build GRCh38
rs542524126 Chr.6: 11770255-11770255 on Build GRCh38
rs45538938 Chr.6: 11770192-11770192 on Build GRCh38
rs370296671 Chr.6: 11770267-11770267 on Build GRCh38
rs769309052 Chr.6: 11770108-11770108 on Build GRCh38
rs748511791 Chr.6: 11770067-11770067 on Build GRCh38
rs536439611 Chr.6: 11770004-11770004 on Build GRCh38
rs937518244 Chr.6: 11770153-11770153 on Build GRCh38
rs2235390 Chr.6: 11770279-11770279 on Build GRCh38
rs749250517 Chr.6: 11770212-11770212 on Build GRCh38
rs549768146 Chr.6: 11770305-11770305 on Build GRCh38
rs754016544 Chr.6: 11770035-11770035 on Build GRCh38
rs1056928902 Chr.6: 11770168-11770168 on Build GRCh38
rs984240005 Chr.6: 11770081-11770081 on Build GRCh38
rs1025908601 Chr.6: 11770244-11770244 on Build GRCh38
rs372863145 Chr.6: 11770169-11770169 on Build GRCh38
rs563544985 Chr.6: 11769976-11769976 on Build GRCh38
rs529831292 Chr.6: 11769945-11769945 on Build GRCh38
rs199516715 Chr.6: 11770101-11770101 on Build GRCh38
rs148298172 Chr.6: 11770102-11770102 on Build GRCh38
rs972372410 Chr.6: 11770034-11770034 on Build GRCh38
rs141763751 Chr.6: 11770127-11770127 on Build GRCh38
rs999978050 Chr.6: 11769991-11769991 on Build GRCh38
rs967905380 Chr.6: 11770333-11770333 on Build GRCh38
rs144530104 Chr.6: 11769969-11769969 on Build GRCh38
rs979368824 Chr.6: 11770137-11770137 on Build GRCh38
rs139558479 Chr.6: 11770278-11770278 on Build GRCh38
rs1023804412 Chr.6: 11769981-11769981 on Build GRCh38
rs184497023 Chr.6: 11770029-11770029 on Build GRCh38
rs750332790 Chr.6: 11770018-11770018 on Build GRCh38
rs548213264 Chr.6: 11770087-11770087 on Build GRCh38
rs921847880 Chr.6: 11769932-11769932 on Build GRCh38
rs958821343 Chr.6: 11770339-11770339 on Build GRCh38
rs896618841 Chr.6: 11769972-11769972 on Build GRCh38
rs898690517 Chr.6: 11770204-11770204 on Build GRCh38
rs368684774 Chr.6: 11770052-11770052 on Build GRCh38
rs969671751 Chr.6: 11769869-11769869 on Build GRCh38
rs1048942198 Chr.6: 11769919-11769919 on Build GRCh38
rs369380870 Chr.6: 11769924-11769924 on Build GRCh38
rs966229310 Chr.6: 11770056-11770056 on Build GRCh38
rs540174240 Chr.6: 11770046-11770046 on Build GRCh38
rs778285671 Chr.6: 11770077-11770077 on Build GRCh38
rs532158096 Chr.6: 11769998-11769998 on Build GRCh38
rs983577541 Chr.6: 11770044-11770044 on Build GRCh38
rs755172853 Chr.6: 11770045-11770045 on Build GRCh38
rs556903967 Chr.6: 11770093-11770093 on Build GRCh38
rs1016882025 Chr.6: 11770324-11770324 on Build GRCh38
rs1054216831 Chr.6: 11770227-11770227 on Build GRCh38
rs898760696 Chr.6: 11769994-11769994 on Build GRCh38
rs944568377 Chr.6: 11769910-11769910 on Build GRCh38
rs920312105 Chr.6: 11769871-11769871 on Build GRCh38
rs923926951 Chr.6: 11770139-11770139 on Build GRCh38
rs765285143 Chr.6: 11770309-11770309 on Build GRCh38
rs902553818 Chr.6: 11769963-11769963 on Build GRCh38
rs1014452554 Chr.6: 11769977-11769977 on Build GRCh38
rs200268228 Chr.6: 11770102-11770102 on Build GRCh38
rs1033148482 Chr.6: 11769992-11769992 on Build GRCh38
rs201390984 Chr.6: 11770125-11770125 on Build GRCh38
rs902829301 Chr.6: 11769989-11769989 on Build GRCh38
rs764604122 Chr.6: 11770189-11770189 on Build GRCh38
rs776167052 Chr.6: 11770101-11770102 on Build GRCh38
rs573607059 Chr.6: 11770215-11770215 on Build GRCh38
rs1028813489 Chr.6: 11770330-11770330 on Build GRCh38
rs774213175 Chr.6: 11770128-11770128 on Build GRCh38
rs886080282 Chr.6: 11770277-11770278 on Build GRCh38
rs9394302 Chr.6: 11769881-11769881 on Build GRCh38
rs377064119 Chr.6: 11770294-11770294 on Build GRCh38
rs375155730 Chr.6: 11769903-11769903 on Build GRCh38
rs888836215 Chr.6: 11770237-11770237 on Build GRCh38
rs191889833 Chr.6: 11770011-11770011 on Build GRCh38
rs774293561 Chr.6: 11769902-11769902 on Build GRCh38
rs984493322 Chr.6: 11770311-11770311 on Build GRCh38
rs748377889 Chr.6: 11769938-11769938 on Build GRCh38
rs571051259 Chr.6: 11769957-11769957 on Build GRCh38
rs911791393 Chr.6: 11769867-11769867 on Build GRCh38
rs780017812 Chr.6: 11770030-11770030 on Build GRCh38
rs540005924 Chr.6: 11770193-11770193 on Build GRCh38
rs983243782 Chr.6: 11769892-11769892 on Build GRCh38
rs910062283 Chr.6: 11769943-11769943 on Build GRCh38
rs1007309502 Chr.6: 11770272-11770272 on Build GRCh38
rs764364031 Chr.6: 11769970-11769970 on Build GRCh38
rs1041423713 Chr.6: 11769985-11769985 on Build GRCh38
rs200946315 Chr.6: 11770075-11770075 on Build GRCh38
rs951415503 Chr.6: 11770310-11770310 on Build GRCh38
rs986061431 Chr.6: 11769866-11769866 on Build GRCh38

View Full Product Details