Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 14117165-14117671 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD83
Entrez Gene ID
9308
Gene Name
CD83 molecule
Gene Aliases
BL11, HB15
UniGene
Hs.595133
Gene Chromosome Location
Chr.6: 14117256-14136918 on Build GRCh38
Transcripts
NM_001040280.1 NM_001251901.1 NM_004233.3

SNP Detail

SNP ID Chromosome Location
rs895527413 Chr.6: 14117406-14117406 on Build GRCh38
rs895976495 Chr.6: 14117225-14117225 on Build GRCh38
rs1013147132 Chr.6: 14117235-14117235 on Build GRCh38
rs372220092 Chr.6: 14117499-14117499 on Build GRCh38
rs912420479 Chr.6: 14117359-14117359 on Build GRCh38
rs924685399 Chr.6: 14117614-14117614 on Build GRCh38
rs999450028 Chr.6: 14117203-14117203 on Build GRCh38
rs561189383 Chr.6: 14117434-14117434 on Build GRCh38
rs1004176940 Chr.6: 14117489-14117489 on Build GRCh38
rs1046529348 Chr.6: 14117443-14117443 on Build GRCh38
rs1002398264 Chr.6: 14117470-14117470 on Build GRCh38
rs113172053 Chr.6: 14117571-14117571 on Build GRCh38
rs1042937339 Chr.6: 14117380-14117380 on Build GRCh38
rs973386263 Chr.6: 14117337-14117337 on Build GRCh38
rs990357471 Chr.6: 14117621-14117621 on Build GRCh38
rs1044518654 Chr.6: 14117648-14117648 on Build GRCh38
rs532335048 Chr.6: 14117532-14117532 on Build GRCh38
rs1027236987 Chr.6: 14117483-14117483 on Build GRCh38
rs1042707657 Chr.6: 14117433-14117433 on Build GRCh38
rs111718881 Chr.6: 14117448-14117448 on Build GRCh38
rs954013325 Chr.6: 14117540-14117540 on Build GRCh38
rs549961010 Chr.6: 14117450-14117450 on Build GRCh38
rs963353018 Chr.6: 14117275-14117275 on Build GRCh38
rs1052694759 Chr.6: 14117601-14117601 on Build GRCh38
rs565399098 Chr.6: 14117615-14117615 on Build GRCh38
rs976576675 Chr.6: 14117609-14117609 on Build GRCh38
rs866028071 Chr.6: 14117343-14117343 on Build GRCh38
rs757975472 Chr.6: 14117192-14117192 on Build GRCh38
rs948984937 Chr.6: 14117630-14117630 on Build GRCh38
rs28384442 Chr.6: 14117216-14117216 on Build GRCh38
rs934318053 Chr.6: 14117400-14117400 on Build GRCh38
rs934579298 Chr.6: 14117567-14117567 on Build GRCh38
rs934458647 Chr.6: 14117444-14117444 on Build GRCh38
rs921384032 Chr.6: 14117533-14117533 on Build GRCh38
rs1017632507 Chr.6: 14117597-14117597 on Build GRCh38
rs966011976 Chr.6: 14117602-14117602 on Build GRCh38
rs976589123 Chr.6: 14117345-14117345 on Build GRCh38
rs769803447 Chr.6: 14117462-14117462 on Build GRCh38
rs986035124 Chr.6: 14117591-14117591 on Build GRCh38
rs765992135 Chr.6: 14117451-14117451 on Build GRCh38
rs952949469 Chr.6: 14117484-14117484 on Build GRCh38
rs982820023 Chr.6: 14117268-14117268 on Build GRCh38
rs978601701 Chr.6: 14117541-14117541 on Build GRCh38
rs1025563958 Chr.6: 14117190-14117190 on Build GRCh38
rs534336653 Chr.6: 14117651-14117651 on Build GRCh38
rs969642755 Chr.6: 14117191-14117191 on Build GRCh38
rs552018093 Chr.6: 14117547-14117547 on Build GRCh38
rs529902839 Chr.6: 14117449-14117449 on Build GRCh38
rs1023164208 Chr.6: 14117258-14117258 on Build GRCh38
rs1015622292 Chr.6: 14117520-14117520 on Build GRCh38
rs975362718 Chr.6: 14117392-14117392 on Build GRCh38
rs553086252 Chr.6: 14117589-14117600 on Build GRCh38
rs1014436760 Chr.6: 14117409-14117409 on Build GRCh38
rs558419279 Chr.6: 14117339-14117339 on Build GRCh38
rs965464386 Chr.6: 14117526-14117526 on Build GRCh38
rs949774666 Chr.6: 14117619-14117619 on Build GRCh38
rs560685332 Chr.6: 14117404-14117404 on Build GRCh38
rs543981131 Chr.6: 14117369-14117369 on Build GRCh38
rs914728324 Chr.6: 14117624-14117624 on Build GRCh38
rs575143143 Chr.6: 14117349-14117349 on Build GRCh38
rs184161495 Chr.6: 14117394-14117394 on Build GRCh38
rs916912941 Chr.6: 14117605-14117605 on Build GRCh38

View Full Product Details