Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 24426112-24426659 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPLD1
Entrez Gene ID
2822
Gene Name
glycosylphosphatidylinositol specific phospholipase D1
Gene Aliases
GPIPLD, GPIPLDM, PIGPLD, PIGPLD1, PLD
UniGene
Hs.533291
Gene Chromosome Location
Chr.6: 24425832-24495163 on Build GRCh38
Transcripts
NM_001503.3 XM_017010753.1 XM_017010754.1 NM_001286264.1 NM_001286266.1 NM_020662.3 NR_104423.1

Gene Symbol
MRS2
Entrez Gene ID
57380
Gene Name
MRS2, magnesium transporter
Gene Aliases
HPT, MRS2L
UniGene
Hs.658612
Gene Chromosome Location
Chr.6: 24402908-24426194 on Build GRCh38
Transcripts
NM_001503.3 XM_017010753.1 XM_017010754.1 NM_001286264.1 NM_001286266.1 NM_020662.3 NR_104423.1

SNP Detail

SNP ID Chromosome Location
rs933609907 Chr.6: 24426367-24426367 on Build GRCh38
rs976338015 Chr.6: 24426596-24426596 on Build GRCh38
rs763672281 Chr.6: 24426568-24426568 on Build GRCh38
rs919238667 Chr.6: 24426467-24426467 on Build GRCh38
rs930701050 Chr.6: 24426486-24426486 on Build GRCh38
rs551791814 Chr.6: 24426418-24426418 on Build GRCh38
rs943624534 Chr.6: 24426523-24426523 on Build GRCh38
rs941227341 Chr.6: 24426409-24426409 on Build GRCh38
rs2298061 Chr.6: 24426592-24426592 on Build GRCh38
rs985512131 Chr.6: 24426617-24426617 on Build GRCh38
rs919283867 Chr.6: 24426137-24426137 on Build GRCh38
rs930861272 Chr.6: 24426160-24426160 on Build GRCh38
rs961263795 Chr.6: 24426626-24426626 on Build GRCh38
rs1021008809 Chr.6: 24426267-24426267 on Build GRCh38
rs140944011 Chr.6: 24426229-24426229 on Build GRCh38
rs1042164555 Chr.6: 24426588-24426588 on Build GRCh38
rs869063760 Chr.6: 24426481-24426482 on Build GRCh38
rs569836037 Chr.6: 24426353-24426356 on Build GRCh38
rs74924371 Chr.6: 24426562-24426562 on Build GRCh38
rs35559840 Chr.6: 24426478-24426479 on Build GRCh38
rs979988906 Chr.6: 24426293-24426293 on Build GRCh38
rs2298062 Chr.6: 24426330-24426330 on Build GRCh38
rs955042230 Chr.6: 24426616-24426616 on Build GRCh38
rs10635598 Chr.6: 24426480-24426481 on Build GRCh38
rs985438482 Chr.6: 24426492-24426492 on Build GRCh38
rs968523304 Chr.6: 24426582-24426582 on Build GRCh38
rs941100766 Chr.6: 24426631-24426631 on Build GRCh38
rs1049506946 Chr.6: 24426389-24426389 on Build GRCh38
rs1009335212 Chr.6: 24426252-24426252 on Build GRCh38
rs889509966 Chr.6: 24426394-24426394 on Build GRCh38
rs763816962 Chr.6: 24426197-24426198 on Build GRCh38
rs1024784472 Chr.6: 24426615-24426615 on Build GRCh38
rs757415248 Chr.6: 24426476-24426476 on Build GRCh38
rs563246639 Chr.6: 24426167-24426167 on Build GRCh38
rs889279491 Chr.6: 24426169-24426169 on Build GRCh38
rs2298060 Chr.6: 24426594-24426594 on Build GRCh38
rs1024818462 Chr.6: 24426337-24426337 on Build GRCh38
rs991959892 Chr.6: 24426336-24426336 on Build GRCh38
rs910769403 Chr.6: 24426515-24426515 on Build GRCh38
rs766682780 Chr.6: 24426595-24426595 on Build GRCh38
rs3216033 Chr.6: 24426477-24426478 on Build GRCh38
rs990867736 Chr.6: 24426197-24426197 on Build GRCh38
rs12210975 Chr.6: 24426346-24426346 on Build GRCh38
rs530932245 Chr.6: 24426524-24426524 on Build GRCh38
rs1040526577 Chr.6: 24426171-24426171 on Build GRCh38
rs1033637500 Chr.6: 24426329-24426329 on Build GRCh38
rs945104202 Chr.6: 24426576-24426576 on Build GRCh38
rs529049590 Chr.6: 24426205-24426205 on Build GRCh38
rs897113621 Chr.6: 24426420-24426420 on Build GRCh38
rs766205741 Chr.6: 24426169-24426174 on Build GRCh38
rs750385979 Chr.6: 24426628-24426629 on Build GRCh38
rs759450152 Chr.6: 24426152-24426152 on Build GRCh38
rs968260816 Chr.6: 24426274-24426274 on Build GRCh38
rs753577021 Chr.6: 24426196-24426196 on Build GRCh38
rs571719333 Chr.6: 24426443-24426443 on Build GRCh38
rs187815194 Chr.6: 24426170-24426170 on Build GRCh38
rs1013527900 Chr.6: 24426610-24426610 on Build GRCh38
rs386698090 Chr.6: 24426592-24426594 on Build GRCh38
rs994044596 Chr.6: 24426485-24426485 on Build GRCh38
rs903544806 Chr.6: 24426305-24426305 on Build GRCh38
rs147852431 Chr.6: 24426476-24426477 on Build GRCh38
rs895026100 Chr.6: 24426606-24426606 on Build GRCh38
rs1001174790 Chr.6: 24426319-24426319 on Build GRCh38

View Full Product Details