Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 38729672-38730217 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAH8
Entrez Gene ID
1769
Gene Name
dynein axonemal heavy chain 8
Gene Aliases
ATPase, hdhc9
UniGene
Hs.520106
Gene Chromosome Location
Chr.6: 38715329-39030798 on Build GRCh38
Transcripts
NM_001206927.1 XM_011514318.2 XM_011514319.2 XM_011514320.2 XM_011514321.1 XM_017010325.1 XM_017010326.1 XM_017010327.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
160516 c.580T>C p.L194L Chr.6: 38729956-38729956 on Build GRCh38
7138373 c.542_543insT p.A184fs*6 Chr.6: 38729918-38729919 on Build GRCh38
7260364 c.529G>A p.E177K Chr.6: 38729905-38729905 on Build GRCh38
7138372 c.542_543insT p.A184fs*6 Chr.6: 38729918-38729919 on Build GRCh38
7260365 c.529G>A p.E177K Chr.6: 38729905-38729905 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs765889198 Chr.6: 38729827-38729827 on Build GRCh38
rs566236960 Chr.6: 38729844-38729844 on Build GRCh38
rs745977267 Chr.6: 38729893-38729893 on Build GRCh38
rs755078339 Chr.6: 38730031-38730031 on Build GRCh38
rs758497061 Chr.6: 38730138-38730138 on Build GRCh38
rs764404672 Chr.6: 38729865-38729865 on Build GRCh38
rs1032623235 Chr.6: 38729949-38729949 on Build GRCh38
rs778634615 Chr.6: 38729943-38729943 on Build GRCh38
rs980394378 Chr.6: 38729978-38729978 on Build GRCh38
rs753974910 Chr.6: 38729934-38729934 on Build GRCh38
rs1051590151 Chr.6: 38730060-38730060 on Build GRCh38
rs1038951930 Chr.6: 38730085-38730085 on Build GRCh38
rs368460226 Chr.6: 38729927-38729927 on Build GRCh38
rs1038010923 Chr.6: 38730102-38730102 on Build GRCh38
rs539090595 Chr.6: 38729782-38729782 on Build GRCh38
rs35954080 Chr.6: 38730058-38730059 on Build GRCh38
rs550885197 Chr.6: 38729729-38729729 on Build GRCh38
rs771905434 Chr.6: 38729973-38729973 on Build GRCh38
rs201672902 Chr.6: 38729928-38729928 on Build GRCh38
rs146727246 Chr.6: 38730017-38730017 on Build GRCh38
rs766201465 Chr.6: 38729995-38730001 on Build GRCh38
rs56284095 Chr.6: 38730063-38730063 on Build GRCh38
rs756356607 Chr.6: 38729907-38729907 on Build GRCh38
rs570330647 Chr.6: 38729753-38729753 on Build GRCh38
rs748063803 Chr.6: 38729958-38729958 on Build GRCh38
rs12212784 Chr.6: 38730068-38730068 on Build GRCh38
rs143348816 Chr.6: 38729976-38729976 on Build GRCh38
rs886324215 Chr.6: 38729778-38729778 on Build GRCh38
rs760195926 Chr.6: 38730014-38730014 on Build GRCh38
rs374870194 Chr.6: 38729824-38729824 on Build GRCh38
rs927435427 Chr.6: 38730114-38730114 on Build GRCh38
rs900467621 Chr.6: 38730097-38730097 on Build GRCh38
rs890265768 Chr.6: 38729801-38729801 on Build GRCh38
rs920548207 Chr.6: 38730150-38730150 on Build GRCh38
rs375701588 Chr.6: 38729777-38729777 on Build GRCh38
rs766245136 Chr.6: 38729846-38729846 on Build GRCh38
rs190192678 Chr.6: 38730032-38730032 on Build GRCh38
rs936972736 Chr.6: 38730152-38730152 on Build GRCh38
rs758736443 Chr.6: 38729854-38729854 on Build GRCh38
rs529520688 Chr.6: 38729708-38729708 on Build GRCh38
rs1047585695 Chr.6: 38729788-38729788 on Build GRCh38
rs770474063 Chr.6: 38730015-38730015 on Build GRCh38
rs941314765 Chr.6: 38729737-38729737 on Build GRCh38
rs757760799 Chr.6: 38729875-38729875 on Build GRCh38
rs917448158 Chr.6: 38729945-38729945 on Build GRCh38
rs752101662 Chr.6: 38729870-38729870 on Build GRCh38
rs1000062856 Chr.6: 38730098-38730098 on Build GRCh38
rs933132633 Chr.6: 38729763-38729763 on Build GRCh38
rs759114831 Chr.6: 38730036-38730036 on Build GRCh38
rs578067889 Chr.6: 38730140-38730140 on Build GRCh38
rs139315945 Chr.6: 38729726-38729726 on Build GRCh38
rs1008831058 Chr.6: 38729821-38729821 on Build GRCh38
rs940609148 Chr.6: 38729781-38729781 on Build GRCh38
rs566967577 Chr.6: 38730043-38730043 on Build GRCh38
rs759251439 Chr.6: 38730025-38730025 on Build GRCh38
rs994246126 Chr.6: 38729783-38729783 on Build GRCh38
rs1032638497 Chr.6: 38729918-38729919 on Build GRCh38
rs936719455 Chr.6: 38729696-38729696 on Build GRCh38
rs1032670688 Chr.6: 38730124-38730124 on Build GRCh38

View Full Product Details