Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 154199281-154199779 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf189
Entrez Gene ID
388701
Gene Name
chromosome 1 open reading frame 189
Gene Aliases
-
UniGene
Hs.104967
Gene Chromosome Location
Chr.1: 154199085-154206365 on Build GRCh38
Transcripts
NM_001010979.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4667355 c.278G>A p.G93D Chr.1: 154199455-154199455 on Build GRCh38
896524 c.235G>C p.E79Q Chr.1: 154199498-154199498 on Build GRCh38
1683790 c.294_295delAG p.R100fs*>2 Chr.1: 154199438-154199439 on Build GRCh38
2208610 c.299G>T p.R100I Chr.1: 154199434-154199434 on Build GRCh38
896525 c.229C>A p.L77M Chr.1: 154199504-154199504 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs933767180 Chr.1: 154199664-154199664 on Build GRCh38
rs972931925 Chr.1: 154199589-154199589 on Build GRCh38
rs749210103 Chr.1: 154199376-154199376 on Build GRCh38
rs751431792 Chr.1: 154199499-154199499 on Build GRCh38
rs779098654 Chr.1: 154199626-154199626 on Build GRCh38
rs538733528 Chr.1: 154199358-154199358 on Build GRCh38
rs543160626 Chr.1: 154199508-154199508 on Build GRCh38
rs372885193 Chr.1: 154199448-154199448 on Build GRCh38
rs778656718 Chr.1: 154199381-154199381 on Build GRCh38
rs993042279 Chr.1: 154199469-154199469 on Build GRCh38
rs760169863 Chr.1: 154199412-154199412 on Build GRCh38
rs774863256 Chr.1: 154199395-154199395 on Build GRCh38
rs369367555 Chr.1: 154199402-154199402 on Build GRCh38
rs905202032 Chr.1: 154199422-154199422 on Build GRCh38
rs202075988 Chr.1: 154199433-154199434 on Build GRCh38
rs769048417 Chr.1: 154199441-154199441 on Build GRCh38
rs563347329 Chr.1: 154199740-154199740 on Build GRCh38
rs551208298 Chr.1: 154199738-154199738 on Build GRCh38
rs149614781 Chr.1: 154199335-154199335 on Build GRCh38
rs768264191 Chr.1: 154199436-154199436 on Build GRCh38
rs1052644122 Chr.1: 154199336-154199336 on Build GRCh38
rs369537164 Chr.1: 154199562-154199562 on Build GRCh38
rs117716884 Chr.1: 154199350-154199350 on Build GRCh38
rs376125797 Chr.1: 154199454-154199454 on Build GRCh38
rs745526744 Chr.1: 154199385-154199385 on Build GRCh38
rs559374635 Chr.1: 154199667-154199667 on Build GRCh38
rs541055444 Chr.1: 154199642-154199642 on Build GRCh38
rs558027833 Chr.1: 154199524-154199524 on Build GRCh38
rs771713837 Chr.1: 154199388-154199388 on Build GRCh38
rs751604138 Chr.1: 154199483-154199483 on Build GRCh38
rs776262778 Chr.1: 154199553-154199553 on Build GRCh38
rs770752085 Chr.1: 154199379-154199379 on Build GRCh38
rs757136330 Chr.1: 154199513-154199513 on Build GRCh38
rs143378215 Chr.1: 154199478-154199478 on Build GRCh38
rs767553621 Chr.1: 154199490-154199490 on Build GRCh38
rs140739159 Chr.1: 154199530-154199530 on Build GRCh38
rs138248174 Chr.1: 154199304-154199304 on Build GRCh38
rs763116420 Chr.1: 154199473-154199473 on Build GRCh38
rs755109450 Chr.1: 154199485-154199485 on Build GRCh38
rs148380319 Chr.1: 154199714-154199714 on Build GRCh38
rs749488994 Chr.1: 154199316-154199316 on Build GRCh38
rs935422244 Chr.1: 154199746-154199746 on Build GRCh38
rs762382622 Chr.1: 154199442-154199442 on Build GRCh38
rs557393252 Chr.1: 154199366-154199366 on Build GRCh38
rs911155400 Chr.1: 154199670-154199670 on Build GRCh38
rs985324481 Chr.1: 154199651-154199651 on Build GRCh38
rs745606878 Chr.1: 154199519-154199519 on Build GRCh38
rs1043714858 Chr.1: 154199726-154199726 on Build GRCh38
rs199607542 Chr.1: 154199551-154199551 on Build GRCh38
rs778602301 Chr.1: 154199517-154199517 on Build GRCh38
rs768325316 Chr.1: 154199549-154199549 on Build GRCh38
rs202093283 Chr.1: 154199525-154199525 on Build GRCh38
rs955411726 Chr.1: 154199648-154199648 on Build GRCh38
rs552949398 Chr.1: 154199340-154199340 on Build GRCh38

View Full Product Details