Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 41937081-41937583 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCND3
Entrez Gene ID
896
Gene Name
cyclin D3
Gene Aliases
-
UniGene
Hs.534307
Gene Chromosome Location
Chr.6: 41934933-42048894 on Build GRCh38
Transcripts
NM_001136017.3 NM_001136125.2 NM_001136126.2 NM_001287427.1 NM_001287434.1 NM_001760.4 XM_011514971.1 XM_011514972.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3874050 c.559G>A p.A187T Chr.6: 41937250-41937250 on Build GRCh38
5663838 c.511C>G p.P171A Chr.6: 41937298-41937298 on Build GRCh38
1079291 c.574G>T p.D192Y Chr.6: 41937235-41937235 on Build GRCh38
4006504 c.465G>A p.V155V Chr.6: 41937344-41937344 on Build GRCh38
5764264 c.499C>T p.R167W Chr.6: 41937310-41937310 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373981901 Chr.6: 41937335-41937335 on Build GRCh38
rs773858961 Chr.6: 41937165-41937165 on Build GRCh38
rs753011743 Chr.6: 41937325-41937325 on Build GRCh38
rs370472300 Chr.6: 41937222-41937222 on Build GRCh38
rs760346974 Chr.6: 41937429-41937429 on Build GRCh38
rs775839403 Chr.6: 41937442-41937442 on Build GRCh38
rs374294883 Chr.6: 41937288-41937288 on Build GRCh38
rs750688882 Chr.6: 41937349-41937349 on Build GRCh38
rs762592347 Chr.6: 41937149-41937149 on Build GRCh38
rs368245852 Chr.6: 41937303-41937303 on Build GRCh38
rs539052412 Chr.6: 41937287-41937287 on Build GRCh38
rs751658895 Chr.6: 41937328-41937328 on Build GRCh38
rs745611061 Chr.6: 41937142-41937142 on Build GRCh38
rs753175357 Chr.6: 41937196-41937196 on Build GRCh38
rs943131258 Chr.6: 41937120-41937120 on Build GRCh38
rs749257225 Chr.6: 41937294-41937294 on Build GRCh38
rs766288220 Chr.6: 41937517-41937517 on Build GRCh38
rs754219667 Chr.6: 41937190-41937190 on Build GRCh38
rs1039859949 Chr.6: 41937134-41937134 on Build GRCh38
rs1053881507 Chr.6: 41937531-41937531 on Build GRCh38
rs371334921 Chr.6: 41937405-41937405 on Build GRCh38
rs747663834 Chr.6: 41937412-41937412 on Build GRCh38
rs905249311 Chr.6: 41937208-41937208 on Build GRCh38
rs764555870 Chr.6: 41937444-41937444 on Build GRCh38
rs769908615 Chr.6: 41937143-41937143 on Build GRCh38
rs772900960 Chr.6: 41937310-41937310 on Build GRCh38
rs761382151 Chr.6: 41937166-41937166 on Build GRCh38
rs974504163 Chr.6: 41937503-41937503 on Build GRCh38
rs915455701 Chr.6: 41937508-41937508 on Build GRCh38
rs781308576 Chr.6: 41937342-41937342 on Build GRCh38
rs945630210 Chr.6: 41937547-41937547 on Build GRCh38
rs753300223 Chr.6: 41937344-41937345 on Build GRCh38
rs138467679 Chr.6: 41937295-41937295 on Build GRCh38
rs768778584 Chr.6: 41937403-41937403 on Build GRCh38
rs763305722 Chr.6: 41937443-41937443 on Build GRCh38
rs767127766 Chr.6: 41937179-41937179 on Build GRCh38
rs964473984 Chr.6: 41937492-41937492 on Build GRCh38
rs376161353 Chr.6: 41937219-41937219 on Build GRCh38
rs780004862 Chr.6: 41937363-41937363 on Build GRCh38
rs911575920 Chr.6: 41937487-41937487 on Build GRCh38
rs950696357 Chr.6: 41937462-41937462 on Build GRCh38
rs3218099 Chr.6: 41937218-41937218 on Build GRCh38
rs775544672 Chr.6: 41937145-41937145 on Build GRCh38
rs761379615 Chr.6: 41937305-41937305 on Build GRCh38
rs987132565 Chr.6: 41937469-41937469 on Build GRCh38
rs550895211 Chr.6: 41937210-41937210 on Build GRCh38
rs554847316 Chr.6: 41937465-41937465 on Build GRCh38
rs755360211 Chr.6: 41937191-41937191 on Build GRCh38
rs921456851 Chr.6: 41937176-41937176 on Build GRCh38
rs759938253 Chr.6: 41937314-41937314 on Build GRCh38
rs3218097 Chr.6: 41937537-41937537 on Build GRCh38
rs763116221 Chr.6: 41937258-41937258 on Build GRCh38
rs763464647 Chr.6: 41937326-41937326 on Build GRCh38
rs147099303 Chr.6: 41937291-41937291 on Build GRCh38
rs765837668 Chr.6: 41937192-41937193 on Build GRCh38
rs763728183 Chr.6: 41937158-41937158 on Build GRCh38
rs764185204 Chr.6: 41937378-41937378 on Build GRCh38
rs936805592 Chr.6: 41937520-41937520 on Build GRCh38
rs758756366 Chr.6: 41937231-41937231 on Build GRCh38
rs765624800 Chr.6: 41937316-41937316 on Build GRCh38
rs946664067 Chr.6: 41937127-41937127 on Build GRCh38
rs2479717 Chr.6: 41937436-41937436 on Build GRCh38
rs976973321 Chr.6: 41937404-41937404 on Build GRCh38
rs756354784 Chr.6: 41937361-41937361 on Build GRCh38
rs1045174139 Chr.6: 41937184-41937184 on Build GRCh38
rs765784325 Chr.6: 41937195-41937195 on Build GRCh38
rs3218098 Chr.6: 41937416-41937416 on Build GRCh38
rs776305334 Chr.6: 41937135-41937135 on Build GRCh38
rs921424356 Chr.6: 41937510-41937510 on Build GRCh38
rs749019972 Chr.6: 41937379-41937379 on Build GRCh38
rs149279084 Chr.6: 41937309-41937309 on Build GRCh38
rs915235464 Chr.6: 41937113-41937113 on Build GRCh38
rs764596308 Chr.6: 41937327-41937327 on Build GRCh38
rs911607333 Chr.6: 41937110-41937110 on Build GRCh38
rs772560568 Chr.6: 41937427-41937427 on Build GRCh38
rs752073158 Chr.6: 41937446-41937446 on Build GRCh38

View Full Product Details