Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 42962125-42962630 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNPY3-GNMT
Entrez Gene ID
107080644
Gene Name
CNPY3-GNMT readthrough
Gene Aliases
-
UniGene
Hs.144914
Gene Chromosome Location
Chr.6: 42929130-42963880 on Build GRCh38
Transcripts
NM_001318856.1 NM_001318857.1 NM_001318858.1 NR_134890.1 NR_134891.1 NR_134892.1 NM_001318865.1 NM_018960.5 NR_134899.1

Gene Symbol
GNMT
Entrez Gene ID
27232
Gene Name
glycine N-methyltransferase
Gene Aliases
HEL-S-182mP
UniGene
Hs.144914
Gene Chromosome Location
Chr.6: 42960754-42963880 on Build GRCh38
Transcripts
NM_001318856.1 NM_001318857.1 NM_001318858.1 NR_134890.1 NR_134891.1 NR_134892.1 NM_001318865.1 NM_018960.5 NR_134899.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
484086 c.260C>T p.A87V Chr.6: 42962265-42962265 on Build GRCh38
7411406 c.316C>A p.P106T Chr.6: 42962321-42962321 on Build GRCh38
3777679 c.285A>G p.A95A Chr.6: 42962290-42962290 on Build GRCh38
4534679 c.210G>A p.V70V Chr.6: 42962215-42962215 on Build GRCh38
3734795 c.260delC p.S88fs*5 Chr.6: 42962265-42962265 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746358380 Chr.6: 42962193-42962193 on Build GRCh38
rs535188125 Chr.6: 42962324-42962324 on Build GRCh38
rs572545021 Chr.6: 42962378-42962378 on Build GRCh38
rs997449610 Chr.6: 42962534-42962535 on Build GRCh38
rs766302988 Chr.6: 42962211-42962211 on Build GRCh38
rs752276656 Chr.6: 42962284-42962284 on Build GRCh38
rs1028072024 Chr.6: 42962553-42962553 on Build GRCh38
rs758121236 Chr.6: 42962301-42962301 on Build GRCh38
rs186962635 Chr.6: 42962177-42962177 on Build GRCh38
rs753572605 Chr.6: 42962237-42962237 on Build GRCh38
rs762374121 Chr.6: 42962256-42962256 on Build GRCh38
rs774442803 Chr.6: 42962204-42962204 on Build GRCh38
rs372514666 Chr.6: 42962196-42962196 on Build GRCh38
rs1028520692 Chr.6: 42962277-42962277 on Build GRCh38
rs901392533 Chr.6: 42962289-42962289 on Build GRCh38
rs766324973 Chr.6: 42962232-42962232 on Build GRCh38
rs143966932 Chr.6: 42962179-42962179 on Build GRCh38
rs777594419 Chr.6: 42962365-42962365 on Build GRCh38
rs751060218 Chr.6: 42962369-42962369 on Build GRCh38
rs966386321 Chr.6: 42962589-42962589 on Build GRCh38
rs543707887 Chr.6: 42962446-42962446 on Build GRCh38
rs572170490 Chr.6: 42962329-42962329 on Build GRCh38
rs756091149 Chr.6: 42962165-42962165 on Build GRCh38
rs763438470 Chr.6: 42962199-42962199 on Build GRCh38
rs138577594 Chr.6: 42962151-42962151 on Build GRCh38
rs113082620 Chr.6: 42962540-42962540 on Build GRCh38
rs773227193 Chr.6: 42962322-42962322 on Build GRCh38
rs781167464 Chr.6: 42962313-42962313 on Build GRCh38
rs558770242 Chr.6: 42962364-42962364 on Build GRCh38
rs377520419 Chr.6: 42962203-42962203 on Build GRCh38
rs780720604 Chr.6: 42962310-42962310 on Build GRCh38
rs985880457 Chr.6: 42962421-42962421 on Build GRCh38
rs778831569 Chr.6: 42962254-42962254 on Build GRCh38
rs749442227 Chr.6: 42962554-42962554 on Build GRCh38
rs760980193 Chr.6: 42962511-42962511 on Build GRCh38
rs993250212 Chr.6: 42962461-42962461 on Build GRCh38
rs541524958 Chr.6: 42962601-42962601 on Build GRCh38
rs374221964 Chr.6: 42962382-42962382 on Build GRCh38
rs3800292 Chr.6: 42962571-42962571 on Build GRCh38
rs1015791139 Chr.6: 42962262-42962262 on Build GRCh38
rs888598421 Chr.6: 42962215-42962215 on Build GRCh38
rs557243794 Chr.6: 42962468-42962468 on Build GRCh38
rs181496341 Chr.6: 42962157-42962157 on Build GRCh38
rs767483781 Chr.6: 42962212-42962212 on Build GRCh38
rs576938593 Chr.6: 42962482-42962482 on Build GRCh38
rs751482570 Chr.6: 42962595-42962595 on Build GRCh38
rs554957004 Chr.6: 42962430-42962430 on Build GRCh38
rs749111811 Chr.6: 42962166-42962166 on Build GRCh38
rs1017066928 Chr.6: 42962189-42962189 on Build GRCh38
rs749747704 Chr.6: 42962383-42962383 on Build GRCh38
rs149056383 Chr.6: 42962330-42962330 on Build GRCh38
rs755468965 Chr.6: 42962361-42962361 on Build GRCh38
rs1030434474 Chr.6: 42962592-42962592 on Build GRCh38
rs59395427 Chr.6: 42962247-42962247 on Build GRCh38
rs752579906 Chr.6: 42962358-42962358 on Build GRCh38
rs574896156 Chr.6: 42962445-42962445 on Build GRCh38
rs1007989413 Chr.6: 42962560-42962560 on Build GRCh38
rs1048462684 Chr.6: 42962509-42962509 on Build GRCh38
rs770379656 Chr.6: 42962309-42962309 on Build GRCh38
rs138006194 Chr.6: 42962317-42962317 on Build GRCh38
rs1037122954 Chr.6: 42962513-42962513 on Build GRCh38
rs760563979 Chr.6: 42962222-42962222 on Build GRCh38
rs938587124 Chr.6: 42962450-42962450 on Build GRCh38
rs775961531 Chr.6: 42962197-42962197 on Build GRCh38
rs754989144 Chr.6: 42962566-42962566 on Build GRCh38
rs116551815 Chr.6: 42962512-42962512 on Build GRCh38
rs924523011 Chr.6: 42962447-42962447 on Build GRCh38
rs546013977 Chr.6: 42962502-42962502 on Build GRCh38
rs778440392 Chr.6: 42962175-42962175 on Build GRCh38
rs772369673 Chr.6: 42962320-42962320 on Build GRCh38
rs779466342 Chr.6: 42962379-42962379 on Build GRCh38
rs761907772 Chr.6: 42962318-42962318 on Build GRCh38
rs915903013 Chr.6: 42962494-42962494 on Build GRCh38
rs759573326 Chr.6: 42962335-42962335 on Build GRCh38
rs764916813 Chr.6: 42962343-42962343 on Build GRCh38
rs201470823 Chr.6: 42962213-42962213 on Build GRCh38
rs894451230 Chr.6: 42962148-42962148 on Build GRCh38
rs758388204 Chr.6: 42962207-42962207 on Build GRCh38
rs746552607 Chr.6: 42962306-42962306 on Build GRCh38
rs769257865 Chr.6: 42962187-42962187 on Build GRCh38
rs749599022 Chr.6: 42962312-42962312 on Build GRCh38
rs754730549 Chr.6: 42962253-42962253 on Build GRCh38
rs570113198 Chr.6: 42962323-42962323 on Build GRCh38
rs535014756 Chr.6: 42962420-42962420 on Build GRCh38
rs753965293 Chr.6: 42962354-42962354 on Build GRCh38

View Full Product Details