Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 44169765-44170175 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPN11
Entrez Gene ID
11131
Gene Name
calpain 11
Gene Aliases
calpain11
UniGene
Hs.225953
Gene Chromosome Location
Chr.6: 44158811-44184402 on Build GRCh38
Transcripts
NM_007058.3 XM_006714984.1 XM_006714985.1 XM_006714987.1 XM_011514274.1 XM_011514275.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
742832 c.388G>C p.D130H Chr.6: 44169954-44169954 on Build GRCh38
5405415 c.402G>A p.G134G Chr.6: 44169968-44169968 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs760060167 Chr.6: 44169983-44169983 on Build GRCh38
rs76813039 Chr.6: 44170096-44170096 on Build GRCh38
rs1003186043 Chr.6: 44169924-44169924 on Build GRCh38
rs377006509 Chr.6: 44169843-44169843 on Build GRCh38
rs757187023 Chr.6: 44169956-44169956 on Build GRCh38
rs900316738 Chr.6: 44170054-44170054 on Build GRCh38
rs751623924 Chr.6: 44169954-44169954 on Build GRCh38
rs575505895 Chr.6: 44170136-44170136 on Build GRCh38
rs781216695 Chr.6: 44169958-44169958 on Build GRCh38
rs531047829 Chr.6: 44169981-44169981 on Build GRCh38
rs764350955 Chr.6: 44170009-44170009 on Build GRCh38
rs769525350 Chr.6: 44169860-44169860 on Build GRCh38
rs774383665 Chr.6: 44170016-44170016 on Build GRCh38
rs797001244 Chr.6: 44169873-44169873 on Build GRCh38
rs917497651 Chr.6: 44169825-44169825 on Build GRCh38
rs764144671 Chr.6: 44169949-44169949 on Build GRCh38
rs185617031 Chr.6: 44169957-44169957 on Build GRCh38
rs763251773 Chr.6: 44169948-44169948 on Build GRCh38
rs768066960 Chr.6: 44169873-44169873 on Build GRCh38
rs150945594 Chr.6: 44170097-44170097 on Build GRCh38
rs927436826 Chr.6: 44169876-44169876 on Build GRCh38
rs749000314 Chr.6: 44169969-44169969 on Build GRCh38
rs755910697 Chr.6: 44169967-44169967 on Build GRCh38
rs748875727 Chr.6: 44169868-44169868 on Build GRCh38
rs765629434 Chr.6: 44169932-44169932 on Build GRCh38
rs752979547 Chr.6: 44169941-44169941 on Build GRCh38
rs1014570863 Chr.6: 44170106-44170106 on Build GRCh38
rs980246934 Chr.6: 44169872-44169872 on Build GRCh38
rs778328078 Chr.6: 44169975-44169975 on Build GRCh38
rs1013166903 Chr.6: 44169795-44169795 on Build GRCh38
rs911792130 Chr.6: 44169966-44169966 on Build GRCh38
rs1045948957 Chr.6: 44170082-44170082 on Build GRCh38
rs761988787 Chr.6: 44170021-44170021 on Build GRCh38
rs189429774 Chr.6: 44169963-44169963 on Build GRCh38
rs1009861199 Chr.6: 44170153-44170153 on Build GRCh38
rs1038906050 Chr.6: 44170045-44170045 on Build GRCh38
rs1032754844 Chr.6: 44170113-44170113 on Build GRCh38
rs1047714726 Chr.6: 44169995-44169995 on Build GRCh38
rs2275745 Chr.6: 44169926-44169926 on Build GRCh38
rs779709745 Chr.6: 44169866-44169866 on Build GRCh38
rs773900019 Chr.6: 44169879-44169879 on Build GRCh38
rs763122090 Chr.6: 44170004-44170004 on Build GRCh38
rs924630047 Chr.6: 44169984-44169984 on Build GRCh38
rs762550958 Chr.6: 44169961-44169961 on Build GRCh38
rs567897947 Chr.6: 44170091-44170091 on Build GRCh38
rs769783832 Chr.6: 44170052-44170052 on Build GRCh38
rs771437989 Chr.6: 44169914-44169914 on Build GRCh38
rs777346104 Chr.6: 44170040-44170040 on Build GRCh38
rs747673026 Chr.6: 44169980-44169980 on Build GRCh38
rs181949159 Chr.6: 44169998-44169998 on Build GRCh38
rs768352317 Chr.6: 44169974-44169974 on Build GRCh38
rs944634311 Chr.6: 44169971-44169971 on Build GRCh38
rs867930235 Chr.6: 44170034-44170034 on Build GRCh38
rs559211706 Chr.6: 44169805-44169805 on Build GRCh38
rs779769200 Chr.6: 44169968-44169968 on Build GRCh38
rs948826050 Chr.6: 44170051-44170051 on Build GRCh38
rs761153528 Chr.6: 44169899-44169899 on Build GRCh38

View Full Product Details