Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 52752765-52753274 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GSTA2
Entrez Gene ID
2939
Gene Name
glutathione S-transferase alpha 2
Gene Aliases
GST2, GSTA2-2, GTA2, GTH2
UniGene
Hs.94107
Gene Chromosome Location
Chr.6: 52750087-52763563 on Build GRCh38
Transcripts
NM_000846.4 XM_011514532.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3628519 c.319C>T p.L107F Chr.6: 52752949-52752949 on Build GRCh38
5870910 c.409delG p.E137fs*4 Chr.6: 52752859-52752859 on Build GRCh38
4823572 c.307G>A p.G103S Chr.6: 52752961-52752961 on Build GRCh38
3715553 c.318C>T p.I106I Chr.6: 52752950-52752950 on Build GRCh38
4004076 c.328C>T p.P110S Chr.6: 52752940-52752940 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367642534 Chr.6: 52752842-52752842 on Build GRCh38
rs369381698 Chr.6: 52752821-52752821 on Build GRCh38
rs115929428 Chr.6: 52753248-52753248 on Build GRCh38
rs192840784 Chr.6: 52752849-52752849 on Build GRCh38
rs200252041 Chr.6: 52752917-52752917 on Build GRCh38
rs534645885 Chr.6: 52753183-52753183 on Build GRCh38
rs371439458 Chr.6: 52753133-52753163 on Build GRCh38
rs915848432 Chr.6: 52752804-52752804 on Build GRCh38
rs746271498 Chr.6: 52752999-52752999 on Build GRCh38
rs761363561 Chr.6: 52752836-52752836 on Build GRCh38
rs780205236 Chr.6: 52752803-52752803 on Build GRCh38
rs766085856 Chr.6: 52752967-52752967 on Build GRCh38
rs746191882 Chr.6: 52752914-52752914 on Build GRCh38
rs560897862 Chr.6: 52752802-52752802 on Build GRCh38
rs767399759 Chr.6: 52752980-52752980 on Build GRCh38
rs373267358 Chr.6: 52752814-52752814 on Build GRCh38
rs368197497 Chr.6: 52753028-52753028 on Build GRCh38
rs773906172 Chr.6: 52752935-52752935 on Build GRCh38
rs189368911 Chr.6: 52753099-52753099 on Build GRCh38
rs60332313 Chr.6: 52752894-52752894 on Build GRCh38
rs771085266 Chr.6: 52752895-52752895 on Build GRCh38
rs533963800 Chr.6: 52753067-52753067 on Build GRCh38
rs6935419 Chr.6: 52753111-52753111 on Build GRCh38
rs768250692 Chr.6: 52752825-52752826 on Build GRCh38
rs773466273 Chr.6: 52752816-52752816 on Build GRCh38
rs202200168 Chr.6: 52752905-52752905 on Build GRCh38
rs766288786 Chr.6: 52752871-52752871 on Build GRCh38
rs2749002 Chr.6: 52753155-52753155 on Build GRCh38
rs200848146 Chr.6: 52752872-52752872 on Build GRCh38
rs371117739 Chr.6: 52752840-52752840 on Build GRCh38
rs911666229 Chr.6: 52753149-52753149 on Build GRCh38
rs997021993 Chr.6: 52752904-52752904 on Build GRCh38
rs142078893 Chr.6: 52752996-52752996 on Build GRCh38
rs1016074436 Chr.6: 52752799-52752799 on Build GRCh38
rs888665771 Chr.6: 52753211-52753211 on Build GRCh38
rs769333289 Chr.6: 52752938-52752938 on Build GRCh38
rs568075839 Chr.6: 52753038-52753038 on Build GRCh38
rs921468533 Chr.6: 52753168-52753168 on Build GRCh38
rs6458864 Chr.6: 52752812-52752812 on Build GRCh38
rs551872534 Chr.6: 52752978-52752978 on Build GRCh38
rs538448319 Chr.6: 52753125-52753125 on Build GRCh38
rs775092149 Chr.6: 52753047-52753047 on Build GRCh38
rs546472526 Chr.6: 52753126-52753156 on Build GRCh38
rs780780470 Chr.6: 52752798-52752798 on Build GRCh38
rs558518583 Chr.6: 52753029-52753029 on Build GRCh38
rs747589919 Chr.6: 52752942-52752944 on Build GRCh38
rs1048623323 Chr.6: 52752918-52752918 on Build GRCh38
rs2234733 Chr.6: 52752852-52752852 on Build GRCh38
rs768002765 Chr.6: 52752826-52752826 on Build GRCh38
rs151112301 Chr.6: 52752877-52752877 on Build GRCh38
rs142983962 Chr.6: 52752869-52752869 on Build GRCh38
rs771582037 Chr.6: 52752948-52752949 on Build GRCh38
rs114762445 Chr.6: 52753220-52753220 on Build GRCh38
rs370446598 Chr.6: 52753020-52753020 on Build GRCh38
rs267601074 Chr.6: 52752950-52752950 on Build GRCh38
rs771732026 Chr.6: 52753143-52753143 on Build GRCh38
rs767333868 Chr.6: 52753040-52753040 on Build GRCh38
rs753849765 Chr.6: 52752989-52752989 on Build GRCh38
rs575354432 Chr.6: 52753161-52753161 on Build GRCh38
rs761059131 Chr.6: 52753209-52753209 on Build GRCh38
rs752679873 Chr.6: 52752981-52752981 on Build GRCh38
rs777927079 Chr.6: 52752890-52752890 on Build GRCh38
rs750085076 Chr.6: 52752867-52752867 on Build GRCh38
rs541882679 Chr.6: 52753005-52753005 on Build GRCh38
rs2234951 Chr.6: 52752940-52752940 on Build GRCh38
rs1032910778 Chr.6: 52753045-52753045 on Build GRCh38
rs759348739 Chr.6: 52752973-52752973 on Build GRCh38
rs376417930 Chr.6: 52752820-52752820 on Build GRCh38
rs976898957 Chr.6: 52753156-52753156 on Build GRCh38
rs959926645 Chr.6: 52752800-52752800 on Build GRCh38
rs760050835 Chr.6: 52752822-52752822 on Build GRCh38
rs2180314 Chr.6: 52752933-52752933 on Build GRCh38
rs772479719 Chr.6: 52753014-52753014 on Build GRCh38
rs776036844 Chr.6: 52752830-52752830 on Build GRCh38
rs748944480 Chr.6: 52752823-52752824 on Build GRCh38
rs756208541 Chr.6: 52752985-52752985 on Build GRCh38
rs746928512 Chr.6: 52752807-52752807 on Build GRCh38
rs13212306 Chr.6: 52752866-52752866 on Build GRCh38
rs768974457 Chr.6: 52753021-52753021 on Build GRCh38
rs200132206 Chr.6: 52752903-52752903 on Build GRCh38
rs749696024 Chr.6: 52752817-52752817 on Build GRCh38
rs757502057 Chr.6: 52752991-52752991 on Build GRCh38
rs772877618 Chr.6: 52753024-52753024 on Build GRCh38
rs754993807 Chr.6: 52752876-52752876 on Build GRCh38
rs779255900 Chr.6: 52752902-52752902 on Build GRCh38
rs988855222 Chr.6: 52753130-52753130 on Build GRCh38
rs376074279 Chr.6: 52752971-52752971 on Build GRCh38
rs776034199 Chr.6: 52752922-52752922 on Build GRCh38
rs755896808 Chr.6: 52752847-52752847 on Build GRCh38
rs762696709 Chr.6: 52752947-52752947 on Build GRCh38
rs772478820 Chr.6: 52752919-52752919 on Build GRCh38
rs754795118 Chr.6: 52752793-52752793 on Build GRCh38
rs752773504 Chr.6: 52752885-52752885 on Build GRCh38
rs777924051 Chr.6: 52752988-52752988 on Build GRCh38

View Full Product Details