Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 52763304-52763822 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GSTA2
Entrez Gene ID
2939
Gene Name
glutathione S-transferase alpha 2
Gene Aliases
GST2, GSTA2-2, GTA2, GTH2
UniGene
Hs.94107
Gene Chromosome Location
Chr.6: 52750087-52763563 on Build GRCh38
Transcripts
NM_000846.4

SNP Detail

SNP ID Chromosome Location
rs927188266 Chr.6: 52763421-52763421 on Build GRCh38
rs111812540 Chr.6: 52763689-52763689 on Build GRCh38
rs990250616 Chr.6: 52763462-52763462 on Build GRCh38
rs73740554 Chr.6: 52763479-52763479 on Build GRCh38
rs374783838 Chr.6: 52763770-52763770 on Build GRCh38
rs976255841 Chr.6: 52763754-52763754 on Build GRCh38
rs778375763 Chr.6: 52763681-52763681 on Build GRCh38
rs2749019 Chr.6: 52763753-52763753 on Build GRCh38
rs1016924805 Chr.6: 52763332-52763332 on Build GRCh38
rs548624225 Chr.6: 52763768-52763768 on Build GRCh38
rs961786452 Chr.6: 52763343-52763343 on Build GRCh38
rs990081009 Chr.6: 52763723-52763723 on Build GRCh38
rs1005784699 Chr.6: 52763530-52763530 on Build GRCh38
rs866487609 Chr.6: 52763632-52763632 on Build GRCh38
rs945825543 Chr.6: 52763473-52763473 on Build GRCh38
rs555576321 Chr.6: 52763772-52763772 on Build GRCh38
rs111873291 Chr.6: 52763600-52763600 on Build GRCh38
rs890072163 Chr.6: 52763529-52763529 on Build GRCh38
rs72667055 Chr.6: 52763645-52763645 on Build GRCh38
rs111255880 Chr.6: 52763705-52763705 on Build GRCh38
rs901344838 Chr.6: 52763487-52763487 on Build GRCh38
rs979989697 Chr.6: 52763375-52763375 on Build GRCh38
rs191820579 Chr.6: 52763387-52763387 on Build GRCh38
rs183947502 Chr.6: 52763696-52763696 on Build GRCh38
rs934125606 Chr.6: 52763508-52763508 on Build GRCh38
rs773896271 Chr.6: 52763461-52763461 on Build GRCh38
rs374809225 Chr.6: 52763401-52763401 on Build GRCh38
rs935681860 Chr.6: 52763430-52763430 on Build GRCh38
rs113908451 Chr.6: 52763676-52763676 on Build GRCh38
rs912879358 Chr.6: 52763467-52763467 on Build GRCh38
rs140258043 Chr.6: 52763627-52763627 on Build GRCh38
rs188338216 Chr.6: 52763784-52763784 on Build GRCh38
rs1024591259 Chr.6: 52763357-52763357 on Build GRCh38
rs922987249 Chr.6: 52763760-52763760 on Build GRCh38
rs73740553 Chr.6: 52763350-52763350 on Build GRCh38
rs534815244 Chr.6: 52763785-52763785 on Build GRCh38
rs1017392363 Chr.6: 52763625-52763625 on Build GRCh38
rs151233101 Chr.6: 52763515-52763515 on Build GRCh38
rs1034132688 Chr.6: 52763703-52763703 on Build GRCh38
rs866764576 Chr.6: 52763639-52763639 on Build GRCh38
rs912922192 Chr.6: 52763740-52763740 on Build GRCh38
rs1001293061 Chr.6: 52763701-52763701 on Build GRCh38
rs146110810 Chr.6: 52763696-52763697 on Build GRCh38
rs971167742 Chr.6: 52763374-52763374 on Build GRCh38
rs770535457 Chr.6: 52763370-52763370 on Build GRCh38
rs539717925 Chr.6: 52763553-52763553 on Build GRCh38
rs112774119 Chr.6: 52763326-52763326 on Build GRCh38

View Full Product Details