Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 63726436-63726943 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EYS
Entrez Gene ID
346007
Gene Name
eyes shut homolog (Drosophila)
Gene Aliases
C6orf178, C6orf179, C6orf180, EGFL10, EGFL11, RP25, SPAM, bA166P24.2, bA307F22.3, bA74E24.1, dJ1018A4.2, dJ22I17.2, dJ303F19.1
UniGene
Hs.25067
Gene Chromosome Location
Chr.6: 63719980-65707225 on Build GRCh38
Transcripts
NM_001142800.1 NM_001292009.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4887626 c.8152C>G p.P2718A Chr.6: 63726663-63726663 on Build GRCh38
6316585 c.8249C>A p.A2750E Chr.6: 63726566-63726566 on Build GRCh38
1568286 c.8162G>A p.R2721K Chr.6: 63726653-63726653 on Build GRCh38
7208704 c.8276A>T p.Q2759L Chr.6: 63726539-63726539 on Build GRCh38
451716 c.8181G>C p.W2727C Chr.6: 63726634-63726634 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs954331122 Chr.6: 63726671-63726671 on Build GRCh38
rs1045626049 Chr.6: 63726820-63726820 on Build GRCh38
rs370845590 Chr.6: 63726589-63726589 on Build GRCh38
rs995862502 Chr.6: 63726570-63726570 on Build GRCh38
rs927932155 Chr.6: 63726786-63726786 on Build GRCh38
rs751629543 Chr.6: 63726615-63726619 on Build GRCh38
rs779983752 Chr.6: 63726641-63726641 on Build GRCh38
rs764229134 Chr.6: 63726596-63726597 on Build GRCh38
rs35504565 Chr.6: 63726672-63726672 on Build GRCh38
rs893966416 Chr.6: 63726511-63726511 on Build GRCh38
rs1030041492 Chr.6: 63726918-63726918 on Build GRCh38
rs778420295 Chr.6: 63726608-63726608 on Build GRCh38
rs377362833 Chr.6: 63726566-63726566 on Build GRCh38
rs891588626 Chr.6: 63726463-63726467 on Build GRCh38
rs773389004 Chr.6: 63726535-63726535 on Build GRCh38
rs998405673 Chr.6: 63726875-63726875 on Build GRCh38
rs1014687591 Chr.6: 63726626-63726626 on Build GRCh38
rs1004568595 Chr.6: 63726896-63726898 on Build GRCh38
rs544276613 Chr.6: 63726492-63726492 on Build GRCh38
rs776605808 Chr.6: 63726792-63726792 on Build GRCh38
rs1001192412 Chr.6: 63726564-63726564 on Build GRCh38
rs369121423 Chr.6: 63726571-63726571 on Build GRCh38
rs894004836 Chr.6: 63726794-63726794 on Build GRCh38
rs794727670 Chr.6: 63726614-63726614 on Build GRCh38
rs868203134 Chr.6: 63726637-63726637 on Build GRCh38
rs566770059 Chr.6: 63726650-63726650 on Build GRCh38
rs533165981 Chr.6: 63726596-63726596 on Build GRCh38
rs925907889 Chr.6: 63726690-63726690 on Build GRCh38
rs976913872 Chr.6: 63726630-63726630 on Build GRCh38
rs1039872513 Chr.6: 63726816-63726816 on Build GRCh38
rs184722374 Chr.6: 63726645-63726645 on Build GRCh38
rs752055698 Chr.6: 63726569-63726569 on Build GRCh38
rs374200609 Chr.6: 63726545-63726545 on Build GRCh38
rs369625012 Chr.6: 63726673-63726673 on Build GRCh38
rs1014108793 Chr.6: 63726494-63726497 on Build GRCh38
rs780632755 Chr.6: 63726664-63726664 on Build GRCh38
rs1011193191 Chr.6: 63726541-63726541 on Build GRCh38
rs767955491 Chr.6: 63726574-63726574 on Build GRCh38
rs768560517 Chr.6: 63726716-63726716 on Build GRCh38
rs747874207 Chr.6: 63726713-63726713 on Build GRCh38
rs1052621803 Chr.6: 63726501-63726501 on Build GRCh38
rs946857404 Chr.6: 63726796-63726796 on Build GRCh38
rs916514276 Chr.6: 63726694-63726694 on Build GRCh38
rs756985941 Chr.6: 63726593-63726593 on Build GRCh38
rs549048238 Chr.6: 63726705-63726705 on Build GRCh38
rs1052941266 Chr.6: 63726790-63726790 on Build GRCh38
rs750011709 Chr.6: 63726624-63726624 on Build GRCh38
rs757841530 Chr.6: 63726632-63726632 on Build GRCh38
rs961817248 Chr.6: 63726627-63726629 on Build GRCh38
rs779154270 Chr.6: 63726510-63726510 on Build GRCh38
rs1048978372 Chr.6: 63726862-63726862 on Build GRCh38
rs761591665 Chr.6: 63726856-63726856 on Build GRCh38
rs747133617 Chr.6: 63726711-63726711 on Build GRCh38
rs983603381 Chr.6: 63726601-63726601 on Build GRCh38
rs577677156 Chr.6: 63726472-63726472 on Build GRCh38
rs964398747 Chr.6: 63726567-63726567 on Build GRCh38
rs886061666 Chr.6: 63726511-63726511 on Build GRCh38
rs73441048 Chr.6: 63726788-63726788 on Build GRCh38
rs905774838 Chr.6: 63726835-63726835 on Build GRCh38
rs147138312 Chr.6: 63726512-63726512 on Build GRCh38
rs887410088 Chr.6: 63726882-63726882 on Build GRCh38

View Full Product Details