Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 63806110-63806613 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EYS
Entrez Gene ID
346007
Gene Name
eyes shut homolog (Drosophila)
Gene Aliases
C6orf178, C6orf179, C6orf180, EGFL10, EGFL11, RP25, SPAM, bA166P24.2, bA307F22.3, bA74E24.1, dJ1018A4.2, dJ22I17.2, dJ303F19.1
UniGene
Hs.25067
Gene Chromosome Location
Chr.6: 63719980-65707225 on Build GRCh38
Transcripts
NM_001142800.1 NM_001292009.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4998552 c.7359C>G p.N2453K Chr.6: 63806242-63806242 on Build GRCh38
5737657 c.7409A>G p.H2470R Chr.6: 63806192-63806192 on Build GRCh38
5405701 c.7401G>A p.Q2467Q Chr.6: 63806200-63806200 on Build GRCh38
6850143 c.7379A>G p.N2460S Chr.6: 63806222-63806222 on Build GRCh38
5982467 c.7276T>C p.Y2426H Chr.6: 63806325-63806325 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754984757 Chr.6: 63806209-63806209 on Build GRCh38
rs776743787 Chr.6: 63806362-63806362 on Build GRCh38
rs768229148 Chr.6: 63806344-63806344 on Build GRCh38
rs551500915 Chr.6: 63806232-63806232 on Build GRCh38
rs761885849 Chr.6: 63806408-63806408 on Build GRCh38
rs571226553 Chr.6: 63806481-63806481 on Build GRCh38
rs191508648 Chr.6: 63806135-63806135 on Build GRCh38
rs1025521794 Chr.6: 63806413-63806413 on Build GRCh38
rs563179129 Chr.6: 63806161-63806161 on Build GRCh38
rs144857339 Chr.6: 63806564-63806564 on Build GRCh38
rs749898029 Chr.6: 63806192-63806192 on Build GRCh38
rs986446306 Chr.6: 63806423-63806423 on Build GRCh38
rs981629158 Chr.6: 63806241-63806241 on Build GRCh38
rs544961803 Chr.6: 63806138-63806138 on Build GRCh38
rs893993795 Chr.6: 63806167-63806167 on Build GRCh38
rs1039566510 Chr.6: 63806438-63806438 on Build GRCh38
rs768000972 Chr.6: 63806250-63806250 on Build GRCh38
rs748614028 Chr.6: 63806242-63806242 on Build GRCh38
rs566843175 Chr.6: 63806245-63806245 on Build GRCh38
rs765245784 Chr.6: 63806416-63806416 on Build GRCh38
rs749650182 Chr.6: 63806249-63806249 on Build GRCh38
rs182819407 Chr.6: 63806262-63806262 on Build GRCh38
rs555891277 Chr.6: 63806401-63806401 on Build GRCh38
rs1051437176 Chr.6: 63806531-63806531 on Build GRCh38
rs537662286 Chr.6: 63806332-63806332 on Build GRCh38
rs1039214683 Chr.6: 63806153-63806153 on Build GRCh38
rs886061669 Chr.6: 63806340-63806340 on Build GRCh38
rs750203967 Chr.6: 63806156-63806156 on Build GRCh38
rs933916000 Chr.6: 63806463-63806463 on Build GRCh38
rs953482333 Chr.6: 63806551-63806551 on Build GRCh38
rs886061668 Chr.6: 63806179-63806179 on Build GRCh38
rs775138535 Chr.6: 63806285-63806285 on Build GRCh38
rs567167108 Chr.6: 63806329-63806329 on Build GRCh38
rs745331821 Chr.6: 63806320-63806320 on Build GRCh38
rs761074070 Chr.6: 63806494-63806494 on Build GRCh38
rs758599195 Chr.6: 63806172-63806172 on Build GRCh38
rs572836860 Chr.6: 63806350-63806350 on Build GRCh38
rs962905685 Chr.6: 63806375-63806375 on Build GRCh38
rs972529432 Chr.6: 63806409-63806409 on Build GRCh38
rs527812465 Chr.6: 63806248-63806248 on Build GRCh38
rs899640885 Chr.6: 63806448-63806448 on Build GRCh38
rs187891419 Chr.6: 63806576-63806576 on Build GRCh38
rs1034996144 Chr.6: 63806286-63806286 on Build GRCh38
rs780311041 Chr.6: 63806194-63806194 on Build GRCh38
rs145184183 Chr.6: 63806207-63806207 on Build GRCh38
rs775877185 Chr.6: 63806317-63806317 on Build GRCh38
rs910848514 Chr.6: 63806546-63806546 on Build GRCh38

View Full Product Details