Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 70415170-70415709 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM135A
Entrez Gene ID
57579
Gene Name
family with sequence similarity 135 member A
Gene Aliases
KIAA1411
UniGene
Hs.211700
Gene Chromosome Location
Chr.6: 70413404-70561174 on Build GRCh38
Transcripts
NM_001105531.2 NM_020819.4 XM_005248743.1 XM_005248744.4 XM_005248745.2 XM_005248748.2 XM_005248749.1 XM_005248751.2 XM_011535994.2 XM_011535995.2 XM_011535996.2 XM_011535997.2 XM_017011124.1 XM_017011125.1 XM_017011126.1 XM_017011127.1 XM_017011128.1 XM_017011129.1 XM_017011130.1 XM_017011131.1 XM_017011132.1 XM_017011133.1 XM_017011134.1 XM_017011135.1

SNP Detail

SNP ID Chromosome Location
rs891870907 Chr.6: 70415484-70415484 on Build GRCh38
rs569164689 Chr.6: 70415537-70415537 on Build GRCh38
rs543225812 Chr.6: 70415414-70415414 on Build GRCh38
rs930948910 Chr.6: 70415212-70415212 on Build GRCh38
rs781033129 Chr.6: 70415481-70415481 on Build GRCh38
rs969283677 Chr.6: 70415367-70415367 on Build GRCh38
rs1050035687 Chr.6: 70415202-70415202 on Build GRCh38
rs751549399 Chr.6: 70415416-70415416 on Build GRCh38
rs905151092 Chr.6: 70415661-70415661 on Build GRCh38
rs545287425 Chr.6: 70415629-70415629 on Build GRCh38
rs908328131 Chr.6: 70415573-70415573 on Build GRCh38
rs1008594216 Chr.6: 70415250-70415250 on Build GRCh38
rs892297324 Chr.6: 70415292-70415292 on Build GRCh38
rs944350652 Chr.6: 70415586-70415586 on Build GRCh38
rs1009388741 Chr.6: 70415485-70415485 on Build GRCh38
rs9294867 Chr.6: 70415513-70415513 on Build GRCh38
rs570882700 Chr.6: 70415213-70415213 on Build GRCh38
rs1049780825 Chr.6: 70415337-70415337 on Build GRCh38
rs1002230730 Chr.6: 70415374-70415374 on Build GRCh38
rs1000570558 Chr.6: 70415418-70415418 on Build GRCh38
rs765203652 Chr.6: 70415345-70415345 on Build GRCh38
rs1024749817 Chr.6: 70415500-70415500 on Build GRCh38
rs976948505 Chr.6: 70415625-70415625 on Build GRCh38
rs754157599 Chr.6: 70415489-70415493 on Build GRCh38
rs546692028 Chr.6: 70415329-70415329 on Build GRCh38
rs1050017226 Chr.6: 70415647-70415647 on Build GRCh38
rs890226870 Chr.6: 70415222-70415222 on Build GRCh38
rs148327999 Chr.6: 70415549-70415549 on Build GRCh38
rs939744413 Chr.6: 70415582-70415582 on Build GRCh38
rs532064124 Chr.6: 70415628-70415628 on Build GRCh38
rs536976396 Chr.6: 70415515-70415515 on Build GRCh38
rs755120991 Chr.6: 70415341-70415341 on Build GRCh38
rs1022649572 Chr.6: 70415330-70415330 on Build GRCh38
rs1002114862 Chr.6: 70415662-70415662 on Build GRCh38
rs908257645 Chr.6: 70415240-70415240 on Build GRCh38
rs533853492 Chr.6: 70415673-70415673 on Build GRCh38
rs566832161 Chr.6: 70415381-70415381 on Build GRCh38
rs960814104 Chr.6: 70415425-70415426 on Build GRCh38
rs1030717935 Chr.6: 70415263-70415263 on Build GRCh38
rs983902931 Chr.6: 70415236-70415236 on Build GRCh38
rs983278403 Chr.6: 70415466-70415466 on Build GRCh38
rs191562186 Chr.6: 70415301-70415301 on Build GRCh38
rs931663043 Chr.6: 70415198-70415198 on Build GRCh38
rs752259819 Chr.6: 70415313-70415338 on Build GRCh38
rs183896787 Chr.6: 70415417-70415417 on Build GRCh38
rs550562385 Chr.6: 70415303-70415303 on Build GRCh38
rs1043563195 Chr.6: 70415655-70415655 on Build GRCh38
rs940175039 Chr.6: 70415271-70415271 on Build GRCh38
rs974238610 Chr.6: 70415499-70415499 on Build GRCh38
rs1033988273 Chr.6: 70415526-70415526 on Build GRCh38

View Full Product Details