Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 70426158-70426605 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM135A
Entrez Gene ID
57579
Gene Name
family with sequence similarity 135 member A
Gene Aliases
KIAA1411
UniGene
Hs.211700
Gene Chromosome Location
Chr.6: 70413404-70561174 on Build GRCh38
Transcripts
NM_001105531.2 NM_001162529.1 NM_020819.4 XM_005248743.1 XM_005248744.4 XM_005248745.2 XM_005248748.2 XM_005248749.1 XM_005248751.2 XM_011535993.2 XM_011535994.2 XM_011535995.2 XM_011535996.2 XM_011535997.2 XM_017011124.1 XM_017011125.1 XM_017011126.1 XM_017011127.1 XM_017011128.1 XM_017011129.1 XM_017011130.1 XM_017011131.1 XM_017011132.1 XM_017011133.1 XM_017011134.1 XM_017011135.1

SNP Detail

SNP ID Chromosome Location
rs368865981 Chr.6: 70426368-70426368 on Build GRCh38
rs892759860 Chr.6: 70426210-70426210 on Build GRCh38
rs1040063157 Chr.6: 70426503-70426503 on Build GRCh38
rs377152323 Chr.6: 70426445-70426445 on Build GRCh38
rs1024161871 Chr.6: 70426316-70426316 on Build GRCh38
rs947162899 Chr.6: 70426223-70426223 on Build GRCh38
rs999561591 Chr.6: 70426567-70426567 on Build GRCh38
rs370623725 Chr.6: 70426449-70426449 on Build GRCh38
rs974836739 Chr.6: 70426317-70426317 on Build GRCh38
rs996388970 Chr.6: 70426186-70426186 on Build GRCh38
rs1044104059 Chr.6: 70426242-70426242 on Build GRCh38
rs938550721 Chr.6: 70426546-70426546 on Build GRCh38
rs764399692 Chr.6: 70426323-70426324 on Build GRCh38
rs987636695 Chr.6: 70426390-70426390 on Build GRCh38
rs922112985 Chr.6: 70426513-70426513 on Build GRCh38
rs1016268268 Chr.6: 70426312-70426312 on Build GRCh38
rs531917784 Chr.6: 70426435-70426435 on Build GRCh38
rs535014173 Chr.6: 70426196-70426196 on Build GRCh38
rs775835586 Chr.6: 70426200-70426203 on Build GRCh38
rs904084877 Chr.6: 70426566-70426566 on Build GRCh38
rs944480645 Chr.6: 70426481-70426481 on Build GRCh38
rs144960952 Chr.6: 70426257-70426262 on Build GRCh38
rs1052877239 Chr.6: 70426574-70426574 on Build GRCh38
rs182493506 Chr.6: 70426285-70426285 on Build GRCh38
rs992214455 Chr.6: 70426305-70426305 on Build GRCh38
rs73474964 Chr.6: 70426250-70426250 on Build GRCh38
rs751554201 Chr.6: 70426384-70426385 on Build GRCh38
rs999932507 Chr.6: 70426268-70426268 on Build GRCh38
rs1029067529 Chr.6: 70426350-70426353 on Build GRCh38
rs914264217 Chr.6: 70426442-70426442 on Build GRCh38
rs1031108636 Chr.6: 70426277-70426277 on Build GRCh38
rs574659843 Chr.6: 70426269-70426269 on Build GRCh38
rs545870851 Chr.6: 70426517-70426517 on Build GRCh38
rs564837635 Chr.6: 70426405-70426405 on Build GRCh38
rs142807421 Chr.6: 70426543-70426543 on Build GRCh38
rs187711319 Chr.6: 70426416-70426416 on Build GRCh38
rs983512002 Chr.6: 70426447-70426447 on Build GRCh38
rs990102034 Chr.6: 70426441-70426441 on Build GRCh38
rs932145259 Chr.6: 70426536-70426536 on Build GRCh38
rs192540982 Chr.6: 70426523-70426523 on Build GRCh38
rs557088399 Chr.6: 70426381-70426381 on Build GRCh38
rs935605411 Chr.6: 70426438-70426438 on Build GRCh38

View Full Product Details