Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 70486050-70486589 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM135A
Entrez Gene ID
57579
Gene Name
family with sequence similarity 135 member A
Gene Aliases
KIAA1411
UniGene
Hs.211700
Gene Chromosome Location
Chr.6: 70413404-70561174 on Build GRCh38
Transcripts
NM_001105531.2 NM_001162529.1 NM_020819.4 XM_005248743.1 XM_005248744.4 XM_005248745.2 XM_005248748.2 XM_005248749.1 XM_005248751.2 XM_011535993.2 XM_011535994.2 XM_011535995.2 XM_011535996.2 XM_011535997.2 XM_017011124.1 XM_017011125.1 XM_017011126.1 XM_017011127.1 XM_017011128.1 XM_017011129.1 XM_017011130.1 XM_017011131.1 XM_017011132.1 XM_017011133.1 XM_017011134.1 XM_017011135.1 XM_017011136.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6658505 c.724C>A p.R242S Chr.6: 70486196-70486196 on Build GRCh38
4987642 c.724C>T p.R242C Chr.6: 70486196-70486196 on Build GRCh38
1081260 c.749G>A p.R250Q Chr.6: 70486221-70486221 on Build GRCh38
5633621 c.736A>G p.R246G Chr.6: 70486208-70486208 on Build GRCh38
1081259 c.730C>A p.L244I Chr.6: 70486202-70486202 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs910973760 Chr.6: 70486097-70486097 on Build GRCh38
rs748090841 Chr.6: 70486521-70486521 on Build GRCh38
rs183667677 Chr.6: 70486314-70486314 on Build GRCh38
rs779495844 Chr.6: 70486254-70486254 on Build GRCh38
rs769475515 Chr.6: 70486212-70486212 on Build GRCh38
rs747858200 Chr.6: 70486270-70486270 on Build GRCh38
rs1053770103 Chr.6: 70486425-70486425 on Build GRCh38
rs565361768 Chr.6: 70486332-70486332 on Build GRCh38
rs1040704804 Chr.6: 70486127-70486127 on Build GRCh38
rs369691884 Chr.6: 70486221-70486221 on Build GRCh38
rs377070236 Chr.6: 70486260-70486260 on Build GRCh38
rs767177273 Chr.6: 70486225-70486225 on Build GRCh38
rs770301005 Chr.6: 70486290-70486290 on Build GRCh38
rs772935543 Chr.6: 70486276-70486276 on Build GRCh38
rs757225680 Chr.6: 70486253-70486253 on Build GRCh38
rs142154928 Chr.6: 70486215-70486215 on Build GRCh38
rs772832673 Chr.6: 70486214-70486214 on Build GRCh38
rs754665325 Chr.6: 70486131-70486131 on Build GRCh38
rs912882928 Chr.6: 70486422-70486425 on Build GRCh38
rs757755738 Chr.6: 70486186-70486186 on Build GRCh38
rs772275248 Chr.6: 70486204-70486204 on Build GRCh38
rs200570801 Chr.6: 70486223-70486223 on Build GRCh38
rs897800489 Chr.6: 70486549-70486549 on Build GRCh38
rs746256854 Chr.6: 70486196-70486196 on Build GRCh38
rs535171868 Chr.6: 70486151-70486151 on Build GRCh38
rs754365053 Chr.6: 70486119-70486119 on Build GRCh38
rs747768113 Chr.6: 70486209-70486209 on Build GRCh38
rs943667846 Chr.6: 70486112-70486116 on Build GRCh38
rs369704366 Chr.6: 70486494-70486494 on Build GRCh38
rs530212458 Chr.6: 70486497-70486497 on Build GRCh38
rs753917224 Chr.6: 70486249-70486249 on Build GRCh38
rs1010135458 Chr.6: 70486211-70486211 on Build GRCh38
rs776373240 Chr.6: 70486205-70486205 on Build GRCh38
rs377003711 Chr.6: 70486142-70486142 on Build GRCh38
rs757478451 Chr.6: 70486359-70486359 on Build GRCh38
rs756217306 Chr.6: 70486157-70486157 on Build GRCh38
rs541248121 Chr.6: 70486470-70486470 on Build GRCh38
rs570025880 Chr.6: 70486552-70486552 on Build GRCh38
rs764218702 Chr.6: 70486248-70486248 on Build GRCh38
rs571372058 Chr.6: 70486366-70486366 on Build GRCh38
rs1009593427 Chr.6: 70486483-70486483 on Build GRCh38
rs867841644 Chr.6: 70486389-70486389 on Build GRCh38
rs751367994 Chr.6: 70486125-70486125 on Build GRCh38
rs953506511 Chr.6: 70486341-70486341 on Build GRCh38
rs972256854 Chr.6: 70486304-70486304 on Build GRCh38
rs769563805 Chr.6: 70486274-70486274 on Build GRCh38
rs777801284 Chr.6: 70486170-70486170 on Build GRCh38
rs766346790 Chr.6: 70486220-70486220 on Build GRCh38
rs976931333 Chr.6: 70486415-70486415 on Build GRCh38
rs180687505 Chr.6: 70486307-70486307 on Build GRCh38
rs1042956222 Chr.6: 70486444-70486444 on Build GRCh38
rs936682356 Chr.6: 70486420-70486420 on Build GRCh38
rs746015316 Chr.6: 70486305-70486305 on Build GRCh38
rs1049444181 Chr.6: 70486077-70486078 on Build GRCh38
rs780255288 Chr.6: 70486267-70486267 on Build GRCh38
rs1014909945 Chr.6: 70486288-70486288 on Build GRCh38
rs142935920 Chr.6: 70486189-70486189 on Build GRCh38
rs868316506 Chr.6: 70486430-70486430 on Build GRCh38
rs1002627364 Chr.6: 70486514-70486514 on Build GRCh38
rs144036955 Chr.6: 70486238-70486238 on Build GRCh38
rs759516112 Chr.6: 70486275-70486275 on Build GRCh38
rs550236677 Chr.6: 70486542-70486542 on Build GRCh38
rs1015437186 Chr.6: 70486547-70486547 on Build GRCh38
rs576341362 Chr.6: 70486313-70486313 on Build GRCh38
rs749844146 Chr.6: 70486178-70486178 on Build GRCh38
rs937275246 Chr.6: 70486475-70486475 on Build GRCh38
rs556915156 Chr.6: 70486509-70486509 on Build GRCh38
rs745966875 Chr.6: 70486564-70486564 on Build GRCh38
rs139238765 Chr.6: 70486230-70486230 on Build GRCh38
rs781495600 Chr.6: 70486148-70486148 on Build GRCh38
rs907047580 Chr.6: 70486505-70486505 on Build GRCh38
rs1014330477 Chr.6: 70486540-70486540 on Build GRCh38
rs1051120234 Chr.6: 70486316-70486316 on Build GRCh38
rs76875901 Chr.6: 70486484-70486484 on Build GRCh38
rs892522139 Chr.6: 70486117-70486117 on Build GRCh38
rs776493209 Chr.6: 70486122-70486123 on Build GRCh38
rs375311087 Chr.6: 70486231-70486231 on Build GRCh38
rs912093061 Chr.6: 70486378-70486378 on Build GRCh38
rs904449517 Chr.6: 70486451-70486451 on Build GRCh38
rs752868530 Chr.6: 70486156-70486156 on Build GRCh38
rs527854119 Chr.6: 70486362-70486362 on Build GRCh38
rs748823560 Chr.6: 70486280-70486280 on Build GRCh38
rs756309054 Chr.6: 70486247-70486247 on Build GRCh38

View Full Product Details