Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 73695650-73696150 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD109
Entrez Gene ID
135228
Gene Name
CD109 molecule
Gene Aliases
CPAMD7, p180, r150
UniGene
Hs.399891
Gene Chromosome Location
Chr.6: 73679192-73828317 on Build GRCh38
Transcripts
NM_001159587.2 NM_001159588.2 NM_133493.4 XM_005248659.3 XM_011535473.1 NR_110889.1

Gene Symbol
LOC101928489
Entrez Gene ID
101928489
Gene Name
uncharacterized LOC101928489
Gene Aliases
-
UniGene
Hs.660059
Gene Chromosome Location
Chr.6: 73693903-73696131 on Build GRCh38
Transcripts
NM_001159587.2 NM_001159588.2 NM_133493.4 XM_005248659.3 XM_011535473.1 NR_110889.1

SNP Detail

SNP ID Chromosome Location
rs569988566 Chr.6: 73695746-73695747 on Build GRCh38
rs554431943 Chr.6: 73695881-73695881 on Build GRCh38
rs933967238 Chr.6: 73695816-73695816 on Build GRCh38
rs762554360 Chr.6: 73695697-73695698 on Build GRCh38
rs762441956 Chr.6: 73695712-73695713 on Build GRCh38
rs181908581 Chr.6: 73695950-73695950 on Build GRCh38
rs988902098 Chr.6: 73695741-73695741 on Build GRCh38
rs868658391 Chr.6: 73695868-73695868 on Build GRCh38
rs915529570 Chr.6: 73695975-73695975 on Build GRCh38
rs141313382 Chr.6: 73695792-73695803 on Build GRCh38
rs943533553 Chr.6: 73695936-73695936 on Build GRCh38
rs969967524 Chr.6: 73695712-73695712 on Build GRCh38
rs957239329 Chr.6: 73695731-73695731 on Build GRCh38
rs979877272 Chr.6: 73695713-73695713 on Build GRCh38
rs955656745 Chr.6: 73696124-73696124 on Build GRCh38
rs894023547 Chr.6: 73695683-73695683 on Build GRCh38
rs925776529 Chr.6: 73695714-73695714 on Build GRCh38
rs1013207559 Chr.6: 73696092-73696092 on Build GRCh38
rs911446202 Chr.6: 73695855-73695855 on Build GRCh38
rs369209299 Chr.6: 73695698-73695699 on Build GRCh38
rs535698880 Chr.6: 73695830-73695830 on Build GRCh38
rs898663098 Chr.6: 73695902-73695902 on Build GRCh38
rs773891421 Chr.6: 73695709-73695710 on Build GRCh38
rs971740232 Chr.6: 73695843-73695843 on Build GRCh38
rs930170895 Chr.6: 73695907-73695907 on Build GRCh38
rs1042435105 Chr.6: 73696003-73696003 on Build GRCh38
rs1004019391 Chr.6: 73695709-73695709 on Build GRCh38
rs895192615 Chr.6: 73696053-73696053 on Build GRCh38
rs1024406384 Chr.6: 73696027-73696028 on Build GRCh38
rs1009610625 Chr.6: 73695697-73695697 on Build GRCh38
rs1032924088 Chr.6: 73696079-73696079 on Build GRCh38
rs1053955171 Chr.6: 73695974-73695974 on Build GRCh38
rs142824304 Chr.6: 73695776-73695776 on Build GRCh38
rs1038560054 Chr.6: 73695681-73695690 on Build GRCh38
rs6149641 Chr.6: 73695793-73695804 on Build GRCh38
rs906552551 Chr.6: 73696011-73696011 on Build GRCh38
rs762257337 Chr.6: 73695710-73695713 on Build GRCh38
rs1001390639 Chr.6: 73696052-73696052 on Build GRCh38
rs9442945 Chr.6: 73696029-73696029 on Build GRCh38
rs886191310 Chr.6: 73695999-73695999 on Build GRCh38
rs958800954 Chr.6: 73695811-73695811 on Build GRCh38
rs866335313 Chr.6: 73696129-73696129 on Build GRCh38
rs141348716 Chr.6: 73695710-73695711 on Build GRCh38
rs954353572 Chr.6: 73695876-73695876 on Build GRCh38
rs565833617 Chr.6: 73695992-73695992 on Build GRCh38
rs572691274 Chr.6: 73695959-73695959 on Build GRCh38
rs878897918 Chr.6: 73696123-73696123 on Build GRCh38
rs1054451865 Chr.6: 73695674-73695674 on Build GRCh38
rs151113002 Chr.6: 73696070-73696070 on Build GRCh38
rs764739426 Chr.6: 73695694-73695705 on Build GRCh38
rs1014346828 Chr.6: 73695707-73695707 on Build GRCh38
rs1012718709 Chr.6: 73695813-73695813 on Build GRCh38
rs963295882 Chr.6: 73696117-73696117 on Build GRCh38
rs1055626627 Chr.6: 73696032-73696032 on Build GRCh38
rs1020341850 Chr.6: 73696123-73696124 on Build GRCh38
rs535640323 Chr.6: 73695833-73695833 on Build GRCh38
rs906644165 Chr.6: 73695705-73695705 on Build GRCh38
rs754263417 Chr.6: 73696063-73696063 on Build GRCh38
rs910265710 Chr.6: 73695885-73695885 on Build GRCh38
rs1016045379 Chr.6: 73696107-73696107 on Build GRCh38
rs189541744 Chr.6: 73695703-73695703 on Build GRCh38
rs568278646 Chr.6: 73695822-73695822 on Build GRCh38

View Full Product Details