Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 73696044-73696553 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD109
Entrez Gene ID
135228
Gene Name
CD109 molecule
Gene Aliases
CPAMD7, p180, r150
UniGene
Hs.399891
Gene Chromosome Location
Chr.6: 73679192-73828317 on Build GRCh38
Transcripts
NM_001159587.2 NM_001159588.2 NM_133493.4 XM_005248659.3 XM_011535473.1 NR_110889.1

Gene Symbol
LOC101928489
Entrez Gene ID
101928489
Gene Name
uncharacterized LOC101928489
Gene Aliases
-
UniGene
Hs.660059
Gene Chromosome Location
Chr.6: 73693903-73696131 on Build GRCh38
Transcripts
NM_001159587.2 NM_001159588.2 NM_133493.4 XM_005248659.3 XM_011535473.1 NR_110889.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6623378 c.28G>A p.A10T Chr.6: 73696243-73696243 on Build GRCh38
4621710 c.42C>T p.C14C Chr.6: 73696257-73696257 on Build GRCh38
4625843 c.25G>A p.A9T Chr.6: 73696240-73696240 on Build GRCh38
6623384 c.37C>T p.L13F Chr.6: 73696252-73696252 on Build GRCh38
6581681 c.3G>T p.M1I Chr.6: 73696218-73696218 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs974073487 Chr.6: 73696308-73696309 on Build GRCh38
rs1022025900 Chr.6: 73696378-73696378 on Build GRCh38
rs778804944 Chr.6: 73696235-73696235 on Build GRCh38
rs951733652 Chr.6: 73696310-73696311 on Build GRCh38
rs368748901 Chr.6: 73696165-73696165 on Build GRCh38
rs941441710 Chr.6: 73696285-73696285 on Build GRCh38
rs749451216 Chr.6: 73696303-73696303 on Build GRCh38
rs755443358 Chr.6: 73696266-73696266 on Build GRCh38
rs1047696471 Chr.6: 73696233-73696233 on Build GRCh38
rs955257876 Chr.6: 73696295-73696295 on Build GRCh38
rs755280995 Chr.6: 73696343-73696343 on Build GRCh38
rs927046645 Chr.6: 73696340-73696340 on Build GRCh38
rs769277474 Chr.6: 73696191-73696191 on Build GRCh38
rs1054060835 Chr.6: 73696349-73696349 on Build GRCh38
rs747990886 Chr.6: 73696267-73696267 on Build GRCh38
rs545840915 Chr.6: 73696270-73696270 on Build GRCh38
rs950904728 Chr.6: 73696345-73696357 on Build GRCh38
rs993256040 Chr.6: 73696312-73696312 on Build GRCh38
rs978216684 Chr.6: 73696394-73696394 on Build GRCh38
rs749960538 Chr.6: 73696252-73696252 on Build GRCh38
rs1009013342 Chr.6: 73696358-73696358 on Build GRCh38
rs939612703 Chr.6: 73696409-73696409 on Build GRCh38
rs955656745 Chr.6: 73696124-73696124 on Build GRCh38
rs549403484 Chr.6: 73696459-73696459 on Build GRCh38
rs73754655 Chr.6: 73696147-73696147 on Build GRCh38
rs989246443 Chr.6: 73696374-73696374 on Build GRCh38
rs989675092 Chr.6: 73696234-73696234 on Build GRCh38
rs949619377 Chr.6: 73696331-73696331 on Build GRCh38
rs1010042694 Chr.6: 73696278-73696278 on Build GRCh38
rs1013207559 Chr.6: 73696092-73696092 on Build GRCh38
rs531317245 Chr.6: 73696420-73696420 on Build GRCh38
rs761773703 Chr.6: 73696245-73696245 on Build GRCh38
rs200017709 Chr.6: 73696226-73696226 on Build GRCh38
rs964839751 Chr.6: 73696367-73696367 on Build GRCh38
rs769799204 Chr.6: 73696299-73696300 on Build GRCh38
rs1027389380 Chr.6: 73696513-73696513 on Build GRCh38
rs369446412 Chr.6: 73696251-73696251 on Build GRCh38
rs542036029 Chr.6: 73696232-73696232 on Build GRCh38
rs186230610 Chr.6: 73696526-73696526 on Build GRCh38
rs545781508 Chr.6: 73696181-73696181 on Build GRCh38
rs978352755 Chr.6: 73696246-73696246 on Build GRCh38
rs74745681 Chr.6: 73696264-73696264 on Build GRCh38
rs777648758 Chr.6: 73696287-73696287 on Build GRCh38
rs560433332 Chr.6: 73696241-73696241 on Build GRCh38
rs769733493 Chr.6: 73696311-73696318 on Build GRCh38
rs1032924088 Chr.6: 73696079-73696079 on Build GRCh38
rs757918232 Chr.6: 73696258-73696258 on Build GRCh38
rs770438976 Chr.6: 73696284-73696284 on Build GRCh38
rs770031240 Chr.6: 73696200-73696200 on Build GRCh38
rs1026158563 Chr.6: 73696145-73696145 on Build GRCh38
rs371757131 Chr.6: 73696195-73696195 on Build GRCh38
rs920184106 Chr.6: 73696223-73696223 on Build GRCh38
rs748625114 Chr.6: 73696272-73696272 on Build GRCh38
rs966877495 Chr.6: 73696387-73696387 on Build GRCh38
rs991259720 Chr.6: 73696410-73696410 on Build GRCh38
rs751959358 Chr.6: 73696263-73696263 on Build GRCh38
rs964006296 Chr.6: 73696307-73696308 on Build GRCh38
rs866335313 Chr.6: 73696129-73696129 on Build GRCh38
rs1017257231 Chr.6: 73696366-73696366 on Build GRCh38
rs776706050 Chr.6: 73696233-73696233 on Build GRCh38
rs1026189506 Chr.6: 73696315-73696315 on Build GRCh38
rs995478532 Chr.6: 73696465-73696465 on Build GRCh38
rs1045421395 Chr.6: 73696337-73696337 on Build GRCh38
rs576353076 Chr.6: 73696178-73696178 on Build GRCh38
rs773370652 Chr.6: 73696215-73696215 on Build GRCh38
rs1010550386 Chr.6: 73696386-73696386 on Build GRCh38
rs878897918 Chr.6: 73696123-73696123 on Build GRCh38
rs557742537 Chr.6: 73696300-73696318 on Build GRCh38
rs151113002 Chr.6: 73696070-73696070 on Build GRCh38
rs574818465 Chr.6: 73696230-73696230 on Build GRCh38
rs963295882 Chr.6: 73696117-73696117 on Build GRCh38
rs564040217 Chr.6: 73696327-73696327 on Build GRCh38
rs1020341850 Chr.6: 73696123-73696124 on Build GRCh38
rs779819372 Chr.6: 73696262-73696262 on Build GRCh38
rs1037523045 Chr.6: 73696300-73696300 on Build GRCh38
rs1055132908 Chr.6: 73696257-73696257 on Build GRCh38
rs986932197 Chr.6: 73696137-73696137 on Build GRCh38
rs945505036 Chr.6: 73696239-73696239 on Build GRCh38
rs370080673 Chr.6: 73696457-73696457 on Build GRCh38
rs562038956 Chr.6: 73696501-73696501 on Build GRCh38
rs758088493 Chr.6: 73696167-73696167 on Build GRCh38
rs763030248 Chr.6: 73696222-73696222 on Build GRCh38
rs928245507 Chr.6: 73696403-73696403 on Build GRCh38
rs1016045379 Chr.6: 73696107-73696107 on Build GRCh38
rs899014869 Chr.6: 73696310-73696310 on Build GRCh38
rs375627612 Chr.6: 73696206-73696206 on Build GRCh38
rs771046714 Chr.6: 73696319-73696319 on Build GRCh38
rs941343767 Chr.6: 73696424-73696424 on Build GRCh38
rs1038423631 Chr.6: 73696453-73696453 on Build GRCh38

View Full Product Details