Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 73697519-73698060 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD109
Entrez Gene ID
135228
Gene Name
CD109 molecule
Gene Aliases
CPAMD7, p180, r150
UniGene
Hs.399891
Gene Chromosome Location
Chr.6: 73679192-73828317 on Build GRCh38
Transcripts
NM_001159587.2 NM_001159588.2 NM_133493.4 XM_005248659.3 XM_011535473.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6107757 c.227C>G p.A76G Chr.6: 73697552-73697552 on Build GRCh38
6623388 c.228_230delAGA p.E77delE Chr.6: 73697553-73697555 on Build GRCh38
484346 c.228A>G p.A76A Chr.6: 73697553-73697553 on Build GRCh38
5727123 c.219C>T p.V73V Chr.6: 73697544-73697544 on Build GRCh38
1329543 c.247+1_247+2insT p.? Chr.6: 73697573-73697574 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1020020064 Chr.6: 73698008-73698008 on Build GRCh38
rs1043073920 Chr.6: 73698023-73698023 on Build GRCh38
rs923821539 Chr.6: 73697636-73697636 on Build GRCh38
rs763405190 Chr.6: 73697553-73697555 on Build GRCh38
rs769670663 Chr.6: 73697589-73697589 on Build GRCh38
rs533495267 Chr.6: 73697735-73697735 on Build GRCh38
rs1011463728 Chr.6: 73697580-73697580 on Build GRCh38
rs72953369 Chr.6: 73697895-73697895 on Build GRCh38
rs935049660 Chr.6: 73697674-73697674 on Build GRCh38
rs77670907 Chr.6: 73697685-73697685 on Build GRCh38
rs967304672 Chr.6: 73697648-73697648 on Build GRCh38
rs1008263037 Chr.6: 73697998-73697998 on Build GRCh38
rs1011140726 Chr.6: 73697964-73697964 on Build GRCh38
rs921152621 Chr.6: 73697816-73697816 on Build GRCh38
rs745949220 Chr.6: 73697548-73697548 on Build GRCh38
rs770785421 Chr.6: 73697596-73697596 on Build GRCh38
rs148053046 Chr.6: 73697873-73697873 on Build GRCh38
rs998614859 Chr.6: 73697668-73697668 on Build GRCh38
rs144858532 Chr.6: 73697574-73697574 on Build GRCh38
rs964186233 Chr.6: 73698011-73698011 on Build GRCh38
rs896263325 Chr.6: 73697701-73697701 on Build GRCh38
rs764476450 Chr.6: 73697581-73697582 on Build GRCh38
rs1019411028 Chr.6: 73697677-73697677 on Build GRCh38
rs939882578 Chr.6: 73697886-73697886 on Build GRCh38
rs528481109 Chr.6: 73697969-73697969 on Build GRCh38
rs549461988 Chr.6: 73697893-73697893 on Build GRCh38
rs772911622 Chr.6: 73697592-73697592 on Build GRCh38
rs540651886 Chr.6: 73697634-73697634 on Build GRCh38
rs371717655 Chr.6: 73697562-73697562 on Build GRCh38
rs758611968 Chr.6: 73697558-73697558 on Build GRCh38
rs545579548 Chr.6: 73697841-73697841 on Build GRCh38
rs921747343 Chr.6: 73697590-73697590 on Build GRCh38
rs575330965 Chr.6: 73697692-73697692 on Build GRCh38
rs375053133 Chr.6: 73697599-73697599 on Build GRCh38
rs117634953 Chr.6: 73697880-73697880 on Build GRCh38
rs947775548 Chr.6: 73697712-73697712 on Build GRCh38
rs1021110472 Chr.6: 73697643-73697643 on Build GRCh38
rs368888917 Chr.6: 73697791-73697791 on Build GRCh38
rs893838274 Chr.6: 73697947-73697947 on Build GRCh38
rs1055062286 Chr.6: 73697936-73697936 on Build GRCh38
rs749198631 Chr.6: 73697593-73697593 on Build GRCh38
rs771639841 Chr.6: 73697842-73697842 on Build GRCh38
rs773994606 Chr.6: 73697598-73697598 on Build GRCh38
rs946511797 Chr.6: 73697624-73697624 on Build GRCh38
rs937991080 Chr.6: 73697916-73697916 on Build GRCh38
rs928054917 Chr.6: 73697906-73697906 on Build GRCh38

View Full Product Details