Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 75237586-75238080 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX7A2
Entrez Gene ID
1347
Gene Name
cytochrome c oxidase subunit 7A2
Gene Aliases
COX7AL, COX7AL1, COXVIIAL, COXVIIa-L, VIIAL
UniGene
Hs.70312
Gene Chromosome Location
Chr.6: 75237675-75243928 on Build GRCh38
Transcripts
NM_001865.3 NR_029466.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6880630 c.332C>T p.P111L Chr.6: 75237946-75237946 on Build GRCh38
1697546 c.236C>T p.P79L Chr.6: 75237946-75237946 on Build GRCh38
484358 c.225G>A p.V75V Chr.6: 75237957-75237957 on Build GRCh38
6007259 c.331C>G p.P111A Chr.6: 75237947-75237947 on Build GRCh38
4857420 c.321G>A p.V107V Chr.6: 75237957-75237957 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs562289269 Chr.6: 75237972-75237972 on Build GRCh38
rs951227869 Chr.6: 75237790-75237791 on Build GRCh38
rs756466895 Chr.6: 75237951-75237951 on Build GRCh38
rs1026807570 Chr.6: 75237780-75237780 on Build GRCh38
rs749796017 Chr.6: 75237909-75237909 on Build GRCh38
rs567368203 Chr.6: 75237765-75237765 on Build GRCh38
rs1046101120 Chr.6: 75237943-75237943 on Build GRCh38
rs765571649 Chr.6: 75237907-75237907 on Build GRCh38
rs139929377 Chr.6: 75237637-75237637 on Build GRCh38
rs768097567 Chr.6: 75237877-75237877 on Build GRCh38
rs760783838 Chr.6: 75237786-75237786 on Build GRCh38
rs766705738 Chr.6: 75238016-75238016 on Build GRCh38
rs796306978 Chr.6: 75237955-75237955 on Build GRCh38
rs1041483976 Chr.6: 75237639-75237639 on Build GRCh38
rs113430068 Chr.6: 75237771-75237771 on Build GRCh38
rs770819113 Chr.6: 75237843-75237843 on Build GRCh38
rs368512375 Chr.6: 75237956-75237956 on Build GRCh38
rs1015021825 Chr.6: 75237808-75237808 on Build GRCh38
rs141924440 Chr.6: 75237969-75237969 on Build GRCh38
rs766867977 Chr.6: 75237919-75237919 on Build GRCh38
rs371480081 Chr.6: 75237967-75237967 on Build GRCh38
rs892130694 Chr.6: 75237932-75237932 on Build GRCh38
rs572469940 Chr.6: 75237801-75237801 on Build GRCh38
rs536503337 Chr.6: 75237865-75237865 on Build GRCh38
rs576647071 Chr.6: 75237879-75237879 on Build GRCh38
rs890752815 Chr.6: 75237694-75237694 on Build GRCh38
rs981864669 Chr.6: 75237893-75237893 on Build GRCh38
rs75996601 Chr.6: 75237910-75237910 on Build GRCh38
rs554336267 Chr.6: 75237776-75237776 on Build GRCh38
rs767534648 Chr.6: 75237933-75237933 on Build GRCh38
rs1060003 Chr.6: 75237869-75237869 on Build GRCh38
rs763329014 Chr.6: 75238012-75238012 on Build GRCh38
rs772881004 Chr.6: 75238010-75238010 on Build GRCh38
rs923129559 Chr.6: 75237610-75237610 on Build GRCh38
rs752029766 Chr.6: 75237922-75237922 on Build GRCh38
rs776175060 Chr.6: 75237990-75237990 on Build GRCh38
rs761516468 Chr.6: 75237895-75237895 on Build GRCh38
rs778922040 Chr.6: 75237851-75237851 on Build GRCh38
rs755065706 Chr.6: 75237927-75237927 on Build GRCh38
rs990393332 Chr.6: 75237939-75237939 on Build GRCh38
rs946221460 Chr.6: 75237990-75237990 on Build GRCh38
rs964322439 Chr.6: 75237768-75237768 on Build GRCh38
rs536951693 Chr.6: 75237630-75237630 on Build GRCh38
rs768203385 Chr.6: 75237985-75237985 on Build GRCh38
rs752963398 Chr.6: 75237950-75237950 on Build GRCh38
rs768990704 Chr.6: 75237948-75237949 on Build GRCh38
rs769367850 Chr.6: 75238005-75238005 on Build GRCh38
rs773507804 Chr.6: 75237909-75237909 on Build GRCh38
rs774891152 Chr.6: 75238033-75238033 on Build GRCh38
rs371056263 Chr.6: 75237813-75237813 on Build GRCh38
rs557868927 Chr.6: 75237857-75237857 on Build GRCh38
rs1008315569 Chr.6: 75237695-75237695 on Build GRCh38
rs747669935 Chr.6: 75237996-75237996 on Build GRCh38
rs983533053 Chr.6: 75237806-75237806 on Build GRCh38
rs745791338 Chr.6: 75237852-75237852 on Build GRCh38
rs778159357 Chr.6: 75237965-75237965 on Build GRCh38
rs949165528 Chr.6: 75237940-75237940 on Build GRCh38

View Full Product Details