Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 75241031-75241543 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX7A2
Entrez Gene ID
1347
Gene Name
cytochrome c oxidase subunit 7A2
Gene Aliases
COX7AL, COX7AL1, COXVIIAL, COXVIIa-L, VIIAL
UniGene
Hs.70312
Gene Chromosome Location
Chr.6: 75237675-75243928 on Build GRCh38
Transcripts
NM_001865.3 NR_029466.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3875543 c.95A>G p.Q32R Chr.6: 75241189-75241189 on Build GRCh38
6634362 c.159C>T p.R53R Chr.6: 75241221-75241221 on Build GRCh38
4356921 c.193A>G p.K65E Chr.6: 75241187-75241187 on Build GRCh38
7094930 c.182C>T p.P61L Chr.6: 75241198-75241198 on Build GRCh38
484359 c.69T>G p.H23Q Chr.6: 75241215-75241215 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778383388 Chr.6: 75241279-75241279 on Build GRCh38
rs1029233995 Chr.6: 75241055-75241055 on Build GRCh38
rs1043425750 Chr.6: 75241345-75241347 on Build GRCh38
rs759725681 Chr.6: 75241447-75241447 on Build GRCh38
rs1017007193 Chr.6: 75241495-75241495 on Build GRCh38
rs748957075 Chr.6: 75241180-75241180 on Build GRCh38
rs918108441 Chr.6: 75241429-75241429 on Build GRCh38
rs760683912 Chr.6: 75241341-75241341 on Build GRCh38
rs751245744 Chr.6: 75241122-75241122 on Build GRCh38
rs189886747 Chr.6: 75241507-75241507 on Build GRCh38
rs1021461014 Chr.6: 75241311-75241311 on Build GRCh38
rs191388851 Chr.6: 75241292-75241292 on Build GRCh38
rs901541728 Chr.6: 75241375-75241375 on Build GRCh38
rs761956234 Chr.6: 75241233-75241233 on Build GRCh38
rs898782299 Chr.6: 75241468-75241468 on Build GRCh38
rs375148077 Chr.6: 75241222-75241222 on Build GRCh38
rs371766385 Chr.6: 75241198-75241198 on Build GRCh38
rs772589871 Chr.6: 75241199-75241199 on Build GRCh38
rs1025641691 Chr.6: 75241254-75241254 on Build GRCh38
rs1019473663 Chr.6: 75241288-75241288 on Build GRCh38
rs770435869 Chr.6: 75241189-75241189 on Build GRCh38
rs898501796 Chr.6: 75241247-75241247 on Build GRCh38
rs924331651 Chr.6: 75241098-75241098 on Build GRCh38
rs888916699 Chr.6: 75241471-75241471 on Build GRCh38
rs767341310 Chr.6: 75241253-75241253 on Build GRCh38
rs948228178 Chr.6: 75241297-75241297 on Build GRCh38
rs777215626 Chr.6: 75241167-75241167 on Build GRCh38
rs761995491 Chr.6: 75241103-75241103 on Build GRCh38
rs747388682 Chr.6: 75241310-75241310 on Build GRCh38
rs780618571 Chr.6: 75241309-75241309 on Build GRCh38
rs756828118 Chr.6: 75241273-75241273 on Build GRCh38
rs756102667 Chr.6: 75241259-75241259 on Build GRCh38
rs750678578 Chr.6: 75241240-75241240 on Build GRCh38
rs745568484 Chr.6: 75241280-75241280 on Build GRCh38
rs139079443 Chr.6: 75241239-75241239 on Build GRCh38
rs752682362 Chr.6: 75241134-75241134 on Build GRCh38
rs948179592 Chr.6: 75241445-75241445 on Build GRCh38
rs896631879 Chr.6: 75241224-75241224 on Build GRCh38
rs371921718 Chr.6: 75241127-75241127 on Build GRCh38
rs569467425 Chr.6: 75241075-75241075 on Build GRCh38
rs773407424 Chr.6: 75241319-75241319 on Build GRCh38
rs372449200 Chr.6: 75241223-75241223 on Build GRCh38
rs200671238 Chr.6: 75241218-75241218 on Build GRCh38
rs554941447 Chr.6: 75241354-75241354 on Build GRCh38
rs754039813 Chr.6: 75241431-75241431 on Build GRCh38
rs924259603 Chr.6: 75241342-75241342 on Build GRCh38
rs778637086 Chr.6: 75241197-75241197 on Build GRCh38
rs577033498 Chr.6: 75241446-75241446 on Build GRCh38
rs759302783 Chr.6: 75241243-75241243 on Build GRCh38
rs558588088 Chr.6: 75241372-75241372 on Build GRCh38
rs184713291 Chr.6: 75241351-75241351 on Build GRCh38
rs781057381 Chr.6: 75241129-75241129 on Build GRCh38
rs1006072137 Chr.6: 75241480-75241480 on Build GRCh38
rs992957010 Chr.6: 75241357-75241357 on Build GRCh38
rs755591432 Chr.6: 75241153-75241153 on Build GRCh38
rs753223498 Chr.6: 75241263-75241263 on Build GRCh38
rs765506648 Chr.6: 75241112-75241112 on Build GRCh38
rs752629299 Chr.6: 75241258-75241258 on Build GRCh38
rs1041453505 Chr.6: 75241086-75241086 on Build GRCh38

View Full Product Details