Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 78937591-78938131 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRAK1BP1
Entrez Gene ID
134728
Gene Name
interleukin 1 receptor associated kinase 1 binding protein 1
Gene Aliases
AIP70, SIMPL
UniGene
Hs.656212
Gene Chromosome Location
Chr.6: 78867486-78979411 on Build GRCh38
Transcripts
NM_017934.5

Gene Symbol
PHIP
Entrez Gene ID
55023
Gene Name
pleckstrin homology domain interacting protein
Gene Aliases
BRWD2, DCAF14, WDR11, ndrp
UniGene
Hs.511817
Gene Chromosome Location
Chr.6: 78934419-79078294 on Build GRCh38
Transcripts
NM_017934.5

SNP Detail

SNP ID Chromosome Location
rs764304290 Chr.6: 78937662-78937662 on Build GRCh38
rs201918575 Chr.6: 78937690-78937690 on Build GRCh38
rs1036848070 Chr.6: 78937887-78937887 on Build GRCh38
rs113074418 Chr.6: 78938002-78938002 on Build GRCh38
rs1004067638 Chr.6: 78937628-78937628 on Build GRCh38
rs5877635 Chr.6: 78937691-78937691 on Build GRCh38
rs200949532 Chr.6: 78937694-78937694 on Build GRCh38
rs542431905 Chr.6: 78937925-78937925 on Build GRCh38
rs768587581 Chr.6: 78937669-78937669 on Build GRCh38
rs1008279311 Chr.6: 78937698-78937698 on Build GRCh38
rs992253027 Chr.6: 78937979-78937979 on Build GRCh38
rs193256159 Chr.6: 78937727-78937727 on Build GRCh38
rs546692899 Chr.6: 78938071-78938071 on Build GRCh38
rs945275968 Chr.6: 78937758-78937771 on Build GRCh38
rs1056117047 Chr.6: 78937839-78937839 on Build GRCh38
rs951751729 Chr.6: 78937619-78937619 on Build GRCh38
rs1025195719 Chr.6: 78938066-78938067 on Build GRCh38
rs773466466 Chr.6: 78938012-78938012 on Build GRCh38
rs751010113 Chr.6: 78938021-78938021 on Build GRCh38
rs937687106 Chr.6: 78937803-78937803 on Build GRCh38
rs951581674 Chr.6: 78937979-78937987 on Build GRCh38
rs969591202 Chr.6: 78938080-78938080 on Build GRCh38
rs948065849 Chr.6: 78937656-78937656 on Build GRCh38
rs769487382 Chr.6: 78937964-78937964 on Build GRCh38
rs1008251506 Chr.6: 78937984-78937984 on Build GRCh38
rs553706950 Chr.6: 78937732-78937732 on Build GRCh38
rs397823390 Chr.6: 78937693-78937693 on Build GRCh38
rs995426853 Chr.6: 78937898-78937898 on Build GRCh38
rs1010761275 Chr.6: 78938016-78938016 on Build GRCh38
rs918130923 Chr.6: 78937654-78937654 on Build GRCh38
rs889753472 Chr.6: 78937978-78937978 on Build GRCh38
rs980765106 Chr.6: 78938081-78938081 on Build GRCh38
rs765811746 Chr.6: 78937818-78937818 on Build GRCh38
rs933107507 Chr.6: 78937741-78937741 on Build GRCh38
rs146682726 Chr.6: 78937869-78937869 on Build GRCh38
rs539506801 Chr.6: 78937786-78937786 on Build GRCh38
rs898399115 Chr.6: 78937648-78937648 on Build GRCh38
rs914756652 Chr.6: 78937764-78937764 on Build GRCh38
rs980801889 Chr.6: 78937792-78937792 on Build GRCh38
rs184598230 Chr.6: 78937959-78937959 on Build GRCh38
rs79535069 Chr.6: 78937695-78937695 on Build GRCh38
rs1007250361 Chr.6: 78937780-78937780 on Build GRCh38
rs72902825 Chr.6: 78937646-78937646 on Build GRCh38
rs546655599 Chr.6: 78937994-78937994 on Build GRCh38
rs556316541 Chr.6: 78937847-78937847 on Build GRCh38
rs527248271 Chr.6: 78937896-78937896 on Build GRCh38
rs143216558 Chr.6: 78937678-78937678 on Build GRCh38
rs956382973 Chr.6: 78937728-78937728 on Build GRCh38
rs761598777 Chr.6: 78937858-78937858 on Build GRCh38
rs570387014 Chr.6: 78937749-78937749 on Build GRCh38
rs926284565 Chr.6: 78937797-78937797 on Build GRCh38
rs76871224 Chr.6: 78937691-78937691 on Build GRCh38

View Full Product Details