Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 79919283-79919804 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELOVL4
Entrez Gene ID
6785
Gene Name
ELOVL fatty acid elongase 4
Gene Aliases
ADMD, CT118, ISQMR, SCA34, STGD2, STGD3
UniGene
Hs.101915
Gene Chromosome Location
Chr.6: 79914812-79947598 on Build GRCh38
Transcripts
NM_022726.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6266498 c.608C>A p.A203E Chr.6: 79919481-79919481 on Build GRCh38
122825 c.604A>C p.T202P Chr.6: 79919485-79919485 on Build GRCh38
6869952 c.573C>T p.I191I Chr.6: 79919516-79919516 on Build GRCh38
4404178 c.655A>C p.T219P Chr.6: 79919434-79919434 on Build GRCh38
5405872 c.602T>C p.L201S Chr.6: 79919487-79919487 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs115885157 Chr.6: 79919372-79919372 on Build GRCh38
rs140405366 Chr.6: 79919523-79919523 on Build GRCh38
rs760857419 Chr.6: 79919449-79919449 on Build GRCh38
rs921908722 Chr.6: 79919666-79919666 on Build GRCh38
rs776412370 Chr.6: 79919447-79919447 on Build GRCh38
rs1039271323 Chr.6: 79919631-79919631 on Build GRCh38
rs700482 Chr.6: 79919659-79919659 on Build GRCh38
rs753542573 Chr.6: 79919593-79919593 on Build GRCh38
rs141364884 Chr.6: 79919431-79919431 on Build GRCh38
rs1015899350 Chr.6: 79919532-79919532 on Build GRCh38
rs776979397 Chr.6: 79919472-79919472 on Build GRCh38
rs777542984 Chr.6: 79919426-79919426 on Build GRCh38
rs751499920 Chr.6: 79919395-79919395 on Build GRCh38
rs754848837 Chr.6: 79919397-79919397 on Build GRCh38
rs755560146 Chr.6: 79919417-79919417 on Build GRCh38
rs373929988 Chr.6: 79919440-79919440 on Build GRCh38
rs554644219 Chr.6: 79919511-79919511 on Build GRCh38
rs766079811 Chr.6: 79919374-79919374 on Build GRCh38
rs756811219 Chr.6: 79919595-79919595 on Build GRCh38
rs387906916 Chr.6: 79919443-79919443 on Build GRCh38
rs771331626 Chr.6: 79919435-79919435 on Build GRCh38
rs759399564 Chr.6: 79919582-79919583 on Build GRCh38
rs890776095 Chr.6: 79919492-79919492 on Build GRCh38
rs958179364 Chr.6: 79919330-79919330 on Build GRCh38
rs924154171 Chr.6: 79919779-79919779 on Build GRCh38
rs754420010 Chr.6: 79919347-79919347 on Build GRCh38
rs982175123 Chr.6: 79919428-79919428 on Build GRCh38
rs114247663 Chr.6: 79919362-79919362 on Build GRCh38
rs949339912 Chr.6: 79919735-79919735 on Build GRCh38
rs1053431559 Chr.6: 79919743-79919743 on Build GRCh38
rs199860277 Chr.6: 79919438-79919438 on Build GRCh38
rs752621101 Chr.6: 79919575-79919575 on Build GRCh38
rs1051466221 Chr.6: 79919691-79919691 on Build GRCh38
rs200665739 Chr.6: 79919399-79919399 on Build GRCh38
rs766171302 Chr.6: 79919502-79919502 on Build GRCh38
rs200457540 Chr.6: 79919370-79919370 on Build GRCh38
rs781245269 Chr.6: 79919546-79919546 on Build GRCh38
rs959478627 Chr.6: 79919632-79919632 on Build GRCh38
rs933213667 Chr.6: 79919684-79919684 on Build GRCh38
rs868460946 Chr.6: 79919510-79919510 on Build GRCh38
rs539166983 Chr.6: 79919341-79919341 on Build GRCh38
rs200118520 Chr.6: 79919471-79919471 on Build GRCh38
rs935102494 Chr.6: 79919707-79919707 on Build GRCh38
rs368830798 Chr.6: 79919598-79919598 on Build GRCh38
rs961246484 Chr.6: 79919545-79919545 on Build GRCh38
rs1026816779 Chr.6: 79919603-79919603 on Build GRCh38
rs747699698 Chr.6: 79919407-79919407 on Build GRCh38
rs371104774 Chr.6: 79919579-79919579 on Build GRCh38
rs776107074 Chr.6: 79919442-79919442 on Build GRCh38
rs767494464 Chr.6: 79919572-79919572 on Build GRCh38
rs1032488136 Chr.6: 79919310-79919310 on Build GRCh38
rs759498674 Chr.6: 79919564-79919564 on Build GRCh38
rs376577129 Chr.6: 79919485-79919485 on Build GRCh38
rs777084890 Chr.6: 79919589-79919589 on Build GRCh38
rs902157331 Chr.6: 79919692-79919692 on Build GRCh38

View Full Product Details